Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24268804-24269304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABGGTA
Entrez Gene ID
5875
Gene Name
Rab geranylgeranyltransferase alpha subunit
Gene Aliases
PTAR3
UniGene
Hs.377992
Gene Chromosome Location
Chr.14: 24265538-24271627 on Build GRCh38
Transcripts
NM_004581.5 NM_182836.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4719925 c.669delC p.N224fs*13 Chr.14: 24269126-24269126 on Build GRCh38
6761722 c.698G>A p.R233H Chr.14: 24269097-24269097 on Build GRCh38
552895 c.648G>C p.Q216H Chr.14: 24269147-24269147 on Build GRCh38
7248921 c.632-10A>G p.? Chr.14: 24269173-24269173 on Build GRCh38
332643 c.637G>C p.E213Q Chr.14: 24269158-24269158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949263702 Chr.14: 24269280-24269280 on Build GRCh38
rs909611762 Chr.14: 24269074-24269074 on Build GRCh38
rs758819720 Chr.14: 24269082-24269082 on Build GRCh38
rs940111239 Chr.14: 24269257-24269257 on Build GRCh38
rs555568550 Chr.14: 24269000-24269000 on Build GRCh38
rs750403065 Chr.14: 24268877-24268877 on Build GRCh38
rs776681034 Chr.14: 24268955-24268955 on Build GRCh38
rs371780561 Chr.14: 24269090-24269090 on Build GRCh38
rs767109692 Chr.14: 24268974-24268974 on Build GRCh38
rs984155198 Chr.14: 24269234-24269234 on Build GRCh38
rs748262293 Chr.14: 24269150-24269150 on Build GRCh38
rs767771354 Chr.14: 24268831-24268831 on Build GRCh38
rs888540606 Chr.14: 24268892-24268892 on Build GRCh38
rs779885726 Chr.14: 24268885-24268885 on Build GRCh38
rs375241144 Chr.14: 24268972-24268972 on Build GRCh38
rs1031958151 Chr.14: 24269083-24269083 on Build GRCh38
rs1131224 Chr.14: 24269096-24269096 on Build GRCh38
rs766253629 Chr.14: 24269032-24269032 on Build GRCh38
rs751369800 Chr.14: 24268886-24268886 on Build GRCh38
rs756742255 Chr.14: 24269065-24269065 on Build GRCh38
rs763769127 Chr.14: 24269060-24269060 on Build GRCh38
rs921517571 Chr.14: 24269219-24269219 on Build GRCh38
rs779317992 Chr.14: 24269079-24269079 on Build GRCh38
rs555890393 Chr.14: 24269066-24269066 on Build GRCh38
rs909883387 Chr.14: 24269243-24269243 on Build GRCh38
rs755893758 Chr.14: 24269050-24269050 on Build GRCh38
rs59227799 Chr.14: 24269173-24269173 on Build GRCh38
rs895872997 Chr.14: 24269279-24269279 on Build GRCh38
rs747290937 Chr.14: 24269021-24269021 on Build GRCh38
rs775697504 Chr.14: 24269007-24269007 on Build GRCh38
rs74037089 Chr.14: 24268999-24268999 on Build GRCh38
rs146912258 Chr.14: 24268987-24268987 on Build GRCh38
rs376558061 Chr.14: 24269185-24269185 on Build GRCh38
rs889801691 Chr.14: 24269230-24269230 on Build GRCh38
rs758313582 Chr.14: 24268883-24268883 on Build GRCh38
rs201557885 Chr.14: 24269178-24269178 on Build GRCh38
rs549174526 Chr.14: 24268899-24268899 on Build GRCh38
rs536913680 Chr.14: 24268949-24268949 on Build GRCh38
rs750976631 Chr.14: 24269081-24269081 on Build GRCh38
rs757844087 Chr.14: 24268997-24268997 on Build GRCh38
rs773140658 Chr.14: 24269166-24269166 on Build GRCh38
rs761827064 Chr.14: 24269030-24269030 on Build GRCh38
rs760686607 Chr.14: 24268869-24268869 on Build GRCh38
rs755647894 Chr.14: 24268918-24268918 on Build GRCh38
rs201499080 Chr.14: 24268921-24268921 on Build GRCh38
rs932885421 Chr.14: 24269224-24269224 on Build GRCh38
rs1131225 Chr.14: 24269084-24269084 on Build GRCh38
rs765178760 Chr.14: 24268872-24268872 on Build GRCh38
rs771895219 Chr.14: 24269177-24269177 on Build GRCh38
rs747307493 Chr.14: 24269089-24269089 on Build GRCh38
rs372223665 Chr.14: 24269040-24269040 on Build GRCh38
rs746176982 Chr.14: 24268935-24268935 on Build GRCh38
rs768862768 Chr.14: 24269025-24269025 on Build GRCh38
rs779417949 Chr.14: 24268998-24268998 on Build GRCh38
rs774307251 Chr.14: 24269039-24269039 on Build GRCh38
rs1037107692 Chr.14: 24269277-24269277 on Build GRCh38
rs774640470 Chr.14: 24268932-24268932 on Build GRCh38
rs976922838 Chr.14: 24269181-24269181 on Build GRCh38
rs775731561 Chr.14: 24268950-24268950 on Build GRCh38
rs752523823 Chr.14: 24269048-24269048 on Build GRCh38
rs762945140 Chr.14: 24268965-24268965 on Build GRCh38
rs367971962 Chr.14: 24268973-24268973 on Build GRCh38
rs538105820 Chr.14: 24269046-24269046 on Build GRCh38
rs200739130 Chr.14: 24269124-24269124 on Build GRCh38
rs754591709 Chr.14: 24268897-24268897 on Build GRCh38
rs780840337 Chr.14: 24268905-24268905 on Build GRCh38
rs567466887 Chr.14: 24268954-24268954 on Build GRCh38
rs142636626 Chr.14: 24268981-24268981 on Build GRCh38
rs749856381 Chr.14: 24268924-24268924 on Build GRCh38
rs368124688 Chr.14: 24268930-24268930 on Build GRCh38
rs139228254 Chr.14: 24269179-24269179 on Build GRCh38
rs772290687 Chr.14: 24268989-24268989 on Build GRCh38
rs752435704 Chr.14: 24268978-24268978 on Build GRCh38
rs760858063 Chr.14: 24268953-24268953 on Build GRCh38
rs752720389 Chr.14: 24268836-24268836 on Build GRCh38
rs760872739 Chr.14: 24269248-24269248 on Build GRCh38
rs191694051 Chr.14: 24268911-24268911 on Build GRCh38
rs769959957 Chr.14: 24269163-24269163 on Build GRCh38
rs1025326786 Chr.14: 24269008-24269008 on Build GRCh38

View Full Product Details