Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 46877201-46877720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MDGA2
Entrez Gene ID
161357
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 2
Gene Aliases
MAMDC1, c14_5286
UniGene
Hs.436380
Gene Chromosome Location
Chr.14: 46836518-47675319 on Build GRCh38
Transcripts
NM_001113498.2 NM_182830.4 XM_011536519.2 XM_011536525.2 XM_017021060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2197736 c.2431C>A p.H811N Chr.14: 46877495-46877495 on Build GRCh38
2197737 c.1537C>A p.H513N Chr.14: 46877495-46877495 on Build GRCh38
6075785 c.1527T>G p.P509P Chr.14: 46877505-46877505 on Build GRCh38
2197738 c.1537C>A p.H513N Chr.14: 46877495-46877495 on Build GRCh38
6075784 c.1527T>G p.P509P Chr.14: 46877505-46877505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772880359 Chr.14: 46877610-46877610 on Build GRCh38
rs762823551 Chr.14: 46877547-46877547 on Build GRCh38
rs1005298967 Chr.14: 46877274-46877274 on Build GRCh38
rs568070086 Chr.14: 46877444-46877447 on Build GRCh38
rs368113251 Chr.14: 46877286-46877286 on Build GRCh38
rs750338631 Chr.14: 46877542-46877542 on Build GRCh38
rs899879037 Chr.14: 46877682-46877682 on Build GRCh38
rs914983620 Chr.14: 46877416-46877416 on Build GRCh38
rs767919146 Chr.14: 46877675-46877675 on Build GRCh38
rs991063529 Chr.14: 46877695-46877695 on Build GRCh38
rs944365122 Chr.14: 46877435-46877435 on Build GRCh38
rs753410909 Chr.14: 46877547-46877547 on Build GRCh38
rs992303923 Chr.14: 46877389-46877389 on Build GRCh38
rs371342215 Chr.14: 46877665-46877665 on Build GRCh38
rs769333148 Chr.14: 46877607-46877607 on Build GRCh38
rs1015007231 Chr.14: 46877644-46877644 on Build GRCh38
rs760120150 Chr.14: 46877487-46877490 on Build GRCh38
rs996342896 Chr.14: 46877236-46877236 on Build GRCh38
rs766400973 Chr.14: 46877331-46877331 on Build GRCh38
rs1049192982 Chr.14: 46877576-46877576 on Build GRCh38
rs969038180 Chr.14: 46877407-46877407 on Build GRCh38
rs569371688 Chr.14: 46877289-46877289 on Build GRCh38
rs1002856056 Chr.14: 46877660-46877660 on Build GRCh38
rs776111306 Chr.14: 46877457-46877457 on Build GRCh38
rs763578745 Chr.14: 46877528-46877531 on Build GRCh38
rs45489507 Chr.14: 46877459-46877459 on Build GRCh38
rs1034953535 Chr.14: 46877689-46877689 on Build GRCh38
rs960500357 Chr.14: 46877376-46877376 on Build GRCh38
rs916270196 Chr.14: 46877390-46877393 on Build GRCh38
rs532779121 Chr.14: 46877252-46877252 on Build GRCh38
rs967905794 Chr.14: 46877339-46877339 on Build GRCh38
rs750545174 Chr.14: 46877462-46877462 on Build GRCh38
rs946449192 Chr.14: 46877424-46877424 on Build GRCh38
rs547842972 Chr.14: 46877351-46877351 on Build GRCh38
rs907230373 Chr.14: 46877648-46877648 on Build GRCh38
rs576724097 Chr.14: 46877512-46877512 on Build GRCh38
rs932157879 Chr.14: 46877570-46877570 on Build GRCh38
rs189307364 Chr.14: 46877319-46877319 on Build GRCh38
rs45554934 Chr.14: 46877358-46877358 on Build GRCh38
rs971383369 Chr.14: 46877305-46877305 on Build GRCh38
rs920180937 Chr.14: 46877432-46877432 on Build GRCh38
rs900732055 Chr.14: 46877470-46877470 on Build GRCh38
rs759680429 Chr.14: 46877469-46877469 on Build GRCh38
rs982058483 Chr.14: 46877317-46877317 on Build GRCh38
rs184430441 Chr.14: 46877255-46877255 on Build GRCh38
rs959290556 Chr.14: 46877690-46877690 on Build GRCh38
rs751405859 Chr.14: 46877557-46877557 on Build GRCh38
rs181217454 Chr.14: 46877431-46877431 on Build GRCh38
rs1040191142 Chr.14: 46877626-46877626 on Build GRCh38
rs766142058 Chr.14: 46877549-46877549 on Build GRCh38
rs755968210 Chr.14: 46877624-46877624 on Build GRCh38
rs999317081 Chr.14: 46877348-46877348 on Build GRCh38
rs536355419 Chr.14: 46877304-46877304 on Build GRCh38
rs923644121 Chr.14: 46877434-46877434 on Build GRCh38
rs533897917 Chr.14: 46877449-46877449 on Build GRCh38
rs912710548 Chr.14: 46877527-46877527 on Build GRCh38
rs899827032 Chr.14: 46877309-46877309 on Build GRCh38

View Full Product Details