Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49623242-49623779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAT2
Entrez Gene ID
4247
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase
Gene Aliases
CDG2A, CDGS2, GLCNACTII, GNT-II, GNT2
UniGene
Hs.93338
Gene Chromosome Location
Chr.14: 49620771-49623481 on Build GRCh38
Transcripts
NM_002408.3

SNP Detail

SNP ID Chromosome Location
rs770759290 Chr.14: 49623733-49623733 on Build GRCh38
rs919869818 Chr.14: 49623270-49623271 on Build GRCh38
rs145595837 Chr.14: 49623494-49623494 on Build GRCh38
rs1046955157 Chr.14: 49623271-49623273 on Build GRCh38
rs865835751 Chr.14: 49623467-49623467 on Build GRCh38
rs1040835491 Chr.14: 49623320-49623320 on Build GRCh38
rs960214333 Chr.14: 49623365-49623365 on Build GRCh38
rs1019321013 Chr.14: 49623669-49623669 on Build GRCh38
rs943741793 Chr.14: 49623272-49623272 on Build GRCh38
rs1055741281 Chr.14: 49623522-49623522 on Build GRCh38
rs34728941 Chr.14: 49623447-49623448 on Build GRCh38
rs964204813 Chr.14: 49623677-49623677 on Build GRCh38
rs1034676616 Chr.14: 49623286-49623286 on Build GRCh38
rs982709211 Chr.14: 49623468-49623468 on Build GRCh38
rs1053301389 Chr.14: 49623603-49623603 on Build GRCh38
rs868540573 Chr.14: 49623445-49623445 on Build GRCh38
rs546919134 Chr.14: 49623402-49623402 on Build GRCh38
rs975731573 Chr.14: 49623429-49623429 on Build GRCh38
rs886050523 Chr.14: 49623278-49623281 on Build GRCh38
rs908353293 Chr.14: 49623480-49623480 on Build GRCh38
rs907073234 Chr.14: 49623488-49623488 on Build GRCh38
rs573436485 Chr.14: 49623271-49623274 on Build GRCh38
rs772829752 Chr.14: 49623726-49623726 on Build GRCh38
rs992831653 Chr.14: 49623372-49623372 on Build GRCh38
rs1011470918 Chr.14: 49623557-49623557 on Build GRCh38
rs537612516 Chr.14: 49623381-49623381 on Build GRCh38
rs886050522 Chr.14: 49623281-49623281 on Build GRCh38
rs879604712 Chr.14: 49623271-49623271 on Build GRCh38
rs908576958 Chr.14: 49623745-49623745 on Build GRCh38
rs533078672 Chr.14: 49623687-49623687 on Build GRCh38
rs557292347 Chr.14: 49623403-49623403 on Build GRCh38
rs36055224 Chr.14: 49623441-49623441 on Build GRCh38
rs375306590 Chr.14: 49623512-49623512 on Build GRCh38
rs1031769834 Chr.14: 49623350-49623350 on Build GRCh38
rs535317346 Chr.14: 49623326-49623326 on Build GRCh38
rs949455346 Chr.14: 49623463-49623463 on Build GRCh38
rs2182078 Chr.14: 49623728-49623728 on Build GRCh38
rs756139772 Chr.14: 49623268-49623270 on Build GRCh38
rs558537786 Chr.14: 49623331-49623331 on Build GRCh38
rs920154273 Chr.14: 49623433-49623433 on Build GRCh38
rs571740661 Chr.14: 49623334-49623334 on Build GRCh38
rs574043827 Chr.14: 49623410-49623410 on Build GRCh38
rs540917754 Chr.14: 49623573-49623573 on Build GRCh38
rs200202235 Chr.14: 49623282-49623282 on Build GRCh38
rs769335073 Chr.14: 49623659-49623659 on Build GRCh38
rs563998184 Chr.14: 49623681-49623681 on Build GRCh38
rs895951770 Chr.14: 49623610-49623610 on Build GRCh38
rs572948832 Chr.14: 49623553-49623553 on Build GRCh38
rs780635680 Chr.14: 49623437-49623437 on Build GRCh38
rs747618664 Chr.14: 49623495-49623495 on Build GRCh38
rs999036686 Chr.14: 49623602-49623602 on Build GRCh38
rs369135991 Chr.14: 49623483-49623483 on Build GRCh38
rs966984804 Chr.14: 49623599-49623599 on Build GRCh38
rs970571302 Chr.14: 49623686-49623686 on Build GRCh38
rs758762548 Chr.14: 49623423-49623423 on Build GRCh38
rs931359725 Chr.14: 49623435-49623435 on Build GRCh38
rs986475641 Chr.14: 49623370-49623370 on Build GRCh38
rs957081723 Chr.14: 49623305-49623305 on Build GRCh38

View Full Product Details