Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51968788-51969323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG2
Entrez Gene ID
54331
Gene Name
G protein subunit gamma 2
Gene Aliases
-
UniGene
Hs.187772
Gene Chromosome Location
Chr.14: 51847131-51969800 on Build GRCh38
Transcripts
NM_001243773.1 NM_001243774.1 NM_053064.4 XM_006720173.2 XM_011536846.2 XM_017021376.1 XM_017021377.1

SNP Detail

SNP ID Chromosome Location
rs751595194 Chr.14: 51969202-51969202 on Build GRCh38
rs866794142 Chr.14: 51969275-51969275 on Build GRCh38
rs1001678487 Chr.14: 51968875-51968875 on Build GRCh38
rs2357306 Chr.14: 51969213-51969213 on Build GRCh38
rs879159701 Chr.14: 51968935-51968935 on Build GRCh38
rs1010805656 Chr.14: 51969270-51969270 on Build GRCh38
rs1012016616 Chr.14: 51969026-51969026 on Build GRCh38
rs964125460 Chr.14: 51969136-51969136 on Build GRCh38
rs45595035 Chr.14: 51969207-51969207 on Build GRCh38
rs1000535633 Chr.14: 51969283-51969283 on Build GRCh38
rs536291974 Chr.14: 51969166-51969166 on Build GRCh38
rs1048098702 Chr.14: 51968813-51968813 on Build GRCh38
rs1025589455 Chr.14: 51969061-51969061 on Build GRCh38
rs907771003 Chr.14: 51969124-51969124 on Build GRCh38
rs746978112 Chr.14: 51969131-51969132 on Build GRCh38
rs1010524683 Chr.14: 51968960-51968960 on Build GRCh38
rs936269410 Chr.14: 51969225-51969225 on Build GRCh38
rs1053928541 Chr.14: 51968974-51968974 on Build GRCh38
rs767520170 Chr.14: 51969055-51969055 on Build GRCh38
rs58238661 Chr.14: 51969207-51969208 on Build GRCh38
rs952634064 Chr.14: 51969067-51969067 on Build GRCh38
rs563143772 Chr.14: 51968876-51968876 on Build GRCh38
rs183790269 Chr.14: 51969083-51969083 on Build GRCh38
rs180852074 Chr.14: 51968865-51968865 on Build GRCh38
rs564578902 Chr.14: 51968821-51968821 on Build GRCh38
rs79707903 Chr.14: 51969215-51969215 on Build GRCh38
rs916073639 Chr.14: 51969257-51969257 on Build GRCh38
rs548903619 Chr.14: 51969004-51969004 on Build GRCh38
rs979063067 Chr.14: 51969017-51969017 on Build GRCh38
rs1032111701 Chr.14: 51969024-51969024 on Build GRCh38
rs895962539 Chr.14: 51968914-51968914 on Build GRCh38
rs529145469 Chr.14: 51968986-51968986 on Build GRCh38
rs896007143 Chr.14: 51969228-51969228 on Build GRCh38
rs76213020 Chr.14: 51969287-51969287 on Build GRCh38
rs567242050 Chr.14: 51969125-51969125 on Build GRCh38
rs904752357 Chr.14: 51969277-51969277 on Build GRCh38
rs143996731 Chr.14: 51969027-51969027 on Build GRCh38
rs766368393 Chr.14: 51969110-51969110 on Build GRCh38
rs1018536638 Chr.14: 51969120-51969120 on Build GRCh38
rs922604009 Chr.14: 51969203-51969203 on Build GRCh38
rs974580106 Chr.14: 51969041-51969041 on Build GRCh38
rs1020935199 Chr.14: 51968989-51968989 on Build GRCh38
rs1037644906 Chr.14: 51969193-51969193 on Build GRCh38
rs551110518 Chr.14: 51969090-51969090 on Build GRCh38
rs763153719 Chr.14: 51969059-51969059 on Build GRCh38
rs1057461742 Chr.14: 51969227-51969227 on Build GRCh38
rs368905627 Chr.14: 51968983-51968983 on Build GRCh38
rs1035825890 Chr.14: 51968890-51968890 on Build GRCh38
rs556056738 Chr.14: 51969297-51969297 on Build GRCh38
rs942017874 Chr.14: 51968858-51968858 on Build GRCh38
rs905930482 Chr.14: 51968873-51968873 on Build GRCh38
rs556793799 Chr.14: 51969208-51969208 on Build GRCh38
rs112698756 Chr.14: 51968851-51968851 on Build GRCh38
rs926109358 Chr.14: 51968878-51968878 on Build GRCh38
rs1005358820 Chr.14: 51969076-51969076 on Build GRCh38
rs964492599 Chr.14: 51968833-51968833 on Build GRCh38
rs966392918 Chr.14: 51968997-51968997 on Build GRCh38
rs933148127 Chr.14: 51969079-51969079 on Build GRCh38

View Full Product Details