Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51999244-51999783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf166
Entrez Gene ID
51637
Gene Name
chromosome 14 open reading frame 166
Gene Aliases
CGI-99, CGI99, CLE, CLE7, LCRP369, RLLM1, hCLE1
UniGene
Hs.534457
Gene Chromosome Location
Chr.14: 51989510-52004702 on Build GRCh38
Transcripts
NM_016039.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1238343 c.398G>A p.G133D Chr.14: 51999732-51999732 on Build GRCh38
1198075 c.433C>T p.R145C Chr.14: 51999767-51999767 on Build GRCh38
4051238 c.437A>G p.H146R Chr.14: 51999771-51999771 on Build GRCh38
4826205 c.430C>T p.Q144* Chr.14: 51999764-51999764 on Build GRCh38
5508412 c.426G>C p.Q142H Chr.14: 51999760-51999760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963306333 Chr.14: 51999289-51999289 on Build GRCh38
rs749836682 Chr.14: 51999664-51999664 on Build GRCh38
rs1034333622 Chr.14: 51999313-51999313 on Build GRCh38
rs778186301 Chr.14: 51999696-51999696 on Build GRCh38
rs1006105965 Chr.14: 51999540-51999540 on Build GRCh38
rs148658934 Chr.14: 51999322-51999322 on Build GRCh38
rs770993994 Chr.14: 51999704-51999704 on Build GRCh38
rs749625437 Chr.14: 51999286-51999286 on Build GRCh38
rs758026845 Chr.14: 51999290-51999290 on Build GRCh38
rs1030291188 Chr.14: 51999381-51999381 on Build GRCh38
rs954879860 Chr.14: 51999416-51999417 on Build GRCh38
rs916763282 Chr.14: 51999468-51999468 on Build GRCh38
rs766726865 Chr.14: 51999683-51999686 on Build GRCh38
rs899744844 Chr.14: 51999498-51999498 on Build GRCh38
rs978358460 Chr.14: 51999418-51999418 on Build GRCh38
rs1033970079 Chr.14: 51999587-51999587 on Build GRCh38
rs147854505 Chr.14: 51999481-51999481 on Build GRCh38
rs184446013 Chr.14: 51999444-51999444 on Build GRCh38
rs200455533 Chr.14: 51999747-51999747 on Build GRCh38
rs945738594 Chr.14: 51999350-51999350 on Build GRCh38
rs189158473 Chr.14: 51999645-51999645 on Build GRCh38
rs900333921 Chr.14: 51999580-51999580 on Build GRCh38
rs374231550 Chr.14: 51999752-51999752 on Build GRCh38
rs776056027 Chr.14: 51999520-51999520 on Build GRCh38
rs543467279 Chr.14: 51999532-51999532 on Build GRCh38
rs199645543 Chr.14: 51999753-51999753 on Build GRCh38
rs1016285877 Chr.14: 51999336-51999336 on Build GRCh38
rs1002754824 Chr.14: 51999269-51999269 on Build GRCh38
rs758423098 Chr.14: 51999667-51999667 on Build GRCh38
rs769728826 Chr.14: 51999604-51999604 on Build GRCh38
rs1037472115 Chr.14: 51999577-51999577 on Build GRCh38
rs934025121 Chr.14: 51999503-51999503 on Build GRCh38
rs778514716 Chr.14: 51999661-51999661 on Build GRCh38
rs768537485 Chr.14: 51999693-51999693 on Build GRCh38
rs749559241 Chr.14: 51999699-51999699 on Build GRCh38
rs748104817 Chr.14: 51999571-51999571 on Build GRCh38
rs1047637920 Chr.14: 51999527-51999527 on Build GRCh38
rs142152189 Chr.14: 51999348-51999348 on Build GRCh38
rs377541470 Chr.14: 51999676-51999676 on Build GRCh38
rs746863322 Chr.14: 51999688-51999688 on Build GRCh38
rs2486656 Chr.14: 51999630-51999630 on Build GRCh38
rs1039646441 Chr.14: 51999490-51999490 on Build GRCh38
rs193286496 Chr.14: 51999424-51999424 on Build GRCh38
rs188730245 Chr.14: 51999349-51999349 on Build GRCh38
rs147282814 Chr.14: 51999728-51999728 on Build GRCh38
rs975248929 Chr.14: 51999359-51999359 on Build GRCh38
rs996240000 Chr.14: 51999598-51999598 on Build GRCh38
rs1002771773 Chr.14: 51999584-51999584 on Build GRCh38
rs1030864968 Chr.14: 51999611-51999611 on Build GRCh38
rs1017539639 Chr.14: 51999561-51999561 on Build GRCh38
rs887202150 Chr.14: 51999529-51999529 on Build GRCh38
rs2486657 Chr.14: 51999597-51999597 on Build GRCh38
rs746584204 Chr.14: 51999455-51999455 on Build GRCh38
rs960945819 Chr.14: 51999614-51999614 on Build GRCh38

View Full Product Details