Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70375587-70376099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNJ2BP
Entrez Gene ID
55333
Gene Name
synaptojanin 2 binding protein
Gene Aliases
ARIP2, OMP25
UniGene
Hs.443661
Gene Chromosome Location
Chr.14: 70366496-70417090 on Build GRCh38
Transcripts
NM_018373.2 NM_001202547.2 NM_001202548.2 NM_001202549.2

Gene Symbol
SYNJ2BP-COX16
Entrez Gene ID
100529257
Gene Name
SYNJ2BP-COX16 readthrough
Gene Aliases
-
UniGene
Hs.443661
Gene Chromosome Location
Chr.14: 70325081-70417090 on Build GRCh38
Transcripts
NM_018373.2 NM_001202547.2 NM_001202548.2 NM_001202549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7237196 c.202-5G>C p.? Chr.14: 70375776-70375776 on Build GRCh38
6418925 c.281T>C p.L94P Chr.14: 70375692-70375692 on Build GRCh38
4577982 c.225C>T p.N75N Chr.14: 70375748-70375748 on Build GRCh38
6379665 c.296G>T p.R99M Chr.14: 70375677-70375677 on Build GRCh38
3983672 c.292C>T p.H98Y Chr.14: 70375681-70375681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893239204 Chr.14: 70375848-70375848 on Build GRCh38
rs563018873 Chr.14: 70375840-70375840 on Build GRCh38
rs747435031 Chr.14: 70375726-70375726 on Build GRCh38
rs548927945 Chr.14: 70375842-70375842 on Build GRCh38
rs973569429 Chr.14: 70375876-70375876 on Build GRCh38
rs766430181 Chr.14: 70375802-70375802 on Build GRCh38
rs755173926 Chr.14: 70375809-70375809 on Build GRCh38
rs373022406 Chr.14: 70375644-70375644 on Build GRCh38
rs758757890 Chr.14: 70375816-70375816 on Build GRCh38
rs2275298 Chr.14: 70376069-70376069 on Build GRCh38
rs186238353 Chr.14: 70376066-70376066 on Build GRCh38
rs766522950 Chr.14: 70375669-70375669 on Build GRCh38
rs748802147 Chr.14: 70375744-70375744 on Build GRCh38
rs866592819 Chr.14: 70375624-70375624 on Build GRCh38
rs867045635 Chr.14: 70375675-70375675 on Build GRCh38
rs943725384 Chr.14: 70375636-70375636 on Build GRCh38
rs867290363 Chr.14: 70375684-70375684 on Build GRCh38
rs934686811 Chr.14: 70375839-70375839 on Build GRCh38
rs201847524 Chr.14: 70375717-70375717 on Build GRCh38
rs950938879 Chr.14: 70375944-70375944 on Build GRCh38
rs201312428 Chr.14: 70375814-70375814 on Build GRCh38
rs375790401 Chr.14: 70375790-70375790 on Build GRCh38
rs551362145 Chr.14: 70375837-70375837 on Build GRCh38
rs975078819 Chr.14: 70376043-70376043 on Build GRCh38
rs778320534 Chr.14: 70375723-70375723 on Build GRCh38
rs773087785 Chr.14: 70375785-70375785 on Build GRCh38
rs750380155 Chr.14: 70375659-70375659 on Build GRCh38
rs758748035 Chr.14: 70375716-70375716 on Build GRCh38
rs539996023 Chr.14: 70375712-70375712 on Build GRCh38
rs199670786 Chr.14: 70375782-70375782 on Build GRCh38
rs765482225 Chr.14: 70375810-70375810 on Build GRCh38
rs761651562 Chr.14: 70375777-70375777 on Build GRCh38
rs190736425 Chr.14: 70376036-70376036 on Build GRCh38
rs771450964 Chr.14: 70375741-70375741 on Build GRCh38
rs779006718 Chr.14: 70375678-70375678 on Build GRCh38
rs759155314 Chr.14: 70375662-70375662 on Build GRCh38
rs753986708 Chr.14: 70375671-70375671 on Build GRCh38
rs180837721 Chr.14: 70376044-70376044 on Build GRCh38
rs530538098 Chr.14: 70375909-70375909 on Build GRCh38
rs564991561 Chr.14: 70375832-70375832 on Build GRCh38
rs755190221 Chr.14: 70375677-70375677 on Build GRCh38
rs532040595 Chr.14: 70375990-70375990 on Build GRCh38
rs751302594 Chr.14: 70375632-70375632 on Build GRCh38
rs567102151 Chr.14: 70376037-70376037 on Build GRCh38
rs773935958 Chr.14: 70375757-70375757 on Build GRCh38
rs756227921 Chr.14: 70375668-70375668 on Build GRCh38
rs541339851 Chr.14: 70375748-70375748 on Build GRCh38
rs753900240 Chr.14: 70375808-70375808 on Build GRCh38
rs936418609 Chr.14: 70376070-70376070 on Build GRCh38
rs2275297 Chr.14: 70376038-70376038 on Build GRCh38
rs774925382 Chr.14: 70375743-70375743 on Build GRCh38
rs943613135 Chr.14: 70375819-70375819 on Build GRCh38

View Full Product Details