Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73076145-73076679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM25
Entrez Gene ID
58517
Gene Name
RNA binding motif protein 25
Gene Aliases
NET52, RED120, RNPC7, S164, Snu71, fSAP94
UniGene
Hs.531106
Gene Chromosome Location
Chr.14: 73058436-73121675 on Build GRCh38
Transcripts
NM_021239.2 XM_011537044.2 XM_011537045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6042799 c.154A>G p.T52A Chr.14: 73076366-73076366 on Build GRCh38
5007060 c.153A>C p.P51P Chr.14: 73076365-73076365 on Build GRCh38
7076087 c.151C>A p.P51T Chr.14: 73076363-73076363 on Build GRCh38
6025497 c.156+10A>G p.? Chr.14: 73076378-73076378 on Build GRCh38
6763905 c.115A>C p.M39L Chr.14: 73076327-73076327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189552693 Chr.14: 73076402-73076402 on Build GRCh38
rs772639104 Chr.14: 73076388-73076388 on Build GRCh38
rs946579023 Chr.14: 73076205-73076205 on Build GRCh38
rs950393746 Chr.14: 73076393-73076393 on Build GRCh38
rs569337113 Chr.14: 73076494-73076494 on Build GRCh38
rs899432320 Chr.14: 73076287-73076287 on Build GRCh38
rs775116869 Chr.14: 73076413-73076413 on Build GRCh38
rs775215816 Chr.14: 73076474-73076481 on Build GRCh38
rs377459249 Chr.14: 73076286-73076286 on Build GRCh38
rs371002071 Chr.14: 73076386-73076386 on Build GRCh38
rs749503437 Chr.14: 73076276-73076276 on Build GRCh38
rs766689839 Chr.14: 73076282-73076283 on Build GRCh38
rs554558335 Chr.14: 73076627-73076627 on Build GRCh38
rs755172876 Chr.14: 73076375-73076375 on Build GRCh38
rs764027495 Chr.14: 73076327-73076327 on Build GRCh38
rs1019289183 Chr.14: 73076457-73076457 on Build GRCh38
rs375783115 Chr.14: 73076272-73076272 on Build GRCh38
rs746964908 Chr.14: 73076183-73076183 on Build GRCh38
rs769079498 Chr.14: 73076280-73076280 on Build GRCh38
rs976285788 Chr.14: 73076487-73076487 on Build GRCh38
rs548213472 Chr.14: 73076239-73076239 on Build GRCh38
rs528812636 Chr.14: 73076293-73076293 on Build GRCh38
rs750525810 Chr.14: 73076361-73076361 on Build GRCh38
rs184988981 Chr.14: 73076180-73076180 on Build GRCh38
rs924630226 Chr.14: 73076622-73076622 on Build GRCh38
rs779274391 Chr.14: 73076385-73076385 on Build GRCh38
rs7144160 Chr.14: 73076609-73076609 on Build GRCh38
rs915046607 Chr.14: 73076592-73076592 on Build GRCh38
rs1043087248 Chr.14: 73076214-73076214 on Build GRCh38
rs377399576 Chr.14: 73076365-73076365 on Build GRCh38
rs780181976 Chr.14: 73076366-73076366 on Build GRCh38
rs762661994 Chr.14: 73076323-73076323 on Build GRCh38
rs539112103 Chr.14: 73076476-73076476 on Build GRCh38
rs759350002 Chr.14: 73076537-73076537 on Build GRCh38
rs780581598 Chr.14: 73076224-73076224 on Build GRCh38
rs1042225510 Chr.14: 73076616-73076616 on Build GRCh38
rs946168316 Chr.14: 73076613-73076613 on Build GRCh38
rs769594718 Chr.14: 73076576-73076576 on Build GRCh38
rs967971145 Chr.14: 73076608-73076608 on Build GRCh38
rs767287584 Chr.14: 73076348-73076348 on Build GRCh38
rs751348397 Chr.14: 73076338-73076338 on Build GRCh38
rs890651239 Chr.14: 73076245-73076245 on Build GRCh38
rs1007678036 Chr.14: 73076263-73076263 on Build GRCh38
rs757288374 Chr.14: 73076347-73076347 on Build GRCh38
rs7144753 Chr.14: 73076653-73076653 on Build GRCh38
rs372939628 Chr.14: 73076489-73076489 on Build GRCh38
rs776631790 Chr.14: 73076387-73076387 on Build GRCh38
rs770976488 Chr.14: 73076216-73076216 on Build GRCh38
rs1052319800 Chr.14: 73076639-73076639 on Build GRCh38
rs959508694 Chr.14: 73076546-73076546 on Build GRCh38
rs376086326 Chr.14: 73076431-73076431 on Build GRCh38
rs772411821 Chr.14: 73076288-73076288 on Build GRCh38
rs769225976 Chr.14: 73076412-73076412 on Build GRCh38
rs369533198 Chr.14: 73076202-73076202 on Build GRCh38
rs151207104 Chr.14: 73076282-73076282 on Build GRCh38
rs868465502 Chr.14: 73076425-73076425 on Build GRCh38
rs990771686 Chr.14: 73076182-73076182 on Build GRCh38
rs995375383 Chr.14: 73076301-73076301 on Build GRCh38
rs572785580 Chr.14: 73076652-73076652 on Build GRCh38
rs1034769225 Chr.14: 73076502-73076502 on Build GRCh38
rs934666545 Chr.14: 73076630-73076630 on Build GRCh38
rs775348644 Chr.14: 73076295-73076295 on Build GRCh38
rs758904508 Chr.14: 73076289-73076289 on Build GRCh38
rs1051839940 Chr.14: 73076243-73076243 on Build GRCh38

View Full Product Details