Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 81092391-81092823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSHR
Entrez Gene ID
7253
Gene Name
thyroid stimulating hormone receptor
Gene Aliases
CHNG1, LGR3, hTSHR-I
UniGene
Hs.160411
Gene Chromosome Location
Chr.14: 80954989-81146302 on Build GRCh38
Transcripts
NM_000369.2 NM_001018036.2 NM_001142626.2 XM_005268037.4 XM_005268039.1 XM_006720245.1 XM_011537119.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5382757 c.523C>T p.L175L Chr.14: 81092586-81092586 on Build GRCh38
4499540 c.544C>T p.L182L Chr.14: 81092607-81092607 on Build GRCh38
4496917 c.485C>T p.P162L Chr.14: 81092548-81092548 on Build GRCh38
4496916 c.485C>T p.P162L Chr.14: 81092548-81092548 on Build GRCh38
4499539 c.544C>T p.L182L Chr.14: 81092607-81092607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368516194 Chr.14: 81092643-81092643 on Build GRCh38
rs112294119 Chr.14: 81092598-81092598 on Build GRCh38
rs753484037 Chr.14: 81092660-81092660 on Build GRCh38
rs892599973 Chr.14: 81092472-81092472 on Build GRCh38
rs371344604 Chr.14: 81092604-81092604 on Build GRCh38
rs535032148 Chr.14: 81092642-81092642 on Build GRCh38
rs954484908 Chr.14: 81092559-81092559 on Build GRCh38
rs779349574 Chr.14: 81092548-81092548 on Build GRCh38
rs557239689 Chr.14: 81092753-81092753 on Build GRCh38
rs786205080 Chr.14: 81092611-81092611 on Build GRCh38
rs759772167 Chr.14: 81092503-81092506 on Build GRCh38
rs775278924 Chr.14: 81092666-81092666 on Build GRCh38
rs906647918 Chr.14: 81092459-81092459 on Build GRCh38
rs879733352 Chr.14: 81092740-81092740 on Build GRCh38
rs773942819 Chr.14: 81092463-81092463 on Build GRCh38
rs762693317 Chr.14: 81092497-81092497 on Build GRCh38
rs775675381 Chr.14: 81092590-81092591 on Build GRCh38
rs375265817 Chr.14: 81092701-81092701 on Build GRCh38
rs907334681 Chr.14: 81092756-81092756 on Build GRCh38
rs962149688 Chr.14: 81092656-81092656 on Build GRCh38
rs776859401 Chr.14: 81092494-81092494 on Build GRCh38
rs968428041 Chr.14: 81092531-81092531 on Build GRCh38
rs112584148 Chr.14: 81092692-81092692 on Build GRCh38
rs570156274 Chr.14: 81092492-81092492 on Build GRCh38
rs751390919 Chr.14: 81092514-81092514 on Build GRCh38
rs748528469 Chr.14: 81092553-81092553 on Build GRCh38
rs771027875 Chr.14: 81092570-81092570 on Build GRCh38
rs779806385 Chr.14: 81092529-81092529 on Build GRCh38
rs121908863 Chr.14: 81092547-81092547 on Build GRCh38
rs767424621 Chr.14: 81092519-81092519 on Build GRCh38
rs548570734 Chr.14: 81092483-81092483 on Build GRCh38
rs2241119 Chr.14: 81092621-81092621 on Build GRCh38
rs749925301 Chr.14: 81092522-81092522 on Build GRCh38
rs1011160460 Chr.14: 81092482-81092482 on Build GRCh38
rs773757026 Chr.14: 81092558-81092558 on Build GRCh38
rs748980987 Chr.14: 81092543-81092543 on Build GRCh38
rs767470807 Chr.14: 81092632-81092632 on Build GRCh38
rs757200998 Chr.14: 81092516-81092516 on Build GRCh38
rs774195333 Chr.14: 81092617-81092617 on Build GRCh38
rs786204790 Chr.14: 81092610-81092611 on Build GRCh38
rs1045057332 Chr.14: 81092443-81092443 on Build GRCh38
rs189484098 Chr.14: 81092748-81092748 on Build GRCh38
rs371708118 Chr.14: 81092528-81092528 on Build GRCh38
rs772600260 Chr.14: 81092557-81092557 on Build GRCh38
rs767877585 Chr.14: 81092515-81092516 on Build GRCh38
rs2075176 Chr.14: 81092462-81092462 on Build GRCh38
rs763951223 Chr.14: 81092503-81092503 on Build GRCh38
rs765994426 Chr.14: 81092649-81092649 on Build GRCh38
rs760589113 Chr.14: 81092797-81092797 on Build GRCh38
rs749779084 Chr.14: 81092420-81092420 on Build GRCh38
rs559831405 Chr.14: 81092417-81092417 on Build GRCh38
rs765431474 Chr.14: 81092612-81092612 on Build GRCh38
rs747039783 Chr.14: 81092564-81092564 on Build GRCh38
rs121908862 Chr.14: 81092563-81092563 on Build GRCh38

View Full Product Details