Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92938363-92938871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPK1
Entrez Gene ID
3705
Gene Name
inositol-tetrakisphosphate 1-kinase
Gene Aliases
ITRPK1
UniGene
Hs.308122
Gene Chromosome Location
Chr.14: 92936914-93115918 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6015604 c.925G>A p.E309K Chr.14: 92938503-92938503 on Build GRCh38
1128169 c.917G>A p.C306Y Chr.14: 92938511-92938511 on Build GRCh38
5437032 c.907_908insT p.C303fs*>13 Chr.14: 92938520-92938521 on Build GRCh38
4519043 c.928_929GG>AA p.G310N Chr.14: 92938499-92938500 on Build GRCh38
6898820 c.938C>T p.T313I Chr.14: 92938490-92938490 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1024786933 Chr.14: 92938400-92938400 on Build GRCh38
rs532646415 Chr.14: 92938654-92938654 on Build GRCh38
rs903304634 Chr.14: 92938741-92938741 on Build GRCh38
rs1042204460 Chr.14: 92938734-92938734 on Build GRCh38
rs568379499 Chr.14: 92938744-92938744 on Build GRCh38
rs747889414 Chr.14: 92938489-92938489 on Build GRCh38
rs910119196 Chr.14: 92938653-92938653 on Build GRCh38
rs772886151 Chr.14: 92938524-92938524 on Build GRCh38
rs754605112 Chr.14: 92938484-92938484 on Build GRCh38
rs530298031 Chr.14: 92938478-92938478 on Build GRCh38
rs771683591 Chr.14: 92938513-92938513 on Build GRCh38
rs8006516 Chr.14: 92938392-92938392 on Build GRCh38
rs935756226 Chr.14: 92938670-92938670 on Build GRCh38
rs749062305 Chr.14: 92938479-92938479 on Build GRCh38
rs964009392 Chr.14: 92938785-92938785 on Build GRCh38
rs964062063 Chr.14: 92938616-92938616 on Build GRCh38
rs548586984 Chr.14: 92938729-92938729 on Build GRCh38
rs201006858 Chr.14: 92938553-92938553 on Build GRCh38
rs1018369543 Chr.14: 92938587-92938587 on Build GRCh38
rs8011546 Chr.14: 92938765-92938765 on Build GRCh38
rs193239474 Chr.14: 92938767-92938767 on Build GRCh38
rs541404317 Chr.14: 92938398-92938398 on Build GRCh38
rs577005835 Chr.14: 92938794-92938794 on Build GRCh38
rs371296846 Chr.14: 92938605-92938605 on Build GRCh38
rs763211487 Chr.14: 92938544-92938544 on Build GRCh38
rs764129225 Chr.14: 92938560-92938560 on Build GRCh38
rs373374574 Chr.14: 92938575-92938575 on Build GRCh38
rs376686350 Chr.14: 92938591-92938591 on Build GRCh38
rs777888345 Chr.14: 92938459-92938459 on Build GRCh38
rs575491658 Chr.14: 92938702-92938702 on Build GRCh38
rs552428984 Chr.14: 92938678-92938678 on Build GRCh38
rs1055461825 Chr.14: 92938483-92938483 on Build GRCh38
rs888643098 Chr.14: 92938725-92938725 on Build GRCh38
rs952825829 Chr.14: 92938583-92938583 on Build GRCh38
rs1012435695 Chr.14: 92938496-92938496 on Build GRCh38
rs776995918 Chr.14: 92938536-92938536 on Build GRCh38
rs370363215 Chr.14: 92938531-92938531 on Build GRCh38
rs945826906 Chr.14: 92938645-92938645 on Build GRCh38
rs893697403 Chr.14: 92938770-92938770 on Build GRCh38
rs572798534 Chr.14: 92938393-92938393 on Build GRCh38
rs143650223 Chr.14: 92938685-92938685 on Build GRCh38
rs1017898982 Chr.14: 92938827-92938827 on Build GRCh38
rs756000302 Chr.14: 92938576-92938576 on Build GRCh38
rs375961844 Chr.14: 92938488-92938488 on Build GRCh38
rs563496820 Chr.14: 92938640-92938640 on Build GRCh38
rs1041553497 Chr.14: 92938650-92938650 on Build GRCh38
rs775562584 Chr.14: 92938542-92938542 on Build GRCh38
rs772730022 Chr.14: 92938642-92938642 on Build GRCh38
rs758469484 Chr.14: 92938434-92938434 on Build GRCh38
rs750339707 Chr.14: 92938466-92938466 on Build GRCh38
rs976910025 Chr.14: 92938598-92938598 on Build GRCh38
rs1025199109 Chr.14: 92938506-92938506 on Build GRCh38
rs994868294 Chr.14: 92938707-92938707 on Build GRCh38
rs747476670 Chr.14: 92938526-92938526 on Build GRCh38
rs906798748 Chr.14: 92938706-92938706 on Build GRCh38
rs534734435 Chr.14: 92938694-92938694 on Build GRCh38
rs112697872 Chr.14: 92938509-92938509 on Build GRCh38
rs202000533 Chr.14: 92938474-92938474 on Build GRCh38
rs760457640 Chr.14: 92938693-92938693 on Build GRCh38
rs1029545173 Chr.14: 92938815-92938815 on Build GRCh38
rs1025805666 Chr.14: 92938709-92938709 on Build GRCh38
rs765699512 Chr.14: 92938537-92938537 on Build GRCh38

View Full Product Details