Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94563704-94564251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINA4
Entrez Gene ID
5267
Gene Name
serpin family A member 4
Gene Aliases
KAL, KLST, KST, PI-4, PI4, kallistatin
UniGene
Hs.719893
Gene Chromosome Location
Chr.14: 94561420-94569913 on Build GRCh38
Transcripts
NM_001289032.1 NM_001289033.1 NM_006215.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3499385 c.344C>T p.S115F Chr.14: 94563826-94563826 on Build GRCh38
7174966 c.400G>A p.G134R Chr.14: 94563882-94563882 on Build GRCh38
3499383 c.304C>G p.L102V Chr.14: 94563786-94563786 on Build GRCh38
7099895 c.561G>C p.K187N Chr.14: 94564043-94564043 on Build GRCh38
4800351 c.440A>G p.H147R Chr.14: 94563922-94563922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753066101 Chr.14: 94563773-94563773 on Build GRCh38
rs750402462 Chr.14: 94563860-94563860 on Build GRCh38
rs766561113 Chr.14: 94564038-94564038 on Build GRCh38
rs539993882 Chr.14: 94563897-94563897 on Build GRCh38
rs1012276175 Chr.14: 94564134-94564134 on Build GRCh38
rs771729669 Chr.14: 94563869-94563869 on Build GRCh38
rs754451695 Chr.14: 94564138-94564138 on Build GRCh38
rs71431647 Chr.14: 94564177-94564177 on Build GRCh38
rs767203821 Chr.14: 94564099-94564099 on Build GRCh38
rs371042106 Chr.14: 94564094-94564094 on Build GRCh38
rs757285900 Chr.14: 94563785-94563785 on Build GRCh38
rs144981852 Chr.14: 94563767-94563767 on Build GRCh38
rs757524773 Chr.14: 94563849-94563849 on Build GRCh38
rs1030794902 Chr.14: 94564178-94564178 on Build GRCh38
rs369378451 Chr.14: 94563964-94563964 on Build GRCh38
rs781081656 Chr.14: 94564010-94564010 on Build GRCh38
rs766351953 Chr.14: 94563979-94563979 on Build GRCh38
rs201586458 Chr.14: 94564105-94564105 on Build GRCh38
rs755874428 Chr.14: 94564108-94564108 on Build GRCh38
rs139198134 Chr.14: 94564031-94564031 on Build GRCh38
rs751750120 Chr.14: 94563845-94563845 on Build GRCh38
rs373176783 Chr.14: 94563966-94563966 on Build GRCh38
rs765961793 Chr.14: 94563821-94563821 on Build GRCh38
rs748691639 Chr.14: 94564032-94564032 on Build GRCh38
rs770778133 Chr.14: 94563873-94563873 on Build GRCh38
rs759642550 Chr.14: 94564056-94564056 on Build GRCh38
rs769083828 Chr.14: 94563954-94563954 on Build GRCh38
rs149939871 Chr.14: 94563759-94563759 on Build GRCh38
rs748638415 Chr.14: 94564152-94564152 on Build GRCh38
rs559001487 Chr.14: 94563780-94563780 on Build GRCh38
rs777710811 Chr.14: 94563908-94563908 on Build GRCh38
rs776734977 Chr.14: 94564053-94564053 on Build GRCh38
rs745486760 Chr.14: 94564011-94564011 on Build GRCh38
rs757079859 Chr.14: 94564006-94564006 on Build GRCh38
rs767668026 Chr.14: 94563896-94563896 on Build GRCh38
rs760440214 Chr.14: 94563818-94563818 on Build GRCh38
rs368977287 Chr.14: 94563868-94563868 on Build GRCh38
rs993683508 Chr.14: 94563895-94563895 on Build GRCh38
rs763557475 Chr.14: 94563886-94563886 on Build GRCh38
rs758464001 Chr.14: 94563901-94563901 on Build GRCh38
rs575149680 Chr.14: 94563836-94563836 on Build GRCh38
rs886611815 Chr.14: 94564078-94564078 on Build GRCh38
rs755118352 Chr.14: 94563990-94563990 on Build GRCh38
rs747841538 Chr.14: 94563813-94563813 on Build GRCh38
rs776431296 Chr.14: 94563875-94563875 on Build GRCh38
rs2144077 Chr.14: 94563929-94563929 on Build GRCh38
rs758792521 Chr.14: 94563774-94563774 on Build GRCh38
rs199872967 Chr.14: 94563740-94563740 on Build GRCh38
rs151234180 Chr.14: 94563730-94563730 on Build GRCh38
rs200620229 Chr.14: 94563835-94563835 on Build GRCh38
rs759880155 Chr.14: 94564181-94564181 on Build GRCh38
rs776537140 Chr.14: 94564099-94564101 on Build GRCh38
rs749254647 Chr.14: 94563729-94563729 on Build GRCh38
rs757314357 Chr.14: 94564097-94564097 on Build GRCh38
rs746738510 Chr.14: 94563919-94563919 on Build GRCh38
rs745567923 Chr.14: 94563936-94563936 on Build GRCh38
rs765599229 Chr.14: 94563830-94563830 on Build GRCh38
rs376441816 Chr.14: 94563876-94563876 on Build GRCh38
rs111617033 Chr.14: 94564019-94564019 on Build GRCh38
rs773592808 Chr.14: 94564162-94564162 on Build GRCh38
rs772299677 Chr.14: 94564033-94564033 on Build GRCh38
rs771926277 Chr.14: 94563816-94563816 on Build GRCh38
rs776342643 Chr.14: 94563827-94563827 on Build GRCh38
rs765441266 Chr.14: 94564081-94564081 on Build GRCh38
rs148176642 Chr.14: 94563959-94563959 on Build GRCh38
rs141982763 Chr.14: 94563965-94563965 on Build GRCh38
rs550133117 Chr.14: 94564002-94564002 on Build GRCh38
rs1045499232 Chr.14: 94564171-94564171 on Build GRCh38
rs542522149 Chr.14: 94563852-94563852 on Build GRCh38
rs772962250 Chr.14: 94563817-94563817 on Build GRCh38
rs957301897 Chr.14: 94564095-94564095 on Build GRCh38
rs548077223 Chr.14: 94564142-94564142 on Build GRCh38
rs768175678 Chr.14: 94564023-94564023 on Build GRCh38
rs982432473 Chr.14: 94563758-94563758 on Build GRCh38
rs900677407 Chr.14: 94563846-94563846 on Build GRCh38
rs749209997 Chr.14: 94563799-94563799 on Build GRCh38
rs1030301546 Chr.14: 94563992-94563992 on Build GRCh38
rs1005567243 Chr.14: 94563870-94563870 on Build GRCh38
rs5509 Chr.14: 94563893-94563893 on Build GRCh38
rs934976944 Chr.14: 94563797-94563797 on Build GRCh38
rs373453583 Chr.14: 94563872-94563872 on Build GRCh38
rs1047416630 Chr.14: 94564188-94564188 on Build GRCh38
rs754012662 Chr.14: 94563988-94563988 on Build GRCh38
rs376130328 Chr.14: 94564034-94564034 on Build GRCh38
rs915785406 Chr.14: 94564136-94564136 on Build GRCh38
rs888023939 Chr.14: 94564223-94564223 on Build GRCh38
rs899161392 Chr.14: 94564125-94564125 on Build GRCh38
rs201138033 Chr.14: 94563775-94563775 on Build GRCh38
rs761003717 Chr.14: 94563968-94563968 on Build GRCh38
rs200334312 Chr.14: 94563733-94563733 on Build GRCh38
rs769156344 Chr.14: 94563940-94563940 on Build GRCh38
rs764068417 Chr.14: 94564001-94564001 on Build GRCh38
rs748969602 Chr.14: 94564119-94564119 on Build GRCh38
rs538199214 Chr.14: 94563828-94563828 on Build GRCh38
rs143645553 Chr.14: 94563900-94563900 on Build GRCh38
rs942339548 Chr.14: 94564224-94564224 on Build GRCh38
rs764474650 Chr.14: 94563841-94563841 on Build GRCh38
rs146511646 Chr.14: 94564167-94564167 on Build GRCh38
rs140377924 Chr.14: 94563753-94563753 on Build GRCh38
rs564953147 Chr.14: 94563925-94563925 on Build GRCh38
rs200475097 Chr.14: 94564112-94564112 on Build GRCh38
rs755854273 Chr.14: 94564014-94564014 on Build GRCh38
rs775822256 Chr.14: 94563882-94563882 on Build GRCh38
rs141308668 Chr.14: 94563812-94563812 on Build GRCh38
rs768543200 Chr.14: 94563806-94563806 on Build GRCh38
rs1006790360 Chr.14: 94563911-94563911 on Build GRCh38
rs755992878 Chr.14: 94563879-94563879 on Build GRCh38
rs768548078 Chr.14: 94564082-94564083 on Build GRCh38
rs758574523 Chr.14: 94564037-94564037 on Build GRCh38
rs372759493 Chr.14: 94563953-94563953 on Build GRCh38
rs756230378 Chr.14: 94563861-94563861 on Build GRCh38
rs74763941 Chr.14: 94564173-94564173 on Build GRCh38
rs368457311 Chr.14: 94563792-94563792 on Build GRCh38
rs533950451 Chr.14: 94564172-94564172 on Build GRCh38
rs760217189 Chr.14: 94563741-94563741 on Build GRCh38
rs13306798 Chr.14: 94564157-94564157 on Build GRCh38
rs747344836 Chr.14: 94564166-94564166 on Build GRCh38
rs1010822223 Chr.14: 94564212-94564212 on Build GRCh38
rs775115901 Chr.14: 94563947-94563947 on Build GRCh38
rs767013382 Chr.14: 94563734-94563734 on Build GRCh38
rs940916193 Chr.14: 94564091-94564091 on Build GRCh38
rs778947204 Chr.14: 94563791-94563791 on Build GRCh38
rs145028164 Chr.14: 94563892-94563892 on Build GRCh38
rs374913307 Chr.14: 94563762-94563762 on Build GRCh38
rs775357027 Chr.14: 94563728-94563728 on Build GRCh38
rs140563422 Chr.14: 94563894-94563894 on Build GRCh38
rs748736850 Chr.14: 94564020-94564020 on Build GRCh38
rs747013149 Chr.14: 94563910-94563911 on Build GRCh38

View Full Product Details