Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102943801-102944301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPB
Entrez Gene ID
9578
Gene Name
CDC42 binding protein kinase beta
Gene Aliases
MRCKB
UniGene
Hs.654634
Gene Chromosome Location
Chr.14: 102932379-103057405 on Build GRCh38
Transcripts
NM_006035.3 XM_005268227.1 XM_005268228.1 XM_005268229.1 XM_005268230.4 XM_011537387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4049228 c.4042G>A p.V1348M Chr.14: 102944257-102944257 on Build GRCh38
3730534 c.4371G>A p.A1457A Chr.14: 102943928-102943928 on Build GRCh38
4889997 c.4364C>A p.A1455E Chr.14: 102943935-102943935 on Build GRCh38
4454082 c.4005A>C p.T1335T Chr.14: 102944294-102944294 on Build GRCh38
6280118 c.4139C>T p.A1380V Chr.14: 102944160-102944160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746411194 Chr.14: 102944066-102944066 on Build GRCh38
rs201500787 Chr.14: 102944270-102944270 on Build GRCh38
rs772682992 Chr.14: 102944039-102944039 on Build GRCh38
rs548093157 Chr.14: 102944092-102944092 on Build GRCh38
rs769143847 Chr.14: 102943831-102943831 on Build GRCh38
rs149319188 Chr.14: 102944164-102944164 on Build GRCh38
rs750575554 Chr.14: 102944081-102944081 on Build GRCh38
rs777304874 Chr.14: 102944130-102944130 on Build GRCh38
rs781495376 Chr.14: 102943982-102943982 on Build GRCh38
rs749803255 Chr.14: 102944021-102944021 on Build GRCh38
rs746522635 Chr.14: 102944257-102944257 on Build GRCh38
rs371972643 Chr.14: 102944131-102944131 on Build GRCh38
rs750198864 Chr.14: 102943853-102943853 on Build GRCh38
rs367693621 Chr.14: 102944036-102944036 on Build GRCh38
rs755548130 Chr.14: 102944107-102944107 on Build GRCh38
rs889223871 Chr.14: 102943907-102943907 on Build GRCh38
rs757617177 Chr.14: 102943973-102943973 on Build GRCh38
rs376515979 Chr.14: 102943842-102943842 on Build GRCh38
rs141912208 Chr.14: 102944088-102944088 on Build GRCh38
rs780645988 Chr.14: 102944006-102944006 on Build GRCh38
rs199903511 Chr.14: 102944196-102944196 on Build GRCh38
rs528455993 Chr.14: 102944114-102944114 on Build GRCh38
rs888287367 Chr.14: 102943836-102943836 on Build GRCh38
rs35828352 Chr.14: 102944258-102944258 on Build GRCh38
rs759334009 Chr.14: 102944160-102944160 on Build GRCh38
rs753163105 Chr.14: 102944243-102944243 on Build GRCh38
rs775515879 Chr.14: 102944174-102944174 on Build GRCh38
rs779327699 Chr.14: 102944113-102944113 on Build GRCh38
rs781726531 Chr.14: 102943904-102943904 on Build GRCh38
rs751692127 Chr.14: 102944064-102944064 on Build GRCh38
rs760541840 Chr.14: 102944042-102944042 on Build GRCh38
rs766354752 Chr.14: 102943950-102943950 on Build GRCh38
rs764317338 Chr.14: 102943961-102943961 on Build GRCh38
rs779419153 Chr.14: 102944237-102944237 on Build GRCh38
rs541329650 Chr.14: 102943928-102943928 on Build GRCh38
rs759807460 Chr.14: 102944274-102944274 on Build GRCh38
rs773912384 Chr.14: 102944028-102944028 on Build GRCh38
rs761922472 Chr.14: 102944067-102944067 on Build GRCh38
rs577521745 Chr.14: 102943941-102943941 on Build GRCh38
rs143564466 Chr.14: 102944248-102944248 on Build GRCh38
rs35906112 Chr.14: 102943861-102943862 on Build GRCh38
rs756136564 Chr.14: 102943869-102943869 on Build GRCh38
rs761505951 Chr.14: 102943934-102943934 on Build GRCh38
rs564119735 Chr.14: 102943930-102943930 on Build GRCh38
rs546461837 Chr.14: 102943897-102943897 on Build GRCh38
rs537476700 Chr.14: 102944180-102944180 on Build GRCh38
rs150321401 Chr.14: 102944048-102944048 on Build GRCh38
rs774671721 Chr.14: 102944173-102944173 on Build GRCh38
rs755279217 Chr.14: 102944235-102944235 on Build GRCh38
rs746689190 Chr.14: 102944139-102944139 on Build GRCh38
rs374533525 Chr.14: 102943898-102943898 on Build GRCh38
rs767538657 Chr.14: 102944179-102944179 on Build GRCh38
rs528394229 Chr.14: 102944220-102944220 on Build GRCh38
rs371032953 Chr.14: 102943843-102943843 on Build GRCh38
rs763037888 Chr.14: 102944277-102944277 on Build GRCh38
rs775799497 Chr.14: 102943916-102943916 on Build GRCh38
rs956441357 Chr.14: 102943900-102943900 on Build GRCh38
rs760320198 Chr.14: 102943949-102943949 on Build GRCh38
rs145737227 Chr.14: 102944068-102944068 on Build GRCh38
rs754160186 Chr.14: 102944223-102944223 on Build GRCh38
rs756382043 Chr.14: 102943998-102943998 on Build GRCh38
rs776355137 Chr.14: 102944150-102944150 on Build GRCh38
rs968226449 Chr.14: 102944060-102944060 on Build GRCh38
rs760866818 Chr.14: 102944219-102944219 on Build GRCh38
rs146207239 Chr.14: 102944093-102944093 on Build GRCh38
rs753678351 Chr.14: 102943952-102943952 on Build GRCh38
rs140245177 Chr.14: 102944026-102944026 on Build GRCh38
rs1015220671 Chr.14: 102944252-102944252 on Build GRCh38
rs776462657 Chr.14: 102944049-102944049 on Build GRCh38
rs770334280 Chr.14: 102943912-102943912 on Build GRCh38
rs572426882 Chr.14: 102943876-102943876 on Build GRCh38
rs376192133 Chr.14: 102943859-102943859 on Build GRCh38
rs529138453 Chr.14: 102944127-102944127 on Build GRCh38
rs770601956 Chr.14: 102944147-102944147 on Build GRCh38
rs745676686 Chr.14: 102944271-102944271 on Build GRCh38
rs775397689 Chr.14: 102944275-102944275 on Build GRCh38
rs752827355 Chr.14: 102943887-102943887 on Build GRCh38
rs777205662 Chr.14: 102944251-102944251 on Build GRCh38
rs140674190 Chr.14: 102943903-102943903 on Build GRCh38
rs190067249 Chr.14: 102944165-102944165 on Build GRCh38
rs747272392 Chr.14: 102943901-102943901 on Build GRCh38
rs566447749 Chr.14: 102943931-102943931 on Build GRCh38
rs151083985 Chr.14: 102943997-102943997 on Build GRCh38
rs773059863 Chr.14: 102943942-102943942 on Build GRCh38
rs1002396118 Chr.14: 102944161-102944161 on Build GRCh38
rs138893409 Chr.14: 102944129-102944129 on Build GRCh38
rs754913461 Chr.14: 102943958-102943958 on Build GRCh38
rs759603636 Chr.14: 102944054-102944054 on Build GRCh38
rs149229554 Chr.14: 102944153-102944153 on Build GRCh38
rs762012281 Chr.14: 102944176-102944176 on Build GRCh38
rs944799076 Chr.14: 102943969-102943969 on Build GRCh38
rs763306847 Chr.14: 102943905-102943905 on Build GRCh38
rs747653502 Chr.14: 102944033-102944033 on Build GRCh38

View Full Product Details