Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 24961943-24962444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNRPN
Entrez Gene ID
6638
Gene Name
small nuclear ribonucleoprotein polypeptide N
Gene Aliases
HCERN3, PWCR, RT-LI, SM-D, SMN, SNRNP-N, SNURF-SNRPN, sm-N
UniGene
Hs.555970
Gene Chromosome Location
Chr.15: 24823637-24978723 on Build GRCh38
Transcripts
NM_003097.4 NM_022805.3 NM_022806.3 NM_022807.3 NM_022808.3 NM_005678.4 NM_022804.2

Gene Symbol
SNURF
Entrez Gene ID
8926
Gene Name
SNRPN upstream reading frame
Gene Aliases
-
UniGene
Hs.564847
Gene Chromosome Location
Chr.15: 24954893-24978723 on Build GRCh38
Transcripts
NM_003097.4 NM_022805.3 NM_022806.3 NM_022807.3 NM_022808.3 NM_005678.4 NM_022804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6521890 c.23T>A p.L8* Chr.15: 24962122-24962122 on Build GRCh38
1608192 c.52G>A p.V18I Chr.15: 24962151-24962151 on Build GRCh38
3956472 c.63G>T p.V21V Chr.15: 24962162-24962162 on Build GRCh38
6142041 c.52G>T p.V18L Chr.15: 24962151-24962151 on Build GRCh38
1323264 c.79C>G p.R27G Chr.15: 24962178-24962178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779030659 Chr.15: 24962083-24962083 on Build GRCh38
rs987504055 Chr.15: 24962088-24962088 on Build GRCh38
rs770705956 Chr.15: 24962116-24962116 on Build GRCh38
rs770772789 Chr.15: 24962223-24962223 on Build GRCh38
rs371527962 Chr.15: 24962134-24962134 on Build GRCh38
rs761310114 Chr.15: 24962253-24962253 on Build GRCh38
rs189091708 Chr.15: 24961976-24961976 on Build GRCh38
rs1055118076 Chr.15: 24961975-24961975 on Build GRCh38
rs967909633 Chr.15: 24962047-24962047 on Build GRCh38
rs543838201 Chr.15: 24962369-24962369 on Build GRCh38
rs961673082 Chr.15: 24962406-24962406 on Build GRCh38
rs766822978 Chr.15: 24962172-24962172 on Build GRCh38
rs371808119 Chr.15: 24962217-24962217 on Build GRCh38
rs1008662286 Chr.15: 24962389-24962389 on Build GRCh38
rs1046393538 Chr.15: 24962010-24962010 on Build GRCh38
rs779706602 Chr.15: 24962140-24962140 on Build GRCh38
rs754147955 Chr.15: 24962178-24962178 on Build GRCh38
rs752792774 Chr.15: 24962203-24962203 on Build GRCh38
rs999455566 Chr.15: 24962020-24962020 on Build GRCh38
rs145306813 Chr.15: 24962208-24962208 on Build GRCh38
rs1046798350 Chr.15: 24962267-24962267 on Build GRCh38
rs996008096 Chr.15: 24962366-24962366 on Build GRCh38
rs954523605 Chr.15: 24962085-24962085 on Build GRCh38
rs181548004 Chr.15: 24962386-24962386 on Build GRCh38
rs996037904 Chr.15: 24962072-24962072 on Build GRCh38
rs902392714 Chr.15: 24962011-24962011 on Build GRCh38
rs776280755 Chr.15: 24962118-24962118 on Build GRCh38
rs1053689792 Chr.15: 24962340-24962340 on Build GRCh38
rs895187699 Chr.15: 24962276-24962276 on Build GRCh38
rs375201041 Chr.15: 24962233-24962233 on Build GRCh38
rs760960743 Chr.15: 24962168-24962168 on Build GRCh38
rs777635512 Chr.15: 24962096-24962096 on Build GRCh38
rs754033392 Chr.15: 24962074-24962074 on Build GRCh38
rs1022219855 Chr.15: 24962358-24962358 on Build GRCh38
rs752589572 Chr.15: 24962410-24962410 on Build GRCh38
rs1032152084 Chr.15: 24962033-24962033 on Build GRCh38
rs755265989 Chr.15: 24962179-24962179 on Build GRCh38
rs773984800 Chr.15: 24962252-24962252 on Build GRCh38
rs426541 Chr.15: 24962126-24962126 on Build GRCh38
rs144974597 Chr.15: 24962273-24962273 on Build GRCh38
rs973073479 Chr.15: 24962413-24962413 on Build GRCh38
rs920173008 Chr.15: 24962420-24962420 on Build GRCh38
rs746807698 Chr.15: 24962100-24962100 on Build GRCh38
rs959624711 Chr.15: 24962012-24962012 on Build GRCh38
rs775243777 Chr.15: 24962239-24962239 on Build GRCh38
rs770006588 Chr.15: 24962217-24962217 on Build GRCh38
rs768170160 Chr.15: 24962150-24962150 on Build GRCh38
rs112194258 Chr.15: 24962390-24962390 on Build GRCh38
rs112824826 Chr.15: 24962109-24962109 on Build GRCh38
rs745335026 Chr.15: 24962299-24962299 on Build GRCh38
rs768223074 Chr.15: 24962251-24962251 on Build GRCh38
rs972979014 Chr.15: 24962188-24962188 on Build GRCh38
rs140705231 Chr.15: 24962234-24962234 on Build GRCh38
rs542255437 Chr.15: 24962099-24962099 on Build GRCh38
rs893762635 Chr.15: 24962365-24962365 on Build GRCh38
rs1013998654 Chr.15: 24961984-24961984 on Build GRCh38
rs11555312 Chr.15: 24962169-24962169 on Build GRCh38
rs199839388 Chr.15: 24962151-24962151 on Build GRCh38
rs376718476 Chr.15: 24962207-24962207 on Build GRCh38
rs755140475 Chr.15: 24962082-24962082 on Build GRCh38
rs199986212 Chr.15: 24962220-24962220 on Build GRCh38
rs762505215 Chr.15: 24962146-24962146 on Build GRCh38
rs772044025 Chr.15: 24962094-24962094 on Build GRCh38
rs902529140 Chr.15: 24962279-24962279 on Build GRCh38
rs957838506 Chr.15: 24962069-24962069 on Build GRCh38
rs775190608 Chr.15: 24962144-24962144 on Build GRCh38
rs769254063 Chr.15: 24962339-24962339 on Build GRCh38
rs886051016 Chr.15: 24962124-24962124 on Build GRCh38
rs954577837 Chr.15: 24962388-24962388 on Build GRCh38
rs1014980739 Chr.15: 24962398-24962398 on Build GRCh38
rs779082047 Chr.15: 24962191-24962191 on Build GRCh38
rs1012622296 Chr.15: 24962315-24962315 on Build GRCh38
rs977885182 Chr.15: 24962055-24962055 on Build GRCh38
rs1022631638 Chr.15: 24962034-24962034 on Build GRCh38
rs766763350 Chr.15: 24962260-24962260 on Build GRCh38
rs753428942 Chr.15: 24962018-24962018 on Build GRCh38
rs931375396 Chr.15: 24961969-24961969 on Build GRCh38
rs907543524 Chr.15: 24962099-24962099 on Build GRCh38

View Full Product Details