Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 26772055-26772546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRB3
Entrez Gene ID
2562
Gene Name
gamma-aminobutyric acid type A receptor beta3 subunit
Gene Aliases
ECA5
UniGene
Hs.302352
Gene Chromosome Location
Chr.15: 26543546-26773788 on Build GRCh38
Transcripts
NM_000814.5 NM_001278631.1 NM_021912.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
960025 c.229G>A p.E77K Chr.15: 26772413-26772413 on Build GRCh38
1372179 c.179C>T p.P60L Chr.15: 26772463-26772463 on Build GRCh38
7122947 c.354C>T p.C118C Chr.15: 26772456-26772456 on Build GRCh38
4686145 c.376G>A p.A126T Chr.15: 26772434-26772434 on Build GRCh38
7122945 c.186C>T p.C62C Chr.15: 26772456-26772456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776070336 Chr.15: 26772507-26772508 on Build GRCh38
rs577808120 Chr.15: 26772077-26772077 on Build GRCh38
rs746317416 Chr.15: 26772513-26772513 on Build GRCh38
rs938882313 Chr.15: 26772285-26772285 on Build GRCh38
rs757803732 Chr.15: 26772504-26772504 on Build GRCh38
rs763579672 Chr.15: 26772261-26772261 on Build GRCh38
rs1042745419 Chr.15: 26772338-26772338 on Build GRCh38
rs1053302761 Chr.15: 26772250-26772250 on Build GRCh38
rs1028119993 Chr.15: 26772373-26772373 on Build GRCh38
rs375978851 Chr.15: 26772369-26772369 on Build GRCh38
rs184297289 Chr.15: 26772143-26772143 on Build GRCh38
rs72708067 Chr.15: 26772404-26772404 on Build GRCh38
rs942053036 Chr.15: 26772081-26772081 on Build GRCh38
rs766026039 Chr.15: 26772381-26772381 on Build GRCh38
rs45486192 Chr.15: 26772106-26772106 on Build GRCh38
rs372750222 Chr.15: 26772366-26772366 on Build GRCh38
rs773067819 Chr.15: 26772483-26772483 on Build GRCh38
rs973954719 Chr.15: 26772085-26772085 on Build GRCh38
rs1012550358 Chr.15: 26772178-26772178 on Build GRCh38
rs760650068 Chr.15: 26772254-26772254 on Build GRCh38
rs770214111 Chr.15: 26772435-26772435 on Build GRCh38
rs746262322 Chr.15: 26772429-26772429 on Build GRCh38
rs764702427 Chr.15: 26772500-26772500 on Build GRCh38
rs976965669 Chr.15: 26772242-26772242 on Build GRCh38
rs531579727 Chr.15: 26772337-26772337 on Build GRCh38
rs778479790 Chr.15: 26772394-26772394 on Build GRCh38
rs935784308 Chr.15: 26772243-26772243 on Build GRCh38
rs376029442 Chr.15: 26772153-26772153 on Build GRCh38
rs574070209 Chr.15: 26772228-26772228 on Build GRCh38
rs1043278195 Chr.15: 26772508-26772508 on Build GRCh38
rs540256645 Chr.15: 26772473-26772473 on Build GRCh38
rs988571432 Chr.15: 26772475-26772475 on Build GRCh38
rs761754263 Chr.15: 26772478-26772478 on Build GRCh38
rs949922748 Chr.15: 26772258-26772258 on Build GRCh38
rs999042646 Chr.15: 26772359-26772359 on Build GRCh38
rs774313933 Chr.15: 26772128-26772128 on Build GRCh38
rs562574616 Chr.15: 26772288-26772288 on Build GRCh38
rs1045623688 Chr.15: 26772278-26772278 on Build GRCh38
rs1026847082 Chr.15: 26772364-26772364 on Build GRCh38
rs766150547 Chr.15: 26772489-26772489 on Build GRCh38
rs775510229 Chr.15: 26772224-26772224 on Build GRCh38
rs904285777 Chr.15: 26772202-26772202 on Build GRCh38
rs905792320 Chr.15: 26772287-26772287 on Build GRCh38
rs752273392 Chr.15: 26772503-26772503 on Build GRCh38
rs755863611 Chr.15: 26772379-26772379 on Build GRCh38
rs775865564 Chr.15: 26772462-26772462 on Build GRCh38
rs774223323 Chr.15: 26772362-26772362 on Build GRCh38
rs899183365 Chr.15: 26772358-26772358 on Build GRCh38
rs974413010 Chr.15: 26772219-26772219 on Build GRCh38
rs965230731 Chr.15: 26772230-26772230 on Build GRCh38
rs916172825 Chr.15: 26772300-26772300 on Build GRCh38
rs749519657 Chr.15: 26772468-26772468 on Build GRCh38
rs778097525 Chr.15: 26772459-26772459 on Build GRCh38
rs776175139 Chr.15: 26772294-26772294 on Build GRCh38
rs542676158 Chr.15: 26772286-26772286 on Build GRCh38
rs151221282 Chr.15: 26772411-26772411 on Build GRCh38
rs754625640 Chr.15: 26772492-26772492 on Build GRCh38
rs912188044 Chr.15: 26772156-26772156 on Build GRCh38
rs894707932 Chr.15: 26772166-26772166 on Build GRCh38
rs8179186 Chr.15: 26772389-26772389 on Build GRCh38
rs760478130 Chr.15: 26772484-26772484 on Build GRCh38
rs942975966 Chr.15: 26772218-26772218 on Build GRCh38
rs938959914 Chr.15: 26772139-26772139 on Build GRCh38
rs772141476 Chr.15: 26772158-26772158 on Build GRCh38
rs753553523 Chr.15: 26772490-26772490 on Build GRCh38
rs370136185 Chr.15: 26772375-26772375 on Build GRCh38
rs768859258 Chr.15: 26772425-26772425 on Build GRCh38
rs868775950 Chr.15: 26772169-26772169 on Build GRCh38
rs756602022 Chr.15: 26772515-26772515 on Build GRCh38
rs547678442 Chr.15: 26772474-26772474 on Build GRCh38
rs747510658 Chr.15: 26772397-26772397 on Build GRCh38
rs934481193 Chr.15: 26772232-26772232 on Build GRCh38
rs371144939 Chr.15: 26772482-26772482 on Build GRCh38
rs960366680 Chr.15: 26772476-26772476 on Build GRCh38
rs1021212633 Chr.15: 26772180-26772180 on Build GRCh38
rs953907811 Chr.15: 26772246-26772246 on Build GRCh38
rs25408 Chr.15: 26772390-26772390 on Build GRCh38
rs963504730 Chr.15: 26772074-26772074 on Build GRCh38
rs764173133 Chr.15: 26772361-26772361 on Build GRCh38
rs200439016 Chr.15: 26772414-26772414 on Build GRCh38
rs1045551246 Chr.15: 26772520-26772520 on Build GRCh38

View Full Product Details