Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48992517-48993052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SECISBP2L
Entrez Gene ID
9728
Gene Name
SECIS binding protein 2 like
Gene Aliases
SBP2L, SLAN
UniGene
Hs.9997
Gene Chromosome Location
Chr.15: 48988638-49046563 on Build GRCh38
Transcripts
NM_001193489.1 NM_014701.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6271637 c.2958C>A p.G986G Chr.15: 48992592-48992592 on Build GRCh38
3734604 c.2698G>A p.E900K Chr.15: 48992717-48992717 on Build GRCh38
6896709 c.2965G>A p.E989K Chr.15: 48992585-48992585 on Build GRCh38
5586522 c.2838G>C p.V946V Chr.15: 48992712-48992712 on Build GRCh38
4251842 c.2786C>T p.S929F Chr.15: 48992629-48992629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757655160 Chr.15: 48992607-48992607 on Build GRCh38
rs754443634 Chr.15: 48992911-48992911 on Build GRCh38
rs1016312081 Chr.15: 48992945-48992945 on Build GRCh38
rs372115192 Chr.15: 48992812-48992812 on Build GRCh38
rs113905266 Chr.15: 48992995-48992995 on Build GRCh38
rs757122762 Chr.15: 48992965-48992965 on Build GRCh38
rs780947292 Chr.15: 48992679-48992679 on Build GRCh38
rs772983096 Chr.15: 48992717-48992717 on Build GRCh38
rs770961521 Chr.15: 48992811-48992811 on Build GRCh38
rs755597602 Chr.15: 48992563-48992568 on Build GRCh38
rs764837115 Chr.15: 48992675-48992675 on Build GRCh38
rs745826156 Chr.15: 48992976-48992976 on Build GRCh38
rs984985320 Chr.15: 48992831-48992831 on Build GRCh38
rs764550754 Chr.15: 48992575-48992575 on Build GRCh38
rs557594280 Chr.15: 48992774-48992774 on Build GRCh38
rs779225819 Chr.15: 48992614-48992614 on Build GRCh38
rs752063029 Chr.15: 48992907-48992907 on Build GRCh38
rs748932940 Chr.15: 48992605-48992613 on Build GRCh38
rs537764723 Chr.15: 48992577-48992577 on Build GRCh38
rs769424220 Chr.15: 48992806-48992806 on Build GRCh38
rs768891202 Chr.15: 48992828-48992828 on Build GRCh38
rs201669301 Chr.15: 48992753-48992753 on Build GRCh38
rs375217888 Chr.15: 48992870-48992870 on Build GRCh38
rs370194787 Chr.15: 48992732-48992732 on Build GRCh38
rs1046930092 Chr.15: 48992880-48992880 on Build GRCh38
rs978211148 Chr.15: 48992647-48992647 on Build GRCh38
rs1016505091 Chr.15: 48992950-48992950 on Build GRCh38
rs377620350 Chr.15: 48992565-48992565 on Build GRCh38
rs905037882 Chr.15: 48992593-48992593 on Build GRCh38
rs757893661 Chr.15: 48992821-48992821 on Build GRCh38
rs368290048 Chr.15: 48992889-48992889 on Build GRCh38
rs942355893 Chr.15: 48992787-48992787 on Build GRCh38
rs765943198 Chr.15: 48992929-48992929 on Build GRCh38
rs747877554 Chr.15: 48992699-48992699 on Build GRCh38
rs777191607 Chr.15: 48992553-48992553 on Build GRCh38
rs752268484 Chr.15: 48992926-48992926 on Build GRCh38
rs553236108 Chr.15: 48992798-48992798 on Build GRCh38
rs370743392 Chr.15: 48992616-48992616 on Build GRCh38
rs773214500 Chr.15: 48992556-48992556 on Build GRCh38
rs978346632 Chr.15: 48993009-48993009 on Build GRCh38
rs766809747 Chr.15: 48992904-48992904 on Build GRCh38
rs750875229 Chr.15: 48992756-48992756 on Build GRCh38
rs575902579 Chr.15: 48992985-48992985 on Build GRCh38
rs372621671 Chr.15: 48992919-48992919 on Build GRCh38
rs777418676 Chr.15: 48992940-48992940 on Build GRCh38
rs762146040 Chr.15: 48992857-48992857 on Build GRCh38
rs34895054 Chr.15: 48992804-48992804 on Build GRCh38
rs200681309 Chr.15: 48992814-48992814 on Build GRCh38
rs769635839 Chr.15: 48992704-48992704 on Build GRCh38
rs772021884 Chr.15: 48992719-48992719 on Build GRCh38
rs1024954939 Chr.15: 48992979-48992979 on Build GRCh38
rs1029117212 Chr.15: 48992820-48992820 on Build GRCh38
rs368413086 Chr.15: 48992771-48992771 on Build GRCh38
rs754118083 Chr.15: 48992751-48992751 on Build GRCh38
rs754730165 Chr.15: 48992551-48992553 on Build GRCh38
rs778954838 Chr.15: 48992970-48992970 on Build GRCh38
rs532203256 Chr.15: 48993003-48993003 on Build GRCh38
rs564723193 Chr.15: 48992999-48992999 on Build GRCh38
rs892413454 Chr.15: 48992785-48992785 on Build GRCh38
rs776948494 Chr.15: 48992856-48992856 on Build GRCh38
rs1031219403 Chr.15: 48993018-48993018 on Build GRCh38
rs770576476 Chr.15: 48992965-48992966 on Build GRCh38
rs146594630 Chr.15: 48992645-48992645 on Build GRCh38
rs949170866 Chr.15: 48992571-48992572 on Build GRCh38
rs749056822 Chr.15: 48992962-48992962 on Build GRCh38
rs374106456 Chr.15: 48992796-48992796 on Build GRCh38
rs543327616 Chr.15: 48992994-48992994 on Build GRCh38
rs917582162 Chr.15: 48992988-48992992 on Build GRCh38
rs757799972 Chr.15: 48992644-48992644 on Build GRCh38
rs140289675 Chr.15: 48992726-48992726 on Build GRCh38
rs17853133 Chr.15: 48992571-48992571 on Build GRCh38
rs1025153028 Chr.15: 48992574-48992574 on Build GRCh38
rs762046923 Chr.15: 48992752-48992752 on Build GRCh38
rs751123921 Chr.15: 48992773-48992773 on Build GRCh38
rs762968155 Chr.15: 48992718-48992718 on Build GRCh38
rs750864237 Chr.15: 48992876-48992876 on Build GRCh38
rs535103584 Chr.15: 48992769-48992769 on Build GRCh38
rs953516488 Chr.15: 48992827-48992827 on Build GRCh38
rs768361689 Chr.15: 48992668-48992668 on Build GRCh38
rs201240214 Chr.15: 48992572-48992574 on Build GRCh38
rs368661434 Chr.15: 48992809-48992809 on Build GRCh38
rs764354379 Chr.15: 48992748-48992748 on Build GRCh38
rs759467573 Chr.15: 48992561-48992561 on Build GRCh38

View Full Product Details