Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49327874-49328384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
UniGene
Hs.712959
Gene Chromosome Location
Chr.15: 49326997-49620931 on Build GRCh38
Transcripts
XM_017021993.1 NM_001001556.2 NM_001289030.1 NM_001289031.1 NM_002044.3 XM_005254279.3 XM_005254280.3 XM_005254284.3 XM_006720461.3 XM_006720462.3 XM_006720463.3 XM_006720464.3 XM_011521441.2 XM_017022062.1 XM_017022063.1 XM_017022064.1 XM_017022065.1 XM_017022066.1

Gene Symbol
GALK2
Entrez Gene ID
2585
Gene Name
galactokinase 2
Gene Aliases
GK2
UniGene
Hs.122006
Gene Chromosome Location
Chr.15: 49155757-49367883 on Build GRCh38
Transcripts
XM_017021993.1 NM_001001556.2 NM_001289030.1 NM_001289031.1 NM_002044.3 XM_005254279.3 XM_005254280.3 XM_005254284.3 XM_006720461.3 XM_006720462.3 XM_006720463.3 XM_006720464.3 XM_011521441.2 XM_017022062.1 XM_017022063.1 XM_017022064.1 XM_017022065.1 XM_017022066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
470756 c.1192C>T p.R398* Chr.15: 49327974-49327974 on Build GRCh38
6505626 c.1310C>T p.P437L Chr.15: 49328125-49328125 on Build GRCh38
3401792 c.1165A>G p.T389A Chr.15: 49327980-49327980 on Build GRCh38
6414487 c.1190C>A p.T397K Chr.15: 49328005-49328005 on Build GRCh38
6142358 c.1336A>T p.T446S Chr.15: 49328118-49328118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199516811 Chr.15: 49328148-49328148 on Build GRCh38
rs538069623 Chr.15: 49328039-49328039 on Build GRCh38
rs781211218 Chr.15: 49328110-49328110 on Build GRCh38
rs200728035 Chr.15: 49328115-49328115 on Build GRCh38
rs1055254 Chr.15: 49328003-49328003 on Build GRCh38
rs760278080 Chr.15: 49328208-49328208 on Build GRCh38
rs764253278 Chr.15: 49328192-49328192 on Build GRCh38
rs756310332 Chr.15: 49328166-49328167 on Build GRCh38
rs761561337 Chr.15: 49328084-49328085 on Build GRCh38
rs917272024 Chr.15: 49328164-49328164 on Build GRCh38
rs867528286 Chr.15: 49328092-49328092 on Build GRCh38
rs776952706 Chr.15: 49328273-49328273 on Build GRCh38
rs767130682 Chr.15: 49327974-49327974 on Build GRCh38
rs987155768 Chr.15: 49327985-49327985 on Build GRCh38
rs750823699 Chr.15: 49327978-49327978 on Build GRCh38
rs757107457 Chr.15: 49328006-49328006 on Build GRCh38
rs774873579 Chr.15: 49328155-49328155 on Build GRCh38
rs765062312 Chr.15: 49328106-49328107 on Build GRCh38
rs939493249 Chr.15: 49327915-49327915 on Build GRCh38
rs752276156 Chr.15: 49328056-49328056 on Build GRCh38
rs770920694 Chr.15: 49327908-49327908 on Build GRCh38
rs576306720 Chr.15: 49328352-49328352 on Build GRCh38
rs749120317 Chr.15: 49327902-49327902 on Build GRCh38
rs952021263 Chr.15: 49328141-49328141 on Build GRCh38
rs758981126 Chr.15: 49328044-49328044 on Build GRCh38
rs373620147 Chr.15: 49328023-49328023 on Build GRCh38
rs751092726 Chr.15: 49327990-49327990 on Build GRCh38
rs1050750712 Chr.15: 49328327-49328327 on Build GRCh38
rs916860225 Chr.15: 49328336-49328336 on Build GRCh38
rs368992857 Chr.15: 49327918-49327918 on Build GRCh38
rs1000036230 Chr.15: 49328215-49328215 on Build GRCh38
rs768167821 Chr.15: 49328187-49328187 on Build GRCh38
rs747073827 Chr.15: 49328122-49328122 on Build GRCh38
rs556418019 Chr.15: 49328088-49328088 on Build GRCh38
rs946133210 Chr.15: 49328183-49328183 on Build GRCh38
rs139485976 Chr.15: 49327981-49327981 on Build GRCh38
rs191170777 Chr.15: 49327922-49327922 on Build GRCh38
rs767081825 Chr.15: 49328055-49328055 on Build GRCh38
rs763844729 Chr.15: 49328080-49328080 on Build GRCh38
rs370217644 Chr.15: 49328095-49328095 on Build GRCh38
rs948274014 Chr.15: 49328341-49328341 on Build GRCh38
rs767278183 Chr.15: 49327994-49327994 on Build GRCh38
rs774828410 Chr.15: 49328189-49328189 on Build GRCh38
rs776896239 Chr.15: 49327960-49327960 on Build GRCh38
rs554876731 Chr.15: 49328333-49328333 on Build GRCh38
rs371198789 Chr.15: 49328024-49328024 on Build GRCh38
rs377181822 Chr.15: 49328013-49328013 on Build GRCh38
rs867379954 Chr.15: 49327980-49327980 on Build GRCh38
rs770088619 Chr.15: 49328031-49328031 on Build GRCh38
rs768844377 Chr.15: 49327939-49327939 on Build GRCh38
rs150920378 Chr.15: 49328035-49328035 on Build GRCh38
rs760074206 Chr.15: 49328177-49328177 on Build GRCh38
rs755806512 Chr.15: 49327996-49327996 on Build GRCh38
rs768211728 Chr.15: 49327918-49327918 on Build GRCh38
rs777379469 Chr.15: 49328002-49328002 on Build GRCh38
rs766949973 Chr.15: 49327984-49327984 on Build GRCh38
rs1057047672 Chr.15: 49328139-49328139 on Build GRCh38
rs773542023 Chr.15: 49328128-49328128 on Build GRCh38
rs746306645 Chr.15: 49328036-49328036 on Build GRCh38
rs376206527 Chr.15: 49328032-49328032 on Build GRCh38
rs766787587 Chr.15: 49328051-49328051 on Build GRCh38
rs182130124 Chr.15: 49328214-49328214 on Build GRCh38
rs755679971 Chr.15: 49328061-49328061 on Build GRCh38
rs147218231 Chr.15: 49328132-49328132 on Build GRCh38
rs768789510 Chr.15: 49328019-49328019 on Build GRCh38
rs983943521 Chr.15: 49328156-49328156 on Build GRCh38
rs760127486 Chr.15: 49328054-49328054 on Build GRCh38
rs745738396 Chr.15: 49328096-49328096 on Build GRCh38
rs916832088 Chr.15: 49327923-49327923 on Build GRCh38
rs750345601 Chr.15: 49328130-49328131 on Build GRCh38
rs918740176 Chr.15: 49328057-49328057 on Build GRCh38
rs754598196 Chr.15: 49327993-49327993 on Build GRCh38
rs762160419 Chr.15: 49327963-49327963 on Build GRCh38
rs760312729 Chr.15: 49327902-49327904 on Build GRCh38
rs757133440 Chr.15: 49328091-49328091 on Build GRCh38
rs754537910 Chr.15: 49327909-49327909 on Build GRCh38
rs780775645 Chr.15: 49328074-49328074 on Build GRCh38
rs767565189 Chr.15: 49328165-49328166 on Build GRCh38
rs752730802 Chr.15: 49328166-49328172 on Build GRCh38
rs776364370 Chr.15: 49327939-49327942 on Build GRCh38
rs984738916 Chr.15: 49328094-49328094 on Build GRCh38
rs775068164 Chr.15: 49328052-49328052 on Build GRCh38
rs748423606 Chr.15: 49328277-49328277 on Build GRCh38
rs1000513428 Chr.15: 49327926-49327926 on Build GRCh38
rs747078307 Chr.15: 49328014-49328014 on Build GRCh38
rs542597283 Chr.15: 49328313-49328313 on Build GRCh38
rs762942743 Chr.15: 49328158-49328159 on Build GRCh38

View Full Product Details