Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58185753-58186225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP9
Entrez Gene ID
366
Gene Name
aquaporin 9
Gene Aliases
AQP-9, HsT17287, SSC1, T17287
UniGene
Hs.104624
Gene Chromosome Location
Chr.15: 58138169-58185911 on Build GRCh38
Transcripts
NM_001320635.1 NM_001320636.1 NM_020980.4

SNP Detail

SNP ID Chromosome Location
rs554195478 Chr.15: 58186009-58186009 on Build GRCh38
rs888943596 Chr.15: 58186191-58186191 on Build GRCh38
rs1036280017 Chr.15: 58185932-58185932 on Build GRCh38
rs763432756 Chr.15: 58185988-58185988 on Build GRCh38
rs866094993 Chr.15: 58185989-58185989 on Build GRCh38
rs569306262 Chr.15: 58185880-58185880 on Build GRCh38
rs192068207 Chr.15: 58186176-58186176 on Build GRCh38
rs750066318 Chr.15: 58186136-58186136 on Build GRCh38
rs1051186452 Chr.15: 58185976-58185976 on Build GRCh38
rs545176483 Chr.15: 58186118-58186118 on Build GRCh38
rs891184877 Chr.15: 58185997-58185997 on Build GRCh38
rs113373867 Chr.15: 58186075-58186075 on Build GRCh38
rs529661319 Chr.15: 58186193-58186193 on Build GRCh38
rs1006961523 Chr.15: 58186000-58186000 on Build GRCh38
rs538443851 Chr.15: 58185883-58185883 on Build GRCh38
rs571767415 Chr.15: 58185954-58185954 on Build GRCh38
rs16939897 Chr.15: 58185818-58185818 on Build GRCh38
rs551728298 Chr.15: 58185901-58185901 on Build GRCh38
rs933161085 Chr.15: 58186185-58186185 on Build GRCh38
rs910675800 Chr.15: 58185839-58185839 on Build GRCh38
rs539661520 Chr.15: 58186043-58186043 on Build GRCh38
rs780631245 Chr.15: 58186107-58186107 on Build GRCh38
rs536130828 Chr.15: 58186054-58186054 on Build GRCh38
rs981164070 Chr.15: 58185840-58185840 on Build GRCh38
rs928347909 Chr.15: 58185842-58185842 on Build GRCh38
rs529389102 Chr.15: 58186000-58186000 on Build GRCh38
rs373389367 Chr.15: 58185902-58185902 on Build GRCh38
rs967527544 Chr.15: 58186070-58186070 on Build GRCh38
rs1801805 Chr.15: 58185807-58185807 on Build GRCh38
rs188463175 Chr.15: 58186011-58186011 on Build GRCh38
rs146774634 Chr.15: 58186093-58186093 on Build GRCh38
rs373789999 Chr.15: 58186108-58186108 on Build GRCh38
rs991239524 Chr.15: 58186135-58186135 on Build GRCh38
rs534318965 Chr.15: 58185966-58185966 on Build GRCh38
rs540760719 Chr.15: 58186183-58186183 on Build GRCh38
rs957885343 Chr.15: 58185792-58185792 on Build GRCh38
rs897698420 Chr.15: 58185950-58185950 on Build GRCh38
rs17240643 Chr.15: 58185868-58185868 on Build GRCh38
rs529526755 Chr.15: 58185841-58185841 on Build GRCh38
rs74016594 Chr.15: 58186174-58186174 on Build GRCh38
rs908865621 Chr.15: 58185900-58185900 on Build GRCh38
rs1036281638 Chr.15: 58186182-58186182 on Build GRCh38
rs1051622936 Chr.15: 58186187-58186187 on Build GRCh38
rs574099962 Chr.15: 58186052-58186052 on Build GRCh38
rs763119704 Chr.15: 58185998-58185999 on Build GRCh38
rs958723075 Chr.15: 58186109-58186109 on Build GRCh38
rs542507852 Chr.15: 58186156-58186156 on Build GRCh38
rs140612392 Chr.15: 58186184-58186184 on Build GRCh38
rs957390359 Chr.15: 58186018-58186018 on Build GRCh38
rs1012275049 Chr.15: 58186032-58186032 on Build GRCh38
rs990396937 Chr.15: 58185802-58185802 on Build GRCh38

View Full Product Details