Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 62647195-62647705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLN2
Entrez Gene ID
83660
Gene Name
talin 2
Gene Aliases
ILWEQ
UniGene
Hs.569438
Gene Chromosome Location
Chr.15: 62390527-62844631 on Build GRCh38
Transcripts
NM_015059.2 XM_005254708.4 XM_005254710.4 XM_005254711.4 XM_005254712.4 XM_005254713.4 XM_005254714.3 XM_005254715.2 XM_006720717.3 XM_017022665.1 XM_017022666.1 XM_017022667.1 XM_017022668.1 XM_017022669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6142570 c.127A>G p.T43A Chr.15: 62647437-62647437 on Build GRCh38
5470371 c.89C>T p.A30V Chr.15: 62647399-62647399 on Build GRCh38
4607295 c.136+3A>G p.? Chr.15: 62647449-62647449 on Build GRCh38
1678517 c.53C>A p.T18N Chr.15: 62647363-62647363 on Build GRCh38
2254583 c.90G>A p.A30A Chr.15: 62647400-62647400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191191909 Chr.15: 62647235-62647235 on Build GRCh38
rs754599334 Chr.15: 62647436-62647436 on Build GRCh38
rs925523711 Chr.15: 62647544-62647544 on Build GRCh38
rs929671612 Chr.15: 62647654-62647654 on Build GRCh38
rs1033950106 Chr.15: 62647234-62647234 on Build GRCh38
rs936931473 Chr.15: 62647553-62647553 on Build GRCh38
rs554667095 Chr.15: 62647405-62647405 on Build GRCh38
rs945556490 Chr.15: 62647254-62647254 on Build GRCh38
rs938979 Chr.15: 62647241-62647241 on Build GRCh38
rs759982549 Chr.15: 62647387-62647387 on Build GRCh38
rs759702004 Chr.15: 62647309-62647309 on Build GRCh38
rs766964116 Chr.15: 62647285-62647285 on Build GRCh38
rs570869418 Chr.15: 62647260-62647260 on Build GRCh38
rs199834807 Chr.15: 62647263-62647263 on Build GRCh38
rs1014006627 Chr.15: 62647575-62647575 on Build GRCh38
rs201936321 Chr.15: 62647395-62647395 on Build GRCh38
rs1006407761 Chr.15: 62647543-62647543 on Build GRCh38
rs779158349 Chr.15: 62647471-62647471 on Build GRCh38
rs543042136 Chr.15: 62647561-62647561 on Build GRCh38
rs775667508 Chr.15: 62647430-62647430 on Build GRCh38
rs62004575 Chr.15: 62647419-62647419 on Build GRCh38
rs753752860 Chr.15: 62647336-62647336 on Build GRCh38
rs377088784 Chr.15: 62647457-62647457 on Build GRCh38
rs746594719 Chr.15: 62647363-62647363 on Build GRCh38
rs764065913 Chr.15: 62647536-62647536 on Build GRCh38
rs888226152 Chr.15: 62647675-62647675 on Build GRCh38
rs550449505 Chr.15: 62647277-62647277 on Build GRCh38
rs772773448 Chr.15: 62647481-62647481 on Build GRCh38
rs374212152 Chr.15: 62647612-62647613 on Build GRCh38
rs1000861063 Chr.15: 62647293-62647293 on Build GRCh38
rs771276314 Chr.15: 62647458-62647458 on Build GRCh38
rs552556907 Chr.15: 62647242-62647242 on Build GRCh38
rs774273131 Chr.15: 62647308-62647308 on Build GRCh38
rs977945740 Chr.15: 62647290-62647291 on Build GRCh38
rs780862989 Chr.15: 62647443-62647443 on Build GRCh38
rs958094700 Chr.15: 62647610-62647611 on Build GRCh38
rs746576084 Chr.15: 62647420-62647420 on Build GRCh38
rs943997861 Chr.15: 62647635-62647635 on Build GRCh38
rs779549971 Chr.15: 62647355-62647355 on Build GRCh38
rs964373254 Chr.15: 62647679-62647679 on Build GRCh38
rs760975468 Chr.15: 62647394-62647394 on Build GRCh38
rs190280271 Chr.15: 62647653-62647653 on Build GRCh38
rs895122897 Chr.15: 62647571-62647571 on Build GRCh38
rs901983241 Chr.15: 62647251-62647251 on Build GRCh38
rs758960977 Chr.15: 62647321-62647321 on Build GRCh38
rs1025017820 Chr.15: 62647572-62647572 on Build GRCh38
rs770072589 Chr.15: 62647397-62647397 on Build GRCh38
rs757119282 Chr.15: 62647402-62647402 on Build GRCh38
rs772531521 Chr.15: 62647239-62647239 on Build GRCh38
rs775700147 Chr.15: 62647482-62647482 on Build GRCh38
rs773569049 Chr.15: 62647282-62647282 on Build GRCh38
rs1017495341 Chr.15: 62647670-62647670 on Build GRCh38
rs759722896 Chr.15: 62647382-62647382 on Build GRCh38
rs539152254 Chr.15: 62647413-62647413 on Build GRCh38
rs770083711 Chr.15: 62647279-62647279 on Build GRCh38
rs751602502 Chr.15: 62647629-62647629 on Build GRCh38
rs771261514 Chr.15: 62647341-62647341 on Build GRCh38
rs372931716 Chr.15: 62647318-62647318 on Build GRCh38
rs144527579 Chr.15: 62647352-62647352 on Build GRCh38
rs967237612 Chr.15: 62647287-62647287 on Build GRCh38
rs548461372 Chr.15: 62647673-62647673 on Build GRCh38
rs182549452 Chr.15: 62647677-62647677 on Build GRCh38
rs746320193 Chr.15: 62647480-62647480 on Build GRCh38
rs910297940 Chr.15: 62647228-62647228 on Build GRCh38
rs747055227 Chr.15: 62647490-62647490 on Build GRCh38
rs764974667 Chr.15: 62647404-62647404 on Build GRCh38
rs371712298 Chr.15: 62647524-62647524 on Build GRCh38
rs371919183 Chr.15: 62647414-62647414 on Build GRCh38
rs760887788 Chr.15: 62647486-62647486 on Build GRCh38
rs763128171 Chr.15: 62647531-62647531 on Build GRCh38
rs897263374 Chr.15: 62647611-62647611 on Build GRCh38
rs573866070 Chr.15: 62647626-62647626 on Build GRCh38
rs887913869 Chr.15: 62647509-62647510 on Build GRCh38
rs562001190 Chr.15: 62647583-62647583 on Build GRCh38
rs763442217 Chr.15: 62647284-62647284 on Build GRCh38
rs111713111 Chr.15: 62647516-62647516 on Build GRCh38
rs745341172 Chr.15: 62647342-62647342 on Build GRCh38
rs775166489 Chr.15: 62647274-62647274 on Build GRCh38
rs951949229 Chr.15: 62647648-62647648 on Build GRCh38
rs775718505 Chr.15: 62647390-62647390 on Build GRCh38
rs147271420 Chr.15: 62647379-62647379 on Build GRCh38
rs368066703 Chr.15: 62647381-62647381 on Build GRCh38
rs753666948 Chr.15: 62647400-62647400 on Build GRCh38
rs767648413 Chr.15: 62647388-62647388 on Build GRCh38
rs182556346 Chr.15: 62647399-62647399 on Build GRCh38
rs757129797 Chr.15: 62647337-62647337 on Build GRCh38
rs774879092 Chr.15: 62647384-62647384 on Build GRCh38
rs536031805 Chr.15: 62647331-62647331 on Build GRCh38
rs1026125995 Chr.15: 62647645-62647645 on Build GRCh38
rs148535228 Chr.15: 62647513-62647513 on Build GRCh38
rs895215801 Chr.15: 62647325-62647325 on Build GRCh38
rs1032448239 Chr.15: 62647585-62647585 on Build GRCh38
rs763916983 Chr.15: 62647332-62647332 on Build GRCh38

View Full Product Details