Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63305838-63306337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APH1B
Entrez Gene ID
83464
Gene Name
aph-1 homolog B, gamma-secretase subunit
Gene Aliases
APH-1B, PRO1328, PSFL, TAAV688
UniGene
Hs.511703
Gene Chromosome Location
Chr.15: 63277519-63309126 on Build GRCh38
Transcripts
NM_001145646.1 NM_031301.3

SNP Detail

SNP ID Chromosome Location
rs573393272 Chr.15: 63306186-63306186 on Build GRCh38
rs200629722 Chr.15: 63305877-63305877 on Build GRCh38
rs199999925 Chr.15: 63305904-63305904 on Build GRCh38
rs200362473 Chr.15: 63306002-63306002 on Build GRCh38
rs201493118 Chr.15: 63306281-63306281 on Build GRCh38
rs200888369 Chr.15: 63306044-63306044 on Build GRCh38
rs201007891 Chr.15: 63306188-63306188 on Build GRCh38
rs201808167 Chr.15: 63306048-63306048 on Build GRCh38
rs745338 Chr.15: 63306231-63306231 on Build GRCh38
rs201197544 Chr.15: 63305939-63305939 on Build GRCh38
rs199721473 Chr.15: 63305997-63305997 on Build GRCh38
rs1033752498 Chr.15: 63305892-63305892 on Build GRCh38
rs200004783 Chr.15: 63305957-63305957 on Build GRCh38
rs149361000 Chr.15: 63305972-63305972 on Build GRCh38
rs201674146 Chr.15: 63306215-63306215 on Build GRCh38
rs199884325 Chr.15: 63305936-63305936 on Build GRCh38
rs201441763 Chr.15: 63306017-63306017 on Build GRCh38
rs202180122 Chr.15: 63305962-63305962 on Build GRCh38
rs202176678 Chr.15: 63306206-63306206 on Build GRCh38
rs57919583 Chr.15: 63306189-63306189 on Build GRCh38
rs138800714 Chr.15: 63305944-63305944 on Build GRCh38
rs927833769 Chr.15: 63306298-63306298 on Build GRCh38
rs554787281 Chr.15: 63305974-63305974 on Build GRCh38
rs187849012 Chr.15: 63306092-63306092 on Build GRCh38
rs150647820 Chr.15: 63305905-63305905 on Build GRCh38
rs922873983 Chr.15: 63305947-63305947 on Build GRCh38
rs200565462 Chr.15: 63306212-63306212 on Build GRCh38
rs867423243 Chr.15: 63306272-63306272 on Build GRCh38
rs199560509 Chr.15: 63306039-63306039 on Build GRCh38
rs557683221 Chr.15: 63305868-63305868 on Build GRCh38
rs184584190 Chr.15: 63305963-63305963 on Build GRCh38
rs200740048 Chr.15: 63306196-63306196 on Build GRCh38
rs961524717 Chr.15: 63306164-63306164 on Build GRCh38
rs199683846 Chr.15: 63306295-63306295 on Build GRCh38
rs202045325 Chr.15: 63305886-63305886 on Build GRCh38
rs201716291 Chr.15: 63306174-63306174 on Build GRCh38
rs201245077 Chr.15: 63305994-63305994 on Build GRCh38
rs1015718802 Chr.15: 63306150-63306150 on Build GRCh38
rs796265681 Chr.15: 63306003-63306003 on Build GRCh38
rs945196098 Chr.15: 63306280-63306280 on Build GRCh38
rs760686418 Chr.15: 63305896-63305896 on Build GRCh38
rs201862046 Chr.15: 63306090-63306090 on Build GRCh38
rs201787473 Chr.15: 63305976-63305976 on Build GRCh38
rs200928901 Chr.15: 63305959-63305959 on Build GRCh38
rs199904531 Chr.15: 63306185-63306185 on Build GRCh38
rs201298219 Chr.15: 63306103-63306103 on Build GRCh38
rs200209058 Chr.15: 63305989-63305989 on Build GRCh38
rs200506623 Chr.15: 63306088-63306088 on Build GRCh38
rs200173599 Chr.15: 63306266-63306266 on Build GRCh38
rs199617451 Chr.15: 63306060-63306060 on Build GRCh38
rs144683364 Chr.15: 63306026-63306026 on Build GRCh38
rs906468731 Chr.15: 63306047-63306047 on Build GRCh38
rs200754743 Chr.15: 63306313-63306313 on Build GRCh38
rs1005635198 Chr.15: 63306142-63306142 on Build GRCh38
rs200254084 Chr.15: 63306191-63306191 on Build GRCh38
rs201739277 Chr.15: 63305930-63305930 on Build GRCh38
rs553366872 Chr.15: 63305975-63305975 on Build GRCh38

View Full Product Details