Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67827899-67828404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKOR1
Entrez Gene ID
390598
Gene Name
SKI family transcriptional corepressor 1
Gene Aliases
CORL1, FUSSEL15, LBXCOR1
UniGene
Hs.451224
Gene Chromosome Location
Chr.15: 67819704-67834561 on Build GRCh38
Transcripts
NM_001258024.1 XM_017022173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4964589 c.2309C>T p.P770L Chr.15: 67828137-67828137 on Build GRCh38
7267522 c.1961_1963delGCG p.G658delG Chr.15: 67828080-67828082 on Build GRCh38
4951830 c.2233C>T p.P745S Chr.15: 67828061-67828061 on Build GRCh38
5963341 c.2161C>T p.R721C Chr.15: 67827989-67827989 on Build GRCh38
6538333 c.2221T>G p.L741V Chr.15: 67828049-67828049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35101671 Chr.15: 67828118-67828118 on Build GRCh38
rs760726956 Chr.15: 67827999-67827999 on Build GRCh38
rs28410827 Chr.15: 67828289-67828289 on Build GRCh38
rs975319792 Chr.15: 67828304-67828304 on Build GRCh38
rs927453006 Chr.15: 67827940-67827940 on Build GRCh38
rs567723005 Chr.15: 67827995-67827995 on Build GRCh38
rs912570520 Chr.15: 67828191-67828191 on Build GRCh38
rs568492767 Chr.15: 67827991-67827991 on Build GRCh38
rs886893181 Chr.15: 67828275-67828275 on Build GRCh38
rs114075979 Chr.15: 67828378-67828378 on Build GRCh38
rs756238272 Chr.15: 67828080-67828082 on Build GRCh38
rs1039612552 Chr.15: 67828203-67828203 on Build GRCh38
rs774689995 Chr.15: 67827987-67827987 on Build GRCh38
rs1057409898 Chr.15: 67828095-67828095 on Build GRCh38
rs757856127 Chr.15: 67827941-67827941 on Build GRCh38
rs972022537 Chr.15: 67828082-67828082 on Build GRCh38
rs762453053 Chr.15: 67828100-67828100 on Build GRCh38
rs1003129090 Chr.15: 67828085-67828085 on Build GRCh38
rs542355123 Chr.15: 67828316-67828316 on Build GRCh38
rs377719980 Chr.15: 67828134-67828134 on Build GRCh38
rs1048130763 Chr.15: 67828251-67828251 on Build GRCh38
rs968570687 Chr.15: 67828308-67828308 on Build GRCh38
rs201412798 Chr.15: 67827946-67827946 on Build GRCh38
rs1034628856 Chr.15: 67828321-67828321 on Build GRCh38
rs200956599 Chr.15: 67828016-67828016 on Build GRCh38
rs993576867 Chr.15: 67828197-67828197 on Build GRCh38
rs531109148 Chr.15: 67827977-67827977 on Build GRCh38
rs552867831 Chr.15: 67828088-67828088 on Build GRCh38
rs1041451147 Chr.15: 67828063-67828063 on Build GRCh38
rs963809197 Chr.15: 67827934-67827934 on Build GRCh38
rs990218937 Chr.15: 67828137-67828137 on Build GRCh38
rs114784960 Chr.15: 67827967-67827967 on Build GRCh38
rs765854785 Chr.15: 67828132-67828132 on Build GRCh38
rs536815632 Chr.15: 67828216-67828216 on Build GRCh38
rs914390367 Chr.15: 67828019-67828019 on Build GRCh38
rs558061270 Chr.15: 67828154-67828154 on Build GRCh38
rs1048161631 Chr.15: 67828047-67828047 on Build GRCh38
rs566896623 Chr.15: 67828064-67828064 on Build GRCh38
rs915556219 Chr.15: 67828352-67828352 on Build GRCh38
rs899298916 Chr.15: 67828138-67828138 on Build GRCh38
rs922494 Chr.15: 67828306-67828306 on Build GRCh38
rs534271073 Chr.15: 67828021-67828021 on Build GRCh38
rs1034493109 Chr.15: 67828112-67828112 on Build GRCh38
rs757002109 Chr.15: 67827918-67827918 on Build GRCh38
rs763937787 Chr.15: 67828010-67828010 on Build GRCh38
rs764893629 Chr.15: 67828026-67828026 on Build GRCh38
rs954702213 Chr.15: 67827971-67827971 on Build GRCh38
rs572824774 Chr.15: 67828356-67828356 on Build GRCh38
rs778055217 Chr.15: 67828189-67828190 on Build GRCh38
rs977206652 Chr.15: 67828310-67828310 on Build GRCh38
rs1015169815 Chr.15: 67828076-67828076 on Build GRCh38
rs551214173 Chr.15: 67827982-67827982 on Build GRCh38
rs376252769 Chr.15: 67827920-67827920 on Build GRCh38
rs552536268 Chr.15: 67828343-67828343 on Build GRCh38
rs1003974282 Chr.15: 67828300-67828300 on Build GRCh38
rs1004326429 Chr.15: 67828074-67828074 on Build GRCh38
rs571472667 Chr.15: 67828182-67828182 on Build GRCh38
rs977856939 Chr.15: 67828183-67828183 on Build GRCh38
rs943931937 Chr.15: 67828200-67828200 on Build GRCh38
rs745963696 Chr.15: 67827989-67827989 on Build GRCh38
rs529049622 Chr.15: 67827929-67827929 on Build GRCh38
rs751490847 Chr.15: 67827939-67827940 on Build GRCh38
rs958840828 Chr.15: 67828324-67828324 on Build GRCh38
rs1013140037 Chr.15: 67828129-67828129 on Build GRCh38
rs1011790585 Chr.15: 67828331-67828331 on Build GRCh38
rs778107476 Chr.15: 67828180-67828180 on Build GRCh38
rs924336887 Chr.15: 67828311-67828311 on Build GRCh38
rs894705403 Chr.15: 67828114-67828114 on Build GRCh38
rs931173443 Chr.15: 67828225-67828225 on Build GRCh38
rs914546277 Chr.15: 67827973-67827973 on Build GRCh38
rs943875593 Chr.15: 67828009-67828009 on Build GRCh38
rs948989116 Chr.15: 67828363-67828363 on Build GRCh38
rs1003096782 Chr.15: 67828317-67828317 on Build GRCh38
rs748619734 Chr.15: 67827947-67827947 on Build GRCh38
rs756447847 Chr.15: 67827954-67827954 on Build GRCh38

View Full Product Details