Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 88898576-88899074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFGE8
Entrez Gene ID
4240
Gene Name
milk fat globule-EGF factor 8 protein
Gene Aliases
BA46, EDIL1, HMFG, HsT19888, MFG-E8, MFGM, OAcGD3S, SED1, SPAG10, hP47
UniGene
Hs.3745
Gene Chromosome Location
Chr.15: 88898683-88913469 on Build GRCh38
Transcripts
NM_001114614.2 NM_001310319.1 NM_001310320.1 NM_001310321.1 NM_005928.3

SNP Detail

SNP ID Chromosome Location
rs1003932262 Chr.15: 88898857-88898857 on Build GRCh38
rs940976434 Chr.15: 88898756-88898756 on Build GRCh38
rs3211278 Chr.15: 88898811-88898811 on Build GRCh38
rs1007968185 Chr.15: 88898845-88898845 on Build GRCh38
rs932193716 Chr.15: 88899009-88899009 on Build GRCh38
rs185683645 Chr.15: 88898668-88898668 on Build GRCh38
rs977239232 Chr.15: 88898723-88898723 on Build GRCh38
rs938554056 Chr.15: 88898749-88898749 on Build GRCh38
rs1802942 Chr.15: 88898844-88898844 on Build GRCh38
rs907973884 Chr.15: 88898938-88898948 on Build GRCh38
rs552953110 Chr.15: 88898994-88898994 on Build GRCh38
rs531226967 Chr.15: 88898944-88898944 on Build GRCh38
rs1049401111 Chr.15: 88898839-88898839 on Build GRCh38
rs1055438869 Chr.15: 88898619-88898619 on Build GRCh38
rs182415985 Chr.15: 88898854-88898854 on Build GRCh38
rs112937603 Chr.15: 88898679-88898679 on Build GRCh38
rs957939862 Chr.15: 88898890-88898890 on Build GRCh38
rs1019430601 Chr.15: 88898635-88898635 on Build GRCh38
rs565269483 Chr.15: 88898783-88898783 on Build GRCh38
rs115586162 Chr.15: 88898820-88898820 on Build GRCh38
rs1802945 Chr.15: 88898848-88898848 on Build GRCh38
rs966681335 Chr.15: 88898639-88898639 on Build GRCh38
rs1021400996 Chr.15: 88898993-88898993 on Build GRCh38
rs538382131 Chr.15: 88899028-88899028 on Build GRCh38
rs138152398 Chr.15: 88898700-88898700 on Build GRCh38
rs552749772 Chr.15: 88898634-88898634 on Build GRCh38
rs1041611003 Chr.15: 88898595-88898595 on Build GRCh38
rs150395576 Chr.15: 88898667-88898667 on Build GRCh38
rs368872939 Chr.15: 88898948-88898948 on Build GRCh38
rs949446085 Chr.15: 88898683-88898683 on Build GRCh38
rs571219227 Chr.15: 88898999-88898999 on Build GRCh38
rs915826381 Chr.15: 88899032-88899032 on Build GRCh38
rs1049451689 Chr.15: 88899015-88899015 on Build GRCh38
rs1019717625 Chr.15: 88898909-88898909 on Build GRCh38
rs8530 Chr.15: 88898947-88898947 on Build GRCh38
rs940846160 Chr.15: 88898958-88898958 on Build GRCh38
rs1002820017 Chr.15: 88898617-88898617 on Build GRCh38
rs978398100 Chr.15: 88898648-88898648 on Build GRCh38
rs975657684 Chr.15: 88898685-88898685 on Build GRCh38
rs115022860 Chr.15: 88899037-88899037 on Build GRCh38
rs540611270 Chr.15: 88898613-88898613 on Build GRCh38
rs1802944 Chr.15: 88898987-88898987 on Build GRCh38
rs538798533 Chr.15: 88898768-88898768 on Build GRCh38
rs562113332 Chr.15: 88898714-88898714 on Build GRCh38
rs973561170 Chr.15: 88898959-88898959 on Build GRCh38
rs982257123 Chr.15: 88898687-88898687 on Build GRCh38
rs930973528 Chr.15: 88898829-88898829 on Build GRCh38
rs886891500 Chr.15: 88898662-88898662 on Build GRCh38
rs541425430 Chr.15: 88898830-88898830 on Build GRCh38
rs571338299 Chr.15: 88898703-88898703 on Build GRCh38
rs747634017 Chr.15: 88899034-88899034 on Build GRCh38
rs982208958 Chr.15: 88898997-88898997 on Build GRCh38
rs902228557 Chr.15: 88899038-88899038 on Build GRCh38
rs1023560660 Chr.15: 88898932-88898932 on Build GRCh38
rs549451279 Chr.15: 88898945-88898945 on Build GRCh38
rs553701505 Chr.15: 88898886-88898886 on Build GRCh38
rs1038356482 Chr.15: 88898796-88898796 on Build GRCh38
rs1032385924 Chr.15: 88898653-88898653 on Build GRCh38
rs990797253 Chr.15: 88898904-88898904 on Build GRCh38
rs772176104 Chr.15: 88898810-88898810 on Build GRCh38
rs928464992 Chr.15: 88898740-88898740 on Build GRCh38
rs902343654 Chr.15: 88898870-88898870 on Build GRCh38
rs908019906 Chr.15: 88898696-88898696 on Build GRCh38
rs1802946 Chr.15: 88898893-88898893 on Build GRCh38
rs115538013 Chr.15: 88898840-88898840 on Build GRCh38
rs995622312 Chr.15: 88898643-88898643 on Build GRCh38
rs551986864 Chr.15: 88898885-88898885 on Build GRCh38
rs11557310 Chr.15: 88898789-88898789 on Build GRCh38
rs368416349 Chr.15: 88898620-88898620 on Build GRCh38

View Full Product Details