Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99974239-99974783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS17
Entrez Gene ID
170691
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 17
Gene Aliases
-
UniGene
Hs.513200
Gene Chromosome Location
Chr.15: 99971438-100342924 on Build GRCh38
Transcripts
NM_139057.3 XM_005254872.3 XM_011521312.2 XM_017021973.1 XM_017021975.1 XM_017021976.1 XM_017021978.1 XM_017021979.1 XM_017021980.1 XM_017021982.1 XM_017021983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3499675 c.3175C>T p.R1059W Chr.15: 99974515-99974515 on Build GRCh38
4488097 c.3274C>T p.P1092S Chr.15: 99974416-99974416 on Build GRCh38
6053809 c.3130G>T p.A1044S Chr.15: 99974560-99974560 on Build GRCh38
6055325 c.3192G>T p.K1064N Chr.15: 99974498-99974498 on Build GRCh38
7211355 c.3278C>T p.P1093L Chr.15: 99974412-99974412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542056339 Chr.15: 99974669-99974669 on Build GRCh38
rs148559983 Chr.15: 99974568-99974568 on Build GRCh38
rs374214853 Chr.15: 99974381-99974381 on Build GRCh38
rs74036647 Chr.15: 99974677-99974677 on Build GRCh38
rs201387097 Chr.15: 99974515-99974515 on Build GRCh38
rs1020568161 Chr.15: 99974687-99974687 on Build GRCh38
rs755550706 Chr.15: 99974447-99974447 on Build GRCh38
rs563364315 Chr.15: 99974675-99974675 on Build GRCh38
rs371570653 Chr.15: 99974526-99974526 on Build GRCh38
rs777329258 Chr.15: 99974459-99974459 on Build GRCh38
rs757020850 Chr.15: 99974571-99974571 on Build GRCh38
rs570787589 Chr.15: 99974659-99974659 on Build GRCh38
rs886050972 Chr.15: 99974546-99974546 on Build GRCh38
rs371297701 Chr.15: 99974311-99974312 on Build GRCh38
rs777030019 Chr.15: 99974534-99974534 on Build GRCh38
rs955574819 Chr.15: 99974656-99974656 on Build GRCh38
rs780853868 Chr.15: 99974466-99974466 on Build GRCh38
rs980363324 Chr.15: 99974297-99974297 on Build GRCh38
rs373899380 Chr.15: 99974412-99974412 on Build GRCh38
rs140274211 Chr.15: 99974465-99974465 on Build GRCh38
rs765072526 Chr.15: 99974435-99974435 on Build GRCh38
rs79095568 Chr.15: 99974278-99974278 on Build GRCh38
rs112214202 Chr.15: 99974552-99974552 on Build GRCh38
rs779509724 Chr.15: 99974448-99974448 on Build GRCh38
rs377535229 Chr.15: 99974611-99974611 on Build GRCh38
rs765663952 Chr.15: 99974489-99974489 on Build GRCh38
rs759825365 Chr.15: 99974536-99974536 on Build GRCh38
rs895103115 Chr.15: 99974741-99974741 on Build GRCh38
rs1051942719 Chr.15: 99974314-99974315 on Build GRCh38
rs746100007 Chr.15: 99974595-99974595 on Build GRCh38
rs58960452 Chr.15: 99974303-99974303 on Build GRCh38
rs139031334 Chr.15: 99974311-99974311 on Build GRCh38
rs1012120799 Chr.15: 99974621-99974621 on Build GRCh38
rs754201774 Chr.15: 99974444-99974444 on Build GRCh38
rs760823694 Chr.15: 99974481-99974481 on Build GRCh38
rs186946807 Chr.15: 99974594-99974594 on Build GRCh38
rs953529961 Chr.15: 99974742-99974742 on Build GRCh38
rs3825894 Chr.15: 99974320-99974320 on Build GRCh38
rs563962465 Chr.15: 99974754-99974754 on Build GRCh38
rs752805401 Chr.15: 99974422-99974422 on Build GRCh38
rs777160312 Chr.15: 99974503-99974503 on Build GRCh38
rs939039374 Chr.15: 99974736-99974736 on Build GRCh38
rs749781134 Chr.15: 99974364-99974364 on Build GRCh38
rs541197112 Chr.15: 99974378-99974378 on Build GRCh38
rs1044670211 Chr.15: 99974661-99974661 on Build GRCh38
rs770889652 Chr.15: 99974533-99974533 on Build GRCh38
rs113965483 Chr.15: 99974737-99974737 on Build GRCh38
rs930329020 Chr.15: 99974660-99974660 on Build GRCh38
rs968692171 Chr.15: 99974592-99974592 on Build GRCh38
rs756484586 Chr.15: 99974396-99974396 on Build GRCh38
rs934150528 Chr.15: 99974403-99974403 on Build GRCh38
rs767614213 Chr.15: 99974436-99974436 on Build GRCh38
rs778582648 Chr.15: 99974406-99974406 on Build GRCh38
rs577051055 Chr.15: 99974620-99974620 on Build GRCh38
rs201896149 Chr.15: 99974415-99974415 on Build GRCh38
rs767596205 Chr.15: 99974387-99974387 on Build GRCh38
rs747870419 Chr.15: 99974371-99974371 on Build GRCh38
rs2573652 Chr.15: 99974409-99974409 on Build GRCh38
rs780775353 Chr.15: 99974512-99974512 on Build GRCh38
rs747077641 Chr.15: 99974331-99974331 on Build GRCh38
rs890248829 Chr.15: 99974315-99974315 on Build GRCh38
rs753229004 Chr.15: 99974452-99974452 on Build GRCh38
rs2581340 Chr.15: 99974356-99974356 on Build GRCh38
rs558417393 Chr.15: 99974346-99974346 on Build GRCh38
rs911713598 Chr.15: 99974633-99974633 on Build GRCh38
rs759480599 Chr.15: 99974424-99974424 on Build GRCh38
rs557893586 Chr.15: 99974617-99974617 on Build GRCh38
rs777496319 Chr.15: 99974389-99974389 on Build GRCh38
rs749048665 Chr.15: 99974579-99974579 on Build GRCh38
rs376675165 Chr.15: 99974483-99974483 on Build GRCh38
rs138196371 Chr.15: 99974274-99974274 on Build GRCh38
rs753320297 Chr.15: 99974494-99974494 on Build GRCh38
rs757473475 Chr.15: 99974384-99974384 on Build GRCh38
rs756890553 Chr.15: 99974506-99974506 on Build GRCh38
rs947952947 Chr.15: 99974750-99974750 on Build GRCh38
rs199498244 Chr.15: 99974373-99974373 on Build GRCh38
rs757883707 Chr.15: 99974453-99974453 on Build GRCh38
rs144817994 Chr.15: 99974405-99974405 on Build GRCh38
rs778731632 Chr.15: 99974601-99974606 on Build GRCh38
rs1007264176 Chr.15: 99974316-99974316 on Build GRCh38
rs376423645 Chr.15: 99974505-99974505 on Build GRCh38
rs201874119 Chr.15: 99974477-99974477 on Build GRCh38
rs749245963 Chr.15: 99974519-99974519 on Build GRCh38
rs1038775149 Chr.15: 99974411-99974411 on Build GRCh38
rs759464229 Chr.15: 99974382-99974382 on Build GRCh38
rs113885511 Chr.15: 99974578-99974578 on Build GRCh38
rs373278935 Chr.15: 99974431-99974431 on Build GRCh38
rs760336301 Chr.15: 99974379-99974379 on Build GRCh38
rs138405311 Chr.15: 99974487-99974487 on Build GRCh38
rs774049674 Chr.15: 99974263-99974263 on Build GRCh38
rs62040995 Chr.15: 99974694-99974694 on Build GRCh38
rs534579896 Chr.15: 99974623-99974623 on Build GRCh38
rs1000204793 Chr.15: 99974717-99974717 on Build GRCh38
rs770072956 Chr.15: 99974609-99974609 on Build GRCh38
rs1017187337 Chr.15: 99974317-99974317 on Build GRCh38
rs766170805 Chr.15: 99974380-99974380 on Build GRCh38
rs372853186 Chr.15: 99974469-99974469 on Build GRCh38
rs987049251 Chr.15: 99974664-99974664 on Build GRCh38
rs773331594 Chr.15: 99974479-99974479 on Build GRCh38
rs766559532 Chr.15: 99974482-99974482 on Build GRCh38
rs886050971 Chr.15: 99974291-99974291 on Build GRCh38
rs982589244 Chr.15: 99974279-99974279 on Build GRCh38
rs1022416989 Chr.15: 99974470-99974470 on Build GRCh38
rs112462169 Chr.15: 99974417-99974417 on Build GRCh38
rs376693261 Chr.15: 99974439-99974439 on Build GRCh38
rs191440300 Chr.15: 99974639-99974639 on Build GRCh38
rs200989311 Chr.15: 99974398-99974398 on Build GRCh38
rs746638782 Chr.15: 99974414-99974414 on Build GRCh38
rs373641684 Chr.15: 99974539-99974539 on Build GRCh38
rs369581271 Chr.15: 99974577-99974577 on Build GRCh38
rs920481855 Chr.15: 99974654-99974654 on Build GRCh38
rs1010520164 Chr.15: 99974678-99974678 on Build GRCh38
rs765090779 Chr.15: 99974425-99974425 on Build GRCh38
rs904947243 Chr.15: 99974665-99974665 on Build GRCh38
rs202106289 Chr.15: 99974467-99974467 on Build GRCh38
rs904737629 Chr.15: 99974709-99974709 on Build GRCh38
rs746697133 Chr.15: 99974462-99974462 on Build GRCh38
rs182634994 Chr.15: 99974574-99974574 on Build GRCh38
rs375751792 Chr.15: 99974590-99974590 on Build GRCh38
rs114338208 Chr.15: 99974635-99974635 on Build GRCh38
rs778954332 Chr.15: 99974586-99974586 on Build GRCh38
rs764344937 Chr.15: 99974614-99974614 on Build GRCh38
rs552244295 Chr.15: 99974427-99974427 on Build GRCh38
rs947418673 Chr.15: 99974342-99974342 on Build GRCh38
rs2581341 Chr.15: 99974289-99974289 on Build GRCh38
rs970324633 Chr.15: 99974295-99974295 on Build GRCh38
rs754906730 Chr.15: 99974583-99974583 on Build GRCh38
rs772111759 Chr.15: 99974478-99974478 on Build GRCh38
rs571067191 Chr.15: 99974275-99974275 on Build GRCh38
rs78682885 Chr.15: 99974471-99974471 on Build GRCh38
rs994687503 Chr.15: 99974321-99974321 on Build GRCh38
rs775402406 Chr.15: 99974451-99974451 on Build GRCh38
rs756676818 Chr.15: 99974362-99974362 on Build GRCh38
rs745497667 Chr.15: 99974514-99974514 on Build GRCh38
rs557053161 Chr.15: 99974312-99974312 on Build GRCh38
rs545799185 Chr.15: 99974345-99974345 on Build GRCh38
rs773998391 Chr.15: 99974367-99974367 on Build GRCh38
rs750652557 Chr.15: 99974438-99974438 on Build GRCh38
rs763372199 Chr.15: 99974542-99974542 on Build GRCh38
rs1030971543 Chr.15: 99974650-99974650 on Build GRCh38
rs747317920 Chr.15: 99974538-99974538 on Build GRCh38
rs760023197 Chr.15: 99974488-99974488 on Build GRCh38
rs1057340919 Chr.15: 99974670-99974670 on Build GRCh38
rs370976322 Chr.15: 99974610-99974610 on Build GRCh38
rs748780450 Chr.15: 99974498-99974499 on Build GRCh38
rs779626937 Chr.15: 99974601-99974601 on Build GRCh38
rs987944775 Chr.15: 99974752-99974752 on Build GRCh38
rs758924018 Chr.15: 99974498-99974498 on Build GRCh38
rs755384317 Chr.15: 99974689-99974689 on Build GRCh38
rs141373128 Chr.15: 99974525-99974525 on Build GRCh38
rs143850369 Chr.15: 99974286-99974286 on Build GRCh38
rs748001114 Chr.15: 99974588-99974588 on Build GRCh38
rs369831239 Chr.15: 99974388-99974388 on Build GRCh38
rs768325336 Chr.15: 99974366-99974366 on Build GRCh38
rs990860148 Chr.15: 99974292-99974292 on Build GRCh38
rs780375981 Chr.15: 99974363-99974363 on Build GRCh38
rs201992797 Chr.15: 99974383-99974383 on Build GRCh38
rs903699905 Chr.15: 99974753-99974753 on Build GRCh38

View Full Product Details