Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1778036-1778540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPSB3
Entrez Gene ID
90864
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
C16orf31, SSB3
UniGene
Hs.592080
Gene Chromosome Location
Chr.16: 1776712-1782605 on Build GRCh38
Transcripts
NM_001324081.1 NM_080861.3 XM_005255673.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6143903 c.368G>A p.R123H Chr.16: 1778258-1778258 on Build GRCh38
1167769 c.233delA p.E78fs*33 Chr.16: 1778506-1778506 on Build GRCh38
471386 c.260C>T p.S87L Chr.16: 1778479-1778479 on Build GRCh38
1227440 c.453G>A p.W151* Chr.16: 1778173-1778173 on Build GRCh38
3932246 c.324T>A p.D108E Chr.16: 1778302-1778302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376486568 Chr.16: 1778172-1778172 on Build GRCh38
rs755568189 Chr.16: 1778376-1778376 on Build GRCh38
rs762992230 Chr.16: 1778442-1778442 on Build GRCh38
rs1043570156 Chr.16: 1778249-1778249 on Build GRCh38
rs199872665 Chr.16: 1778206-1778206 on Build GRCh38
rs751539032 Chr.16: 1778405-1778405 on Build GRCh38
rs745349141 Chr.16: 1778270-1778270 on Build GRCh38
rs751359894 Chr.16: 1778091-1778091 on Build GRCh38
rs765936051 Chr.16: 1778115-1778115 on Build GRCh38
rs760434523 Chr.16: 1778456-1778456 on Build GRCh38
rs375566827 Chr.16: 1778428-1778428 on Build GRCh38
rs771733248 Chr.16: 1778107-1778107 on Build GRCh38
rs200328531 Chr.16: 1778076-1778076 on Build GRCh38
rs958273887 Chr.16: 1778346-1778346 on Build GRCh38
rs372022665 Chr.16: 1778435-1778435 on Build GRCh38
rs776048536 Chr.16: 1778116-1778116 on Build GRCh38
rs773188529 Chr.16: 1778377-1778388 on Build GRCh38
rs542354620 Chr.16: 1778242-1778242 on Build GRCh38
rs762079810 Chr.16: 1778123-1778123 on Build GRCh38
rs766442687 Chr.16: 1778212-1778212 on Build GRCh38
rs765413989 Chr.16: 1778317-1778317 on Build GRCh38
rs575732450 Chr.16: 1778265-1778265 on Build GRCh38
rs533566196 Chr.16: 1778396-1778396 on Build GRCh38
rs201470257 Chr.16: 1778365-1778365 on Build GRCh38
rs773904830 Chr.16: 1778389-1778389 on Build GRCh38
rs748723630 Chr.16: 1778275-1778275 on Build GRCh38
rs546828797 Chr.16: 1778426-1778426 on Build GRCh38
rs759985124 Chr.16: 1778070-1778070 on Build GRCh38
rs768351257 Chr.16: 1778438-1778438 on Build GRCh38
rs746224127 Chr.16: 1778494-1778494 on Build GRCh38
rs372948589 Chr.16: 1778227-1778227 on Build GRCh38
rs771892457 Chr.16: 1778152-1778152 on Build GRCh38
rs767832584 Chr.16: 1778126-1778126 on Build GRCh38
rs146273979 Chr.16: 1778263-1778263 on Build GRCh38
rs73483721 Chr.16: 1778500-1778500 on Build GRCh38
rs779630743 Chr.16: 1778383-1778383 on Build GRCh38
rs778576287 Chr.16: 1778475-1778475 on Build GRCh38
rs374237453 Chr.16: 1778353-1778353 on Build GRCh38
rs757071950 Chr.16: 1778097-1778097 on Build GRCh38
rs368103171 Chr.16: 1778363-1778363 on Build GRCh38
rs774972965 Chr.16: 1778191-1778191 on Build GRCh38
rs769386743 Chr.16: 1778271-1778271 on Build GRCh38
rs138713221 Chr.16: 1778090-1778090 on Build GRCh38
rs758340257 Chr.16: 1778330-1778330 on Build GRCh38
rs745600765 Chr.16: 1778372-1778372 on Build GRCh38
rs770188079 Chr.16: 1778497-1778497 on Build GRCh38
rs769068508 Chr.16: 1778180-1778180 on Build GRCh38
rs750661270 Chr.16: 1778204-1778204 on Build GRCh38
rs766098083 Chr.16: 1778461-1778461 on Build GRCh38
rs757427432 Chr.16: 1778077-1778077 on Build GRCh38
rs755820733 Chr.16: 1778417-1778417 on Build GRCh38
rs754902144 Chr.16: 1778145-1778145 on Build GRCh38
rs369818441 Chr.16: 1778429-1778429 on Build GRCh38
rs762239308 Chr.16: 1778197-1778197 on Build GRCh38
rs747972453 Chr.16: 1778327-1778328 on Build GRCh38
rs77388088 Chr.16: 1778508-1778508 on Build GRCh38
rs761605176 Chr.16: 1778512-1778512 on Build GRCh38
rs370198184 Chr.16: 1778412-1778412 on Build GRCh38
rs746719041 Chr.16: 1778158-1778158 on Build GRCh38
rs201565187 Chr.16: 1778069-1778069 on Build GRCh38
rs199857506 Chr.16: 1778450-1778450 on Build GRCh38
rs367827865 Chr.16: 1778099-1778099 on Build GRCh38
rs748625016 Chr.16: 1778385-1778385 on Build GRCh38
rs763479942 Chr.16: 1778179-1778179 on Build GRCh38
rs747795229 Chr.16: 1778106-1778106 on Build GRCh38
rs764001598 Chr.16: 1778404-1778404 on Build GRCh38
rs62040027 Chr.16: 1778154-1778154 on Build GRCh38
rs748784762 Chr.16: 1778425-1778425 on Build GRCh38
rs755133398 Chr.16: 1778218-1778218 on Build GRCh38
rs750714283 Chr.16: 1778131-1778131 on Build GRCh38
rs973637073 Chr.16: 1778395-1778395 on Build GRCh38
rs753883696 Chr.16: 1778213-1778213 on Build GRCh38
rs772726555 Chr.16: 1778386-1778386 on Build GRCh38
rs371769096 Chr.16: 1778392-1778392 on Build GRCh38
rs768061667 Chr.16: 1778203-1778203 on Build GRCh38
rs202190320 Chr.16: 1778333-1778333 on Build GRCh38
rs751381805 Chr.16: 1778338-1778338 on Build GRCh38
rs377461244 Chr.16: 1778075-1778075 on Build GRCh38
rs910521337 Chr.16: 1778083-1778083 on Build GRCh38
rs768381046 Chr.16: 1778165-1778166 on Build GRCh38
rs762941312 Chr.16: 1778400-1778420 on Build GRCh38
rs776700162 Chr.16: 1778393-1778393 on Build GRCh38
rs766944869 Chr.16: 1778060-1778060 on Build GRCh38
rs529163255 Chr.16: 1778081-1778081 on Build GRCh38
rs745969511 Chr.16: 1778089-1778089 on Build GRCh38
rs772022282 Chr.16: 1778516-1778516 on Build GRCh38
rs775581867 Chr.16: 1778397-1778397 on Build GRCh38
rs770373858 Chr.16: 1778161-1778161 on Build GRCh38
rs200890401 Chr.16: 1778439-1778439 on Build GRCh38
rs780787493 Chr.16: 1778229-1778246 on Build GRCh38
rs774021746 Chr.16: 1778103-1778103 on Build GRCh38
rs897537779 Chr.16: 1778230-1778230 on Build GRCh38
rs74002455 Chr.16: 1778281-1778281 on Build GRCh38
rs753180296 Chr.16: 1778073-1778073 on Build GRCh38
rs111388867 Chr.16: 1778080-1778080 on Build GRCh38
rs376234209 Chr.16: 1778140-1778140 on Build GRCh38
rs367742723 Chr.16: 1778424-1778424 on Build GRCh38
rs1030425848 Chr.16: 1778277-1778277 on Build GRCh38
rs145688103 Chr.16: 1778061-1778061 on Build GRCh38
rs867572923 Chr.16: 1778415-1778415 on Build GRCh38
rs1017887346 Chr.16: 1778334-1778334 on Build GRCh38
rs141651024 Chr.16: 1778478-1778478 on Build GRCh38
rs759526708 Chr.16: 1778314-1778314 on Build GRCh38
rs768440158 Chr.16: 1778098-1778098 on Build GRCh38
rs142760336 Chr.16: 1778507-1778507 on Build GRCh38
rs769550829 Chr.16: 1778347-1778347 on Build GRCh38
rs141701472 Chr.16: 1778146-1778146 on Build GRCh38
rs576953359 Chr.16: 1778122-1778122 on Build GRCh38
rs371589211 Chr.16: 1778101-1778101 on Build GRCh38
rs754518685 Chr.16: 1778471-1778471 on Build GRCh38
rs765708572 Chr.16: 1778071-1778071 on Build GRCh38
rs375261591 Chr.16: 1778096-1778096 on Build GRCh38
rs775651913 Chr.16: 1778419-1778420 on Build GRCh38
rs758172975 Chr.16: 1778240-1778240 on Build GRCh38
rs760144688 Chr.16: 1778114-1778114 on Build GRCh38
rs772897374 Chr.16: 1778113-1778113 on Build GRCh38
rs774718098 Chr.16: 1778274-1778274 on Build GRCh38
rs774494665 Chr.16: 1778509-1778509 on Build GRCh38
rs369515288 Chr.16: 1778219-1778219 on Build GRCh38
rs761386215 Chr.16: 1778059-1778059 on Build GRCh38
rs368805218 Chr.16: 1778455-1778455 on Build GRCh38
rs756258686 Chr.16: 1778139-1778139 on Build GRCh38
rs894119818 Chr.16: 1778159-1778159 on Build GRCh38
rs781365988 Chr.16: 1778481-1778481 on Build GRCh38
rs559780982 Chr.16: 1778268-1778268 on Build GRCh38
rs530459268 Chr.16: 1778470-1778470 on Build GRCh38
rs372168834 Chr.16: 1778151-1778151 on Build GRCh38
rs150535227 Chr.16: 1778479-1778479 on Build GRCh38
rs777394709 Chr.16: 1778156-1778156 on Build GRCh38
rs369677526 Chr.16: 1778445-1778445 on Build GRCh38
rs752570163 Chr.16: 1778325-1778325 on Build GRCh38
rs766487340 Chr.16: 1778308-1778308 on Build GRCh38
rs910805735 Chr.16: 1778119-1778119 on Build GRCh38
rs760930732 Chr.16: 1778294-1778294 on Build GRCh38
rs993213775 Chr.16: 1778272-1778272 on Build GRCh38
rs746600669 Chr.16: 1778253-1778253 on Build GRCh38
rs746449813 Chr.16: 1778098-1778098 on Build GRCh38
rs367618296 Chr.16: 1778074-1778074 on Build GRCh38
rs369022550 Chr.16: 1778288-1778288 on Build GRCh38
rs200130779 Chr.16: 1778121-1778121 on Build GRCh38
rs770705396 Chr.16: 1778407-1778422 on Build GRCh38
rs377110031 Chr.16: 1778451-1778451 on Build GRCh38

View Full Product Details