Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2784897-2785388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS33
Entrez Gene ID
260429
Gene Name
protease, serine 33
Gene Aliases
EOS
UniGene
Hs.280658
Gene Chromosome Location
Chr.16: 2783953-2787759 on Build GRCh38
Transcripts
NM_152891.2 XM_011522451.2 XM_011522452.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1377096 c.557C>A p.P186Q Chr.16: 2785129-2785129 on Build GRCh38
4059759 c.538C>A p.L180I Chr.16: 2785148-2785148 on Build GRCh38
7337766 c.557C>T p.P186L Chr.16: 2785129-2785129 on Build GRCh38
4632120 c.573C>T p.R191R Chr.16: 2785113-2785113 on Build GRCh38
5368354 c.661C>A p.Q221K Chr.16: 2785025-2785025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760754621 Chr.16: 2784972-2784972 on Build GRCh38
rs911248455 Chr.16: 2785155-2785155 on Build GRCh38
rs973683757 Chr.16: 2785166-2785166 on Build GRCh38
rs767837680 Chr.16: 2785328-2785328 on Build GRCh38
rs779216851 Chr.16: 2784989-2784989 on Build GRCh38
rs753094874 Chr.16: 2785330-2785330 on Build GRCh38
rs759329567 Chr.16: 2785045-2785045 on Build GRCh38
rs895933958 Chr.16: 2785151-2785151 on Build GRCh38
rs763915375 Chr.16: 2784975-2784975 on Build GRCh38
rs755866079 Chr.16: 2785223-2785224 on Build GRCh38
rs1012851101 Chr.16: 2785154-2785154 on Build GRCh38
rs554016491 Chr.16: 2785079-2785079 on Build GRCh38
rs761719514 Chr.16: 2785186-2785186 on Build GRCh38
rs914923491 Chr.16: 2785243-2785243 on Build GRCh38
rs748854900 Chr.16: 2785034-2785034 on Build GRCh38
rs12599918 Chr.16: 2785188-2785188 on Build GRCh38
rs895902605 Chr.16: 2785319-2785319 on Build GRCh38
rs770290645 Chr.16: 2785037-2785037 on Build GRCh38
rs751956445 Chr.16: 2785099-2785099 on Build GRCh38
rs1011425967 Chr.16: 2785345-2785345 on Build GRCh38
rs1056251575 Chr.16: 2785301-2785301 on Build GRCh38
rs752991810 Chr.16: 2785158-2785158 on Build GRCh38
rs866481869 Chr.16: 2785084-2785084 on Build GRCh38
rs764121705 Chr.16: 2785065-2785065 on Build GRCh38
rs755029066 Chr.16: 2785089-2785089 on Build GRCh38
rs370114600 Chr.16: 2785116-2785116 on Build GRCh38
rs1012144607 Chr.16: 2785098-2785098 on Build GRCh38
rs573561118 Chr.16: 2785193-2785193 on Build GRCh38
rs372511608 Chr.16: 2785105-2785105 on Build GRCh38
rs974807921 Chr.16: 2785230-2785230 on Build GRCh38
rs762983726 Chr.16: 2785195-2785195 on Build GRCh38
rs916135963 Chr.16: 2784990-2784990 on Build GRCh38
rs1046306723 Chr.16: 2785254-2785254 on Build GRCh38
rs963930408 Chr.16: 2785159-2785159 on Build GRCh38
rs773326023 Chr.16: 2785191-2785191 on Build GRCh38
rs776631245 Chr.16: 2785182-2785182 on Build GRCh38
rs371706924 Chr.16: 2785010-2785010 on Build GRCh38
rs866577680 Chr.16: 2785269-2785269 on Build GRCh38
rs538480085 Chr.16: 2784960-2784960 on Build GRCh38
rs756629711 Chr.16: 2785332-2785332 on Build GRCh38
rs746843281 Chr.16: 2785169-2785169 on Build GRCh38
rs757242287 Chr.16: 2784984-2784984 on Build GRCh38
rs755251116 Chr.16: 2785111-2785111 on Build GRCh38
rs753869322 Chr.16: 2785073-2785073 on Build GRCh38
rs757238975 Chr.16: 2785286-2785286 on Build GRCh38
rs1047716068 Chr.16: 2785221-2785221 on Build GRCh38
rs534239129 Chr.16: 2785060-2785060 on Build GRCh38
rs761962479 Chr.16: 2785076-2785076 on Build GRCh38
rs369502268 Chr.16: 2784997-2784997 on Build GRCh38
rs545476745 Chr.16: 2785259-2785259 on Build GRCh38
rs911624582 Chr.16: 2785215-2785215 on Build GRCh38
rs547079835 Chr.16: 2785218-2785218 on Build GRCh38
rs549098676 Chr.16: 2784923-2784923 on Build GRCh38
rs187976130 Chr.16: 2785320-2785320 on Build GRCh38
rs745401260 Chr.16: 2785342-2785342 on Build GRCh38
rs771890022 Chr.16: 2785142-2785142 on Build GRCh38
rs779865717 Chr.16: 2785344-2785344 on Build GRCh38
rs775473326 Chr.16: 2785053-2785053 on Build GRCh38
rs989482633 Chr.16: 2785146-2785146 on Build GRCh38
rs769249795 Chr.16: 2785014-2785014 on Build GRCh38
rs778240819 Chr.16: 2785271-2785271 on Build GRCh38
rs758764868 Chr.16: 2785009-2785009 on Build GRCh38
rs756639459 Chr.16: 2785119-2785119 on Build GRCh38
rs540558401 Chr.16: 2785337-2785337 on Build GRCh38
rs991674105 Chr.16: 2784976-2784976 on Build GRCh38
rs995468441 Chr.16: 2785356-2785356 on Build GRCh38
rs1042582668 Chr.16: 2785133-2785133 on Build GRCh38
rs942959955 Chr.16: 2784995-2784995 on Build GRCh38
rs998874503 Chr.16: 2785141-2785141 on Build GRCh38
rs564286625 Chr.16: 2785264-2785264 on Build GRCh38
rs759419641 Chr.16: 2784941-2784941 on Build GRCh38
rs950873119 Chr.16: 2785189-2785189 on Build GRCh38
rs1023005268 Chr.16: 2785156-2785156 on Build GRCh38
rs771837149 Chr.16: 2785047-2785047 on Build GRCh38
rs886064602 Chr.16: 2785083-2785083 on Build GRCh38
rs770727194 Chr.16: 2785129-2785129 on Build GRCh38
rs766613362 Chr.16: 2785202-2785202 on Build GRCh38
rs778625644 Chr.16: 2785125-2785125 on Build GRCh38
rs774153655 Chr.16: 2785038-2785038 on Build GRCh38
rs1023348261 Chr.16: 2785338-2785338 on Build GRCh38
rs757932285 Chr.16: 2785343-2785343 on Build GRCh38
rs894835456 Chr.16: 2785235-2785235 on Build GRCh38
rs374910362 Chr.16: 2785011-2785011 on Build GRCh38
rs970548683 Chr.16: 2785106-2785106 on Build GRCh38
rs768277213 Chr.16: 2785178-2785178 on Build GRCh38
rs753990780 Chr.16: 2784981-2784981 on Build GRCh38
rs745350400 Chr.16: 2785140-2785140 on Build GRCh38
rs568853302 Chr.16: 2784950-2784950 on Build GRCh38
rs571467804 Chr.16: 2785023-2785023 on Build GRCh38
rs369059939 Chr.16: 2785086-2785086 on Build GRCh38
rs781730827 Chr.16: 2785115-2785115 on Build GRCh38
rs369289564 Chr.16: 2785088-2785088 on Build GRCh38
rs939176424 Chr.16: 2785296-2785296 on Build GRCh38
rs775241646 Chr.16: 2785145-2785145 on Build GRCh38
rs755376356 Chr.16: 2785223-2785223 on Build GRCh38
rs752477591 Chr.16: 2785133-2785133 on Build GRCh38

View Full Product Details