Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3022065-3022562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF12A
Entrez Gene ID
51330
Gene Name
TNF receptor superfamily member 12A
Gene Aliases
CD266, FN14, TWEAKR
UniGene
Hs.355899
Gene Chromosome Location
Chr.16: 3020312-3022383 on Build GRCh38
Transcripts
NM_016639.2

SNP Detail

SNP ID Chromosome Location
rs949304010 Chr.16: 3022483-3022483 on Build GRCh38
rs921599752 Chr.16: 3022311-3022311 on Build GRCh38
rs1044326 Chr.16: 3022126-3022126 on Build GRCh38
rs1035656833 Chr.16: 3022225-3022225 on Build GRCh38
rs968466693 Chr.16: 3022507-3022507 on Build GRCh38
rs1022399544 Chr.16: 3022492-3022492 on Build GRCh38
rs146183720 Chr.16: 3022367-3022367 on Build GRCh38
rs565565983 Chr.16: 3022488-3022488 on Build GRCh38
rs3760087 Chr.16: 3022308-3022308 on Build GRCh38
rs35510228 Chr.16: 3022459-3022460 on Build GRCh38
rs1005175924 Chr.16: 3022284-3022284 on Build GRCh38
rs1009727489 Chr.16: 3022468-3022468 on Build GRCh38
rs931703652 Chr.16: 3022092-3022092 on Build GRCh38
rs751473512 Chr.16: 3022456-3022456 on Build GRCh38
rs542372828 Chr.16: 3022328-3022328 on Build GRCh38
rs1003914622 Chr.16: 3022459-3022459 on Build GRCh38
rs530903792 Chr.16: 3022353-3022353 on Build GRCh38
rs978387467 Chr.16: 3022097-3022097 on Build GRCh38
rs978420103 Chr.16: 3022307-3022307 on Build GRCh38
rs1003810930 Chr.16: 3022219-3022219 on Build GRCh38
rs142178519 Chr.16: 3022516-3022519 on Build GRCh38
rs979144554 Chr.16: 3022430-3022430 on Build GRCh38
rs558709641 Chr.16: 3022440-3022440 on Build GRCh38
rs190588947 Chr.16: 3022171-3022171 on Build GRCh38
rs1049118982 Chr.16: 3022257-3022257 on Build GRCh38
rs940498002 Chr.16: 3022166-3022166 on Build GRCh38
rs528594951 Chr.16: 3022172-3022172 on Build GRCh38
rs987733059 Chr.16: 3022451-3022451 on Build GRCh38
rs769271557 Chr.16: 3022443-3022443 on Build GRCh38
rs999917095 Chr.16: 3022534-3022534 on Build GRCh38
rs931613736 Chr.16: 3022314-3022314 on Build GRCh38
rs930696892 Chr.16: 3022442-3022443 on Build GRCh38
rs577469148 Chr.16: 3022113-3022113 on Build GRCh38
rs35992248 Chr.16: 3022537-3022537 on Build GRCh38
rs921882576 Chr.16: 3022106-3022106 on Build GRCh38
rs1023640218 Chr.16: 3022209-3022209 on Build GRCh38
rs34461776 Chr.16: 3022349-3022350 on Build GRCh38
rs940515633 Chr.16: 3022419-3022419 on Build GRCh38
rs546376536 Chr.16: 3022121-3022121 on Build GRCh38
rs892700999 Chr.16: 3022466-3022466 on Build GRCh38
rs1052302958 Chr.16: 3022162-3022162 on Build GRCh38
rs988234132 Chr.16: 3022278-3022278 on Build GRCh38
rs926435228 Chr.16: 3022522-3022522 on Build GRCh38
rs551945375 Chr.16: 3022442-3022442 on Build GRCh38
rs770624108 Chr.16: 3022216-3022216 on Build GRCh38
rs776017302 Chr.16: 3022464-3022464 on Build GRCh38
rs1029559163 Chr.16: 3022405-3022405 on Build GRCh38
rs905466860 Chr.16: 3022218-3022219 on Build GRCh38
rs563553036 Chr.16: 3022397-3022397 on Build GRCh38
rs887769110 Chr.16: 3022282-3022282 on Build GRCh38
rs953186763 Chr.16: 3022417-3022417 on Build GRCh38
rs905209193 Chr.16: 3022447-3022447 on Build GRCh38
rs937441924 Chr.16: 3022538-3022538 on Build GRCh38
rs935284603 Chr.16: 3022149-3022149 on Build GRCh38
rs532284218 Chr.16: 3022439-3022439 on Build GRCh38
rs905965620 Chr.16: 3022226-3022226 on Build GRCh38
rs1002006199 Chr.16: 3022250-3022250 on Build GRCh38
rs763478858 Chr.16: 3022494-3022494 on Build GRCh38
rs539590413 Chr.16: 3022107-3022107 on Build GRCh38
rs114883839 Chr.16: 3022343-3022343 on Build GRCh38
rs994874030 Chr.16: 3022195-3022195 on Build GRCh38
rs911760963 Chr.16: 3022399-3022399 on Build GRCh38
rs776141192 Chr.16: 3022293-3022293 on Build GRCh38
rs764531855 Chr.16: 3022504-3022504 on Build GRCh38
rs924938367 Chr.16: 3022135-3022135 on Build GRCh38
rs28595195 Chr.16: 3022090-3022090 on Build GRCh38

View Full Product Details