Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3141679-3142173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF213
Entrez Gene ID
7760
Gene Name
zinc finger protein 213
Gene Aliases
CR53, ZKSCAN21, ZSCAN53
UniGene
Hs.115284
Gene Chromosome Location
Chr.16: 3135056-3142804 on Build GRCh38
Transcripts
NM_001134655.1 NM_004220.2 NR_104432.1

SNP Detail

SNP ID Chromosome Location
rs945174300 Chr.16: 3141949-3141949 on Build GRCh38
rs34621102 Chr.16: 3141864-3141864 on Build GRCh38
rs377083396 Chr.16: 3141815-3141815 on Build GRCh38
rs979520497 Chr.16: 3141897-3141897 on Build GRCh38
rs945036467 Chr.16: 3141893-3141893 on Build GRCh38
rs1023316259 Chr.16: 3141801-3141802 on Build GRCh38
rs972292876 Chr.16: 3141882-3141882 on Build GRCh38
rs957983420 Chr.16: 3142129-3142129 on Build GRCh38
rs894734884 Chr.16: 3141983-3141983 on Build GRCh38
rs775133610 Chr.16: 3142087-3142087 on Build GRCh38
rs561506719 Chr.16: 3141942-3141942 on Build GRCh38
rs940593000 Chr.16: 3141699-3141699 on Build GRCh38
rs913822565 Chr.16: 3141880-3141880 on Build GRCh38
rs559539648 Chr.16: 3141871-3141871 on Build GRCh38
rs1016352917 Chr.16: 3141868-3141868 on Build GRCh38
rs146370668 Chr.16: 3142074-3142074 on Build GRCh38
rs954534921 Chr.16: 3142081-3142081 on Build GRCh38
rs545921448 Chr.16: 3141853-3141853 on Build GRCh38
rs541843584 Chr.16: 3141926-3141926 on Build GRCh38
rs956922788 Chr.16: 3141869-3141869 on Build GRCh38
rs934072645 Chr.16: 3141944-3141944 on Build GRCh38
rs531258592 Chr.16: 3141961-3141961 on Build GRCh38
rs572843483 Chr.16: 3141918-3141918 on Build GRCh38
rs923151305 Chr.16: 3141902-3141902 on Build GRCh38
rs374016789 Chr.16: 3141871-3141872 on Build GRCh38
rs754495193 Chr.16: 3141843-3141843 on Build GRCh38
rs907435977 Chr.16: 3141784-3141784 on Build GRCh38
rs544461396 Chr.16: 3141997-3141997 on Build GRCh38
rs146774741 Chr.16: 3141875-3141876 on Build GRCh38
rs12444830 Chr.16: 3142070-3142070 on Build GRCh38
rs1041924998 Chr.16: 3141782-3141782 on Build GRCh38
rs911698696 Chr.16: 3141978-3141978 on Build GRCh38
rs566986634 Chr.16: 3142075-3142075 on Build GRCh38
rs968718552 Chr.16: 3141938-3141938 on Build GRCh38
rs181090924 Chr.16: 3141735-3141735 on Build GRCh38
rs1056025107 Chr.16: 3141977-3141977 on Build GRCh38
rs985981748 Chr.16: 3141928-3141928 on Build GRCh38
rs187033990 Chr.16: 3142053-3142053 on Build GRCh38
rs1003151959 Chr.16: 3141792-3141792 on Build GRCh38
rs374298776 Chr.16: 3141927-3141927 on Build GRCh38
rs141597981 Chr.16: 3141758-3141758 on Build GRCh38
rs1003281531 Chr.16: 3142083-3142084 on Build GRCh38
rs904905753 Chr.16: 3141806-3141806 on Build GRCh38
rs920492483 Chr.16: 3142055-3142055 on Build GRCh38
rs1042260457 Chr.16: 3141965-3141965 on Build GRCh38
rs913633845 Chr.16: 3141948-3141948 on Build GRCh38
rs779350913 Chr.16: 3141940-3141940 on Build GRCh38
rs907347679 Chr.16: 3141966-3141966 on Build GRCh38
rs536739337 Chr.16: 3141855-3141855 on Build GRCh38
rs936301537 Chr.16: 3141941-3141941 on Build GRCh38
rs1006216204 Chr.16: 3142032-3142032 on Build GRCh38
rs191472806 Chr.16: 3142060-3142060 on Build GRCh38
rs985928731 Chr.16: 3142105-3142105 on Build GRCh38
rs1016300189 Chr.16: 3142054-3142054 on Build GRCh38
rs910466302 Chr.16: 3142120-3142120 on Build GRCh38
rs899098589 Chr.16: 3141765-3141765 on Build GRCh38
rs1003468197 Chr.16: 3141970-3141970 on Build GRCh38
rs940556249 Chr.16: 3142013-3142013 on Build GRCh38

View Full Product Details