Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23468769-23469263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGA2
Entrez Gene ID
23062
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 2
Gene Aliases
VEAR
UniGene
Hs.460336
Gene Chromosome Location
Chr.16: 23463542-23510494 on Build GRCh38
Transcripts
NM_015044.4 XM_017023074.1 XM_017023075.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
968775 c.1683G>A p.M561I Chr.16: 23468934-23468934 on Build GRCh38
6670652 c.1643C>T p.P548L Chr.16: 23468974-23468974 on Build GRCh38
4059399 c.1685C>A p.P562H Chr.16: 23468932-23468932 on Build GRCh38
4686846 c.1630G>A p.V544M Chr.16: 23468987-23468987 on Build GRCh38
3983062 c.1685C>T p.P562L Chr.16: 23468932-23468932 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1022071556 Chr.16: 23469074-23469074 on Build GRCh38
rs1000997231 Chr.16: 23469168-23469168 on Build GRCh38
rs769550020 Chr.16: 23468919-23468919 on Build GRCh38
rs906058681 Chr.16: 23468907-23468907 on Build GRCh38
rs747526759 Chr.16: 23468878-23468878 on Build GRCh38
rs754024013 Chr.16: 23468974-23468974 on Build GRCh38
rs150611801 Chr.16: 23468993-23468993 on Build GRCh38
rs781654354 Chr.16: 23469047-23469047 on Build GRCh38
rs780306056 Chr.16: 23469018-23469018 on Build GRCh38
rs755527794 Chr.16: 23469028-23469028 on Build GRCh38
rs139733534 Chr.16: 23468961-23468961 on Build GRCh38
rs747334082 Chr.16: 23469025-23469025 on Build GRCh38
rs924919396 Chr.16: 23468807-23468808 on Build GRCh38
rs190827792 Chr.16: 23469164-23469164 on Build GRCh38
rs758590159 Chr.16: 23469014-23469014 on Build GRCh38
rs757590565 Chr.16: 23468846-23468846 on Build GRCh38
rs1010252818 Chr.16: 23469068-23469068 on Build GRCh38
rs766104456 Chr.16: 23468937-23468937 on Build GRCh38
rs186192750 Chr.16: 23468815-23468815 on Build GRCh38
rs35347972 Chr.16: 23468973-23468973 on Build GRCh38
rs942300224 Chr.16: 23468847-23468847 on Build GRCh38
rs747271425 Chr.16: 23468848-23468849 on Build GRCh38
rs549533668 Chr.16: 23468829-23468829 on Build GRCh38
rs765564100 Chr.16: 23468983-23468983 on Build GRCh38
rs758682994 Chr.16: 23468856-23468856 on Build GRCh38
rs551451850 Chr.16: 23469108-23469108 on Build GRCh38
rs907702433 Chr.16: 23468920-23468920 on Build GRCh38
rs771198946 Chr.16: 23469002-23469003 on Build GRCh38
rs567557631 Chr.16: 23469228-23469228 on Build GRCh38
rs923214959 Chr.16: 23468835-23468835 on Build GRCh38
rs146396049 Chr.16: 23468891-23468891 on Build GRCh38
rs377173596 Chr.16: 23468981-23468981 on Build GRCh38
rs747635091 Chr.16: 23468896-23468896 on Build GRCh38
rs571917334 Chr.16: 23469122-23469122 on Build GRCh38
rs554442149 Chr.16: 23469185-23469185 on Build GRCh38
rs866903061 Chr.16: 23468875-23468875 on Build GRCh38
rs964362662 Chr.16: 23468808-23468808 on Build GRCh38
rs762699555 Chr.16: 23469154-23469154 on Build GRCh38
rs947737366 Chr.16: 23469233-23469233 on Build GRCh38
rs1018158559 Chr.16: 23469117-23469117 on Build GRCh38
rs976533050 Chr.16: 23469140-23469140 on Build GRCh38
rs762601848 Chr.16: 23468933-23468933 on Build GRCh38
rs904502862 Chr.16: 23469090-23469090 on Build GRCh38
rs746299571 Chr.16: 23468855-23468855 on Build GRCh38
rs780398494 Chr.16: 23468864-23468864 on Build GRCh38
rs1010498719 Chr.16: 23469126-23469126 on Build GRCh38
rs772738100 Chr.16: 23468930-23468930 on Build GRCh38
rs896102657 Chr.16: 23469118-23469118 on Build GRCh38
rs751048578 Chr.16: 23469207-23469207 on Build GRCh38
rs776158618 Chr.16: 23468893-23468893 on Build GRCh38
rs954594365 Chr.16: 23469238-23469238 on Build GRCh38
rs1007254975 Chr.16: 23469094-23469094 on Build GRCh38
rs965189018 Chr.16: 23469132-23469132 on Build GRCh38
rs200028402 Chr.16: 23468853-23468853 on Build GRCh38
rs891861172 Chr.16: 23468985-23468985 on Build GRCh38
rs1047589394 Chr.16: 23468872-23468872 on Build GRCh38
rs774242311 Chr.16: 23468942-23468942 on Build GRCh38
rs866120320 Chr.16: 23468972-23468972 on Build GRCh38

View Full Product Details