Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24154594-24155090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCB
Entrez Gene ID
5579
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
UniGene
Hs.460355
Gene Chromosome Location
Chr.16: 23835979-24220611 on Build GRCh38
Transcripts
NM_002738.6 NM_212535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5537611 c.1096G>A p.E366K Chr.16: 24154714-24154714 on Build GRCh38
3701066 c.1143T>C p.D381D Chr.16: 24154761-24154761 on Build GRCh38
6078788 c.1238_1239TG>AT p.M413N Chr.16: 24154856-24154857 on Build GRCh38
968891 c.1206C>T p.F402F Chr.16: 24154824-24154824 on Build GRCh38
1478688 c.1093G>A p.D365N Chr.16: 24154711-24154711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201855972 Chr.16: 24154856-24154856 on Build GRCh38
rs1039740393 Chr.16: 24154668-24154668 on Build GRCh38
rs141827066 Chr.16: 24154818-24154818 on Build GRCh38
rs929487920 Chr.16: 24154619-24154619 on Build GRCh38
rs267604474 Chr.16: 24154786-24154786 on Build GRCh38
rs745985737 Chr.16: 24154903-24154903 on Build GRCh38
rs749354695 Chr.16: 24154746-24154746 on Build GRCh38
rs74569287 Chr.16: 24155019-24155019 on Build GRCh38
rs141758720 Chr.16: 24154723-24154723 on Build GRCh38
rs750377270 Chr.16: 24154699-24154699 on Build GRCh38
rs758085883 Chr.16: 24154703-24154703 on Build GRCh38
rs868712349 Chr.16: 24154700-24154700 on Build GRCh38
rs766266252 Chr.16: 24154839-24154839 on Build GRCh38
rs757544207 Chr.16: 24154673-24154673 on Build GRCh38
rs200238437 Chr.16: 24154768-24154768 on Build GRCh38
rs763837460 Chr.16: 24154801-24154801 on Build GRCh38
rs772277246 Chr.16: 24154790-24154790 on Build GRCh38
rs432998 Chr.16: 24154809-24154809 on Build GRCh38
rs777912410 Chr.16: 24154860-24154860 on Build GRCh38
rs963422706 Chr.16: 24154921-24154921 on Build GRCh38
rs866023052 Chr.16: 24154765-24154765 on Build GRCh38
rs771422439 Chr.16: 24154643-24154643 on Build GRCh38
rs779072714 Chr.16: 24154897-24154897 on Build GRCh38
rs778704504 Chr.16: 24154680-24154680 on Build GRCh38
rs760382807 Chr.16: 24155045-24155045 on Build GRCh38
rs1002934805 Chr.16: 24154908-24154908 on Build GRCh38
rs763892410 Chr.16: 24154663-24154663 on Build GRCh38
rs764831474 Chr.16: 24154817-24154817 on Build GRCh38
rs368085619 Chr.16: 24154639-24154639 on Build GRCh38
rs537453687 Chr.16: 24155015-24155015 on Build GRCh38
rs375177555 Chr.16: 24154637-24154637 on Build GRCh38
rs774978908 Chr.16: 24154646-24154646 on Build GRCh38
rs774387524 Chr.16: 24154870-24154871 on Build GRCh38
rs756297068 Chr.16: 24154716-24154716 on Build GRCh38
rs760425879 Chr.16: 24154658-24154658 on Build GRCh38
rs566340714 Chr.16: 24154920-24154920 on Build GRCh38
rs764476534 Chr.16: 24154629-24154629 on Build GRCh38
rs757246331 Chr.16: 24154878-24154878 on Build GRCh38
rs548921325 Chr.16: 24154951-24154951 on Build GRCh38
rs772094439 Chr.16: 24155030-24155030 on Build GRCh38
rs759548439 Chr.16: 24154894-24154894 on Build GRCh38
rs761166216 Chr.16: 24154795-24154795 on Build GRCh38
rs147005947 Chr.16: 24155002-24155002 on Build GRCh38
rs776487939 Chr.16: 24154803-24154803 on Build GRCh38
rs540401484 Chr.16: 24154934-24154934 on Build GRCh38
rs1001164834 Chr.16: 24154713-24154713 on Build GRCh38
rs779917484 Chr.16: 24154707-24154707 on Build GRCh38
rs746420133 Chr.16: 24154780-24154780 on Build GRCh38
rs372069305 Chr.16: 24154649-24154649 on Build GRCh38
rs148312044 Chr.16: 24154709-24154709 on Build GRCh38
rs374599996 Chr.16: 24154877-24154877 on Build GRCh38
rs369508594 Chr.16: 24154855-24154855 on Build GRCh38
rs375689115 Chr.16: 24154674-24154674 on Build GRCh38
rs910340038 Chr.16: 24155065-24155065 on Build GRCh38
rs753371346 Chr.16: 24154664-24154664 on Build GRCh38
rs943763817 Chr.16: 24155037-24155037 on Build GRCh38
rs34973008 Chr.16: 24154681-24154682 on Build GRCh38
rs184106438 Chr.16: 24154654-24154654 on Build GRCh38
rs956137812 Chr.16: 24154924-24154924 on Build GRCh38
rs543064850 Chr.16: 24154767-24154767 on Build GRCh38
rs549019167 Chr.16: 24155031-24155031 on Build GRCh38
rs202161442 Chr.16: 24154722-24154722 on Build GRCh38
rs774656581 Chr.16: 24154773-24154773 on Build GRCh38
rs775901586 Chr.16: 24154793-24154793 on Build GRCh38
rs1031772683 Chr.16: 24154912-24154912 on Build GRCh38
rs1034073456 Chr.16: 24154910-24154910 on Build GRCh38

View Full Product Details