Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 26137084-26137581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS3ST4
Entrez Gene ID
9951
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 4
Gene Aliases
3-OST-4, 30ST4, 3OST4, h3-OST-4
UniGene
Hs.655275
Gene Chromosome Location
Chr.16: 25692026-26137688 on Build GRCh38
Transcripts
NM_006040.2

SNP Detail

SNP ID Chromosome Location
rs975566925 Chr.16: 26137517-26137517 on Build GRCh38
rs985088047 Chr.16: 26137547-26137547 on Build GRCh38
rs947025070 Chr.16: 26137206-26137206 on Build GRCh38
rs533331029 Chr.16: 26137109-26137109 on Build GRCh38
rs965623930 Chr.16: 26137215-26137215 on Build GRCh38
rs950498972 Chr.16: 26137163-26137163 on Build GRCh38
rs1028604276 Chr.16: 26137534-26137534 on Build GRCh38
rs142186247 Chr.16: 26137146-26137146 on Build GRCh38
rs1054998096 Chr.16: 26137350-26137350 on Build GRCh38
rs1012237189 Chr.16: 26137375-26137376 on Build GRCh38
rs544330128 Chr.16: 26137423-26137423 on Build GRCh38
rs560629149 Chr.16: 26137170-26137170 on Build GRCh38
rs748042501 Chr.16: 26137443-26137443 on Build GRCh38
rs912690739 Chr.16: 26137208-26137208 on Build GRCh38
rs956868806 Chr.16: 26137470-26137470 on Build GRCh38
rs886200264 Chr.16: 26137441-26137441 on Build GRCh38
rs187186466 Chr.16: 26137199-26137199 on Build GRCh38
rs192115142 Chr.16: 26137213-26137213 on Build GRCh38
rs989464190 Chr.16: 26137299-26137299 on Build GRCh38
rs958253343 Chr.16: 26137539-26137539 on Build GRCh38
rs148325442 Chr.16: 26137413-26137413 on Build GRCh38
rs180868679 Chr.16: 26137172-26137172 on Build GRCh38
rs994482924 Chr.16: 26137387-26137387 on Build GRCh38
rs914008581 Chr.16: 26137309-26137309 on Build GRCh38
rs548748870 Chr.16: 26137175-26137175 on Build GRCh38
rs868775292 Chr.16: 26137386-26137386 on Build GRCh38
rs749011903 Chr.16: 26137233-26137233 on Build GRCh38
rs867903504 Chr.16: 26137125-26137125 on Build GRCh38
rs533993158 Chr.16: 26137328-26137328 on Build GRCh38
rs906318434 Chr.16: 26137339-26137339 on Build GRCh38
rs989509008 Chr.16: 26137551-26137551 on Build GRCh38
rs532220699 Chr.16: 26137326-26137326 on Build GRCh38
rs183084739 Chr.16: 26137223-26137223 on Build GRCh38
rs555521311 Chr.16: 26137398-26137398 on Build GRCh38
rs1026090361 Chr.16: 26137133-26137133 on Build GRCh38
rs921509064 Chr.16: 26137227-26137227 on Build GRCh38
rs141492623 Chr.16: 26137494-26137494 on Build GRCh38
rs190550569 Chr.16: 26137367-26137367 on Build GRCh38
rs1031503944 Chr.16: 26137415-26137415 on Build GRCh38
rs1041201206 Chr.16: 26137301-26137301 on Build GRCh38
rs975702850 Chr.16: 26137221-26137221 on Build GRCh38
rs985120556 Chr.16: 26137211-26137211 on Build GRCh38
rs1022531913 Chr.16: 26137525-26137525 on Build GRCh38
rs1018695814 Chr.16: 26137489-26137489 on Build GRCh38
rs943825937 Chr.16: 26137281-26137281 on Build GRCh38
rs185978143 Chr.16: 26137340-26137340 on Build GRCh38
rs368828901 Chr.16: 26137353-26137353 on Build GRCh38

View Full Product Details