Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28977817-28978310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPNS1
Entrez Gene ID
83985
Gene Name
sphingolipid transporter 1 (putative)
Gene Aliases
HSpin1, LAT, PP2030, SPIN1, SPINL, nrs
UniGene
Hs.617449
Gene Chromosome Location
Chr.16: 28974767-28984769 on Build GRCh38
Transcripts
NM_001142448.1 NM_001142449.1 NM_001142450.1 NM_001142451.1 NM_032038.2 XM_006721096.3 XM_017023757.1 XM_017023758.1 XM_017023759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3110516 c.372G>A p.R124R Chr.16: 28977972-28977972 on Build GRCh38
3110514 c.308-3T>C p.? Chr.16: 28977905-28977905 on Build GRCh38
6311214 c.388G>T p.G130W Chr.16: 28977988-28977988 on Build GRCh38
7131122 c.370C>T p.R124W Chr.16: 28977970-28977970 on Build GRCh38
3110517 c.378T>C p.Y126Y Chr.16: 28977978-28977978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753871768 Chr.16: 28977903-28977903 on Build GRCh38
rs527289754 Chr.16: 28977870-28977870 on Build GRCh38
rs775213002 Chr.16: 28978278-28978278 on Build GRCh38
rs770869551 Chr.16: 28978228-28978228 on Build GRCh38
rs905893741 Chr.16: 28978251-28978251 on Build GRCh38
rs1035920735 Chr.16: 28977974-28977974 on Build GRCh38
rs1002817260 Chr.16: 28977917-28977917 on Build GRCh38
rs764095000 Chr.16: 28977902-28977902 on Build GRCh38
rs776668343 Chr.16: 28978061-28978061 on Build GRCh38
rs778204826 Chr.16: 28977987-28977987 on Build GRCh38
rs756519960 Chr.16: 28977975-28977975 on Build GRCh38
rs370939688 Chr.16: 28978029-28978029 on Build GRCh38
rs946988387 Chr.16: 28978214-28978214 on Build GRCh38
rs911462002 Chr.16: 28978199-28978199 on Build GRCh38
rs943603984 Chr.16: 28978083-28978083 on Build GRCh38
rs780615062 Chr.16: 28978231-28978231 on Build GRCh38
rs985906565 Chr.16: 28978178-28978178 on Build GRCh38
rs775618063 Chr.16: 28978056-28978056 on Build GRCh38
rs759587868 Chr.16: 28978286-28978286 on Build GRCh38
rs756431623 Chr.16: 28978095-28978095 on Build GRCh38
rs368432030 Chr.16: 28978092-28978092 on Build GRCh38
rs749975785 Chr.16: 28977934-28977934 on Build GRCh38
rs976332027 Chr.16: 28978097-28978097 on Build GRCh38
rs368155910 Chr.16: 28978080-28978080 on Build GRCh38
rs777107125 Chr.16: 28978073-28978074 on Build GRCh38
rs896156837 Chr.16: 28977848-28977848 on Build GRCh38
rs764869232 Chr.16: 28977931-28977931 on Build GRCh38
rs187109604 Chr.16: 28978169-28978169 on Build GRCh38
rs749827751 Chr.16: 28977988-28977988 on Build GRCh38
rs934001351 Chr.16: 28978177-28978177 on Build GRCh38
rs201559262 Chr.16: 28978075-28978075 on Build GRCh38
rs770932119 Chr.16: 28978017-28978017 on Build GRCh38
rs1043871621 Chr.16: 28978222-28978222 on Build GRCh38
rs761844642 Chr.16: 28978073-28978073 on Build GRCh38
rs374695118 Chr.16: 28978060-28978060 on Build GRCh38
rs778981533 Chr.16: 28978023-28978023 on Build GRCh38
rs968471587 Chr.16: 28978038-28978038 on Build GRCh38
rs749934007 Chr.16: 28978081-28978081 on Build GRCh38
rs972793064 Chr.16: 28978143-28978143 on Build GRCh38
rs761354214 Chr.16: 28977919-28977919 on Build GRCh38
rs745996135 Chr.16: 28978039-28978039 on Build GRCh38
rs758082098 Chr.16: 28977939-28977939 on Build GRCh38
rs201026483 Chr.16: 28978058-28978058 on Build GRCh38
rs371536107 Chr.16: 28978082-28978082 on Build GRCh38
rs769633424 Chr.16: 28978244-28978244 on Build GRCh38
rs1035591210 Chr.16: 28978255-28978255 on Build GRCh38
rs753101006 Chr.16: 28977965-28977965 on Build GRCh38
rs374161103 Chr.16: 28977951-28977951 on Build GRCh38
rs779631079 Chr.16: 28977942-28977942 on Build GRCh38
rs759810098 Chr.16: 28977863-28977863 on Build GRCh38
rs772339108 Chr.16: 28978047-28978047 on Build GRCh38
rs923535183 Chr.16: 28978162-28978162 on Build GRCh38
rs867421690 Chr.16: 28978005-28978005 on Build GRCh38
rs745358323 Chr.16: 28978236-28978236 on Build GRCh38
rs566246453 Chr.16: 28977901-28977901 on Build GRCh38
rs200363122 Chr.16: 28977881-28977881 on Build GRCh38
rs775345211 Chr.16: 28977874-28977874 on Build GRCh38
rs145845819 Chr.16: 28978254-28978254 on Build GRCh38
rs766023300 Chr.16: 28978084-28978084 on Build GRCh38
rs556306732 Chr.16: 28978216-28978216 on Build GRCh38

View Full Product Details