Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30553333-30553847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF764
Entrez Gene ID
92595
Gene Name
zinc finger protein 764
Gene Aliases
-
UniGene
Hs.132227
Gene Chromosome Location
Chr.16: 30553764-30558321 on Build GRCh38
Transcripts
NM_001172679.1 NM_033410.3

SNP Detail

SNP ID Chromosome Location
rs777495849 Chr.16: 30553600-30553600 on Build GRCh38
rs376516845 Chr.16: 30553610-30553610 on Build GRCh38
rs529336131 Chr.16: 30553684-30553684 on Build GRCh38
rs966943898 Chr.16: 30553589-30553589 on Build GRCh38
rs893696670 Chr.16: 30553361-30553362 on Build GRCh38
rs1010606873 Chr.16: 30553366-30553366 on Build GRCh38
rs565869747 Chr.16: 30553635-30553635 on Build GRCh38
rs116994224 Chr.16: 30553601-30553601 on Build GRCh38
rs187289162 Chr.16: 30553595-30553595 on Build GRCh38
rs778700774 Chr.16: 30553789-30553790 on Build GRCh38
rs886374435 Chr.16: 30553364-30553364 on Build GRCh38
rs901709576 Chr.16: 30553405-30553405 on Build GRCh38
rs983638761 Chr.16: 30553524-30553524 on Build GRCh38
rs1050013559 Chr.16: 30553747-30553747 on Build GRCh38
rs1034731869 Chr.16: 30553468-30553468 on Build GRCh38
rs543044105 Chr.16: 30553544-30553544 on Build GRCh38
rs551590997 Chr.16: 30553675-30553675 on Build GRCh38
rs907758853 Chr.16: 30553739-30553739 on Build GRCh38
rs930635610 Chr.16: 30553695-30553695 on Build GRCh38
rs752571493 Chr.16: 30553439-30553439 on Build GRCh38
rs985199480 Chr.16: 30553716-30553716 on Build GRCh38
rs578077849 Chr.16: 30553384-30553384 on Build GRCh38
rs544589696 Chr.16: 30553795-30553795 on Build GRCh38
rs990905526 Chr.16: 30553631-30553631 on Build GRCh38
rs991392281 Chr.16: 30553481-30553481 on Build GRCh38
rs1040353517 Chr.16: 30553816-30553816 on Build GRCh38
rs753890009 Chr.16: 30553358-30553358 on Build GRCh38
rs754802503 Chr.16: 30553416-30553416 on Build GRCh38
rs765167860 Chr.16: 30553434-30553434 on Build GRCh38
rs915052528 Chr.16: 30553777-30553777 on Build GRCh38
rs946636129 Chr.16: 30553792-30553792 on Build GRCh38
rs922008681 Chr.16: 30553649-30553649 on Build GRCh38
rs562355862 Chr.16: 30553578-30553578 on Build GRCh38
rs532940702 Chr.16: 30553657-30553657 on Build GRCh38
rs11867032 Chr.16: 30553471-30553471 on Build GRCh38
rs115627122 Chr.16: 30553498-30553498 on Build GRCh38
rs78345703 Chr.16: 30553592-30553592 on Build GRCh38
rs940464692 Chr.16: 30553374-30553374 on Build GRCh38
rs1027927947 Chr.16: 30553485-30553485 on Build GRCh38
rs151316158 Chr.16: 30553433-30553433 on Build GRCh38
rs866990277 Chr.16: 30553706-30553706 on Build GRCh38

View Full Product Details