Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66885878-66886386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDP2
Entrez Gene ID
57546
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 2
Gene Aliases
PPM2B, PPM2C2
UniGene
Hs.632214
Gene Chromosome Location
Chr.16: 66880397-66891101 on Build GRCh38
Transcripts
NM_020786.2 XM_005256064.4 XM_005256066.4 XM_005256067.4 XM_005256068.4 XM_017023506.1

SNP Detail

SNP ID Chromosome Location
rs895257431 Chr.16: 66886309-66886309 on Build GRCh38
rs914240464 Chr.16: 66886223-66886223 on Build GRCh38
rs962641272 Chr.16: 66886008-66886008 on Build GRCh38
rs986625127 Chr.16: 66886059-66886059 on Build GRCh38
rs965671724 Chr.16: 66886172-66886172 on Build GRCh38
rs990810119 Chr.16: 66885972-66885972 on Build GRCh38
rs894476771 Chr.16: 66885959-66885959 on Build GRCh38
rs1035518059 Chr.16: 66885935-66885935 on Build GRCh38
rs1007720705 Chr.16: 66886124-66886124 on Build GRCh38
rs564255767 Chr.16: 66886025-66886025 on Build GRCh38
rs1016870730 Chr.16: 66885996-66885996 on Build GRCh38
rs918475093 Chr.16: 66886034-66886034 on Build GRCh38
rs998691957 Chr.16: 66886085-66886085 on Build GRCh38
rs751132627 Chr.16: 66885903-66885903 on Build GRCh38
rs555661367 Chr.16: 66886003-66886003 on Build GRCh38
rs756443244 Chr.16: 66885911-66885911 on Build GRCh38
rs1054147414 Chr.16: 66886217-66886217 on Build GRCh38
rs557209341 Chr.16: 66886302-66886302 on Build GRCh38
rs184521572 Chr.16: 66886316-66886316 on Build GRCh38
rs544385087 Chr.16: 66886022-66886022 on Build GRCh38
rs1045663372 Chr.16: 66886004-66886004 on Build GRCh38
rs201365694 Chr.16: 66885910-66885910 on Build GRCh38
rs1003681399 Chr.16: 66885929-66885929 on Build GRCh38
rs1043041637 Chr.16: 66886244-66886244 on Build GRCh38
rs1045738849 Chr.16: 66886319-66886319 on Build GRCh38
rs772772137 Chr.16: 66885970-66885970 on Build GRCh38
rs903108980 Chr.16: 66886331-66886331 on Build GRCh38
rs1056543944 Chr.16: 66886306-66886306 on Build GRCh38
rs17767794 Chr.16: 66886026-66886026 on Build GRCh38
rs1017296172 Chr.16: 66886147-66886147 on Build GRCh38
rs758081116 Chr.16: 66886087-66886087 on Build GRCh38
rs982976123 Chr.16: 66886277-66886277 on Build GRCh38
rs1033225408 Chr.16: 66886098-66886098 on Build GRCh38
rs1004138791 Chr.16: 66886305-66886305 on Build GRCh38
rs902725215 Chr.16: 66886299-66886299 on Build GRCh38
rs575530494 Chr.16: 66886013-66886013 on Build GRCh38
rs915943341 Chr.16: 66886311-66886311 on Build GRCh38
rs777534873 Chr.16: 66886155-66886155 on Build GRCh38
rs1055832224 Chr.16: 66885922-66885922 on Build GRCh38
rs942415908 Chr.16: 66886182-66886182 on Build GRCh38
rs955384653 Chr.16: 66886047-66886047 on Build GRCh38
rs951590238 Chr.16: 66886275-66886275 on Build GRCh38
rs756831474 Chr.16: 66885905-66885905 on Build GRCh38
rs892922188 Chr.16: 66886115-66886115 on Build GRCh38
rs764594294 Chr.16: 66885917-66885917 on Build GRCh38
rs910989621 Chr.16: 66886160-66886160 on Build GRCh38
rs991962654 Chr.16: 66886307-66886307 on Build GRCh38
rs193235873 Chr.16: 66886235-66886235 on Build GRCh38

View Full Product Details