Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87406154-87406649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCCHC14
Entrez Gene ID
23174
Gene Name
zinc finger CCHC-type containing 14
Gene Aliases
BDG-29, BDG29
UniGene
Hs.156231
Gene Chromosome Location
Chr.16: 87406246-87492948 on Build GRCh38
Transcripts
NM_015144.2

SNP Detail

SNP ID Chromosome Location
rs542747332 Chr.16: 87406526-87406526 on Build GRCh38
rs960697533 Chr.16: 87406222-87406222 on Build GRCh38
rs190157490 Chr.16: 87406207-87406207 on Build GRCh38
rs975097823 Chr.16: 87406587-87406587 on Build GRCh38
rs988072385 Chr.16: 87406224-87406224 on Build GRCh38
rs1005820555 Chr.16: 87406369-87406369 on Build GRCh38
rs1040151247 Chr.16: 87406258-87406258 on Build GRCh38
rs898689606 Chr.16: 87406474-87406474 on Build GRCh38
rs945773977 Chr.16: 87406280-87406280 on Build GRCh38
rs906839864 Chr.16: 87406353-87406353 on Build GRCh38
rs1025795200 Chr.16: 87406491-87406491 on Build GRCh38
rs1017028823 Chr.16: 87406398-87406398 on Build GRCh38
rs1028896255 Chr.16: 87406588-87406588 on Build GRCh38
rs557303043 Chr.16: 87406186-87406186 on Build GRCh38
rs1035105904 Chr.16: 87406437-87406440 on Build GRCh38
rs544711096 Chr.16: 87406457-87406457 on Build GRCh38
rs990015566 Chr.16: 87406603-87406603 on Build GRCh38
rs781675357 Chr.16: 87406399-87406399 on Build GRCh38
rs554791108 Chr.16: 87406218-87406218 on Build GRCh38
rs76663018 Chr.16: 87406621-87406621 on Build GRCh38
rs887203957 Chr.16: 87406501-87406501 on Build GRCh38
rs372573627 Chr.16: 87406539-87406539 on Build GRCh38
rs1056043836 Chr.16: 87406471-87406471 on Build GRCh38
rs78333183 Chr.16: 87406602-87406602 on Build GRCh38
rs1045383264 Chr.16: 87406330-87406330 on Build GRCh38
rs990332353 Chr.16: 87406257-87406257 on Build GRCh38
rs913035628 Chr.16: 87406613-87406613 on Build GRCh38
rs965959826 Chr.16: 87406209-87406209 on Build GRCh38
rs1020849036 Chr.16: 87406489-87406489 on Build GRCh38
rs979354082 Chr.16: 87406533-87406533 on Build GRCh38
rs563285778 Chr.16: 87406462-87406462 on Build GRCh38
rs577441616 Chr.16: 87406423-87406423 on Build GRCh38
rs530495518 Chr.16: 87406279-87406279 on Build GRCh38
rs887380768 Chr.16: 87406352-87406352 on Build GRCh38
rs140953251 Chr.16: 87406518-87406518 on Build GRCh38
rs923045913 Chr.16: 87406616-87406617 on Build GRCh38
rs540390766 Chr.16: 87406281-87406281 on Build GRCh38
rs967106527 Chr.16: 87406617-87406617 on Build GRCh38
rs921799606 Chr.16: 87406249-87406249 on Build GRCh38
rs528311176 Chr.16: 87406578-87406578 on Build GRCh38
rs980835212 Chr.16: 87406188-87406188 on Build GRCh38
rs913841260 Chr.16: 87406232-87406232 on Build GRCh38
rs1047297833 Chr.16: 87406309-87406309 on Build GRCh38
rs573107274 Chr.16: 87406256-87406256 on Build GRCh38
rs995671075 Chr.16: 87406477-87406477 on Build GRCh38
rs1002203906 Chr.16: 87406401-87406401 on Build GRCh38
rs202052816 Chr.16: 87406611-87406612 on Build GRCh38
rs575686246 Chr.16: 87406203-87406203 on Build GRCh38
rs558842515 Chr.16: 87406356-87406356 on Build GRCh38
rs986818029 Chr.16: 87406195-87406195 on Build GRCh38
rs910350858 Chr.16: 87406552-87406552 on Build GRCh38
rs563454071 Chr.16: 87406192-87406192 on Build GRCh38
rs964036992 Chr.16: 87406563-87406563 on Build GRCh38
rs974648205 Chr.16: 87406236-87406236 on Build GRCh38

View Full Product Details