Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3471309-3471823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA22
Entrez Gene ID
84690
Gene Name
spermatogenesis associated 22
Gene Aliases
NYD-SP20, NYDSP20
UniGene
Hs.351068
Gene Chromosome Location
Chr.17: 3440011-3513852 on Build GRCh38
Transcripts
NM_001170695.1 NM_001170696.1 NM_001170697.1 NM_001170698.1 NM_001170699.1 NM_001321336.1 NM_001321337.1 NM_032598.4

SNP Detail

SNP ID Chromosome Location
rs759553659 Chr.17: 3471586-3471586 on Build GRCh38
rs969248396 Chr.17: 3471615-3471615 on Build GRCh38
rs539697214 Chr.17: 3471765-3471765 on Build GRCh38
rs373027923 Chr.17: 3471392-3471392 on Build GRCh38
rs1052017343 Chr.17: 3471768-3471768 on Build GRCh38
rs549774289 Chr.17: 3471632-3471632 on Build GRCh38
rs890708524 Chr.17: 3471369-3471369 on Build GRCh38
rs900889728 Chr.17: 3471759-3471759 on Build GRCh38
rs764552771 Chr.17: 3471726-3471726 on Build GRCh38
rs866366119 Chr.17: 3471757-3471757 on Build GRCh38
rs888199187 Chr.17: 3471518-3471518 on Build GRCh38
rs575756416 Chr.17: 3471611-3471611 on Build GRCh38
rs980546691 Chr.17: 3471433-3471433 on Build GRCh38
rs376775846 Chr.17: 3471384-3471384 on Build GRCh38
rs1051980355 Chr.17: 3471356-3471358 on Build GRCh38
rs1010703629 Chr.17: 3471610-3471610 on Build GRCh38
rs569670848 Chr.17: 3471635-3471635 on Build GRCh38
rs917808722 Chr.17: 3471405-3471405 on Build GRCh38
rs745795548 Chr.17: 3471383-3471383 on Build GRCh38
rs78232924 Chr.17: 3471510-3471510 on Build GRCh38
rs146343382 Chr.17: 3471670-3471670 on Build GRCh38
rs762159534 Chr.17: 3471417-3471417 on Build GRCh38
rs904727537 Chr.17: 3471548-3471548 on Build GRCh38
rs894560060 Chr.17: 3471559-3471559 on Build GRCh38
rs1048553668 Chr.17: 3471517-3471517 on Build GRCh38
rs900818772 Chr.17: 3471555-3471555 on Build GRCh38
rs752159395 Chr.17: 3471774-3471774 on Build GRCh38
rs953146591 Chr.17: 3471782-3471782 on Build GRCh38
rs1039442546 Chr.17: 3471750-3471750 on Build GRCh38
rs968799103 Chr.17: 3471431-3471431 on Build GRCh38
rs933661814 Chr.17: 3471760-3471760 on Build GRCh38
rs1010777343 Chr.17: 3471783-3471783 on Build GRCh38
rs76430945 Chr.17: 3471391-3471391 on Build GRCh38
rs150770000 Chr.17: 3471679-3471680 on Build GRCh38
rs904779843 Chr.17: 3471681-3471681 on Build GRCh38
rs905014575 Chr.17: 3471802-3471802 on Build GRCh38
rs781749538 Chr.17: 3471519-3471522 on Build GRCh38
rs770011187 Chr.17: 3471475-3471475 on Build GRCh38
rs1030815124 Chr.17: 3471394-3471394 on Build GRCh38
rs936121393 Chr.17: 3471698-3471698 on Build GRCh38
rs375422076 Chr.17: 3471541-3471542 on Build GRCh38
rs1030895597 Chr.17: 3471584-3471584 on Build GRCh38
rs114929279 Chr.17: 3471677-3471677 on Build GRCh38
rs533767147 Chr.17: 3471672-3471672 on Build GRCh38
rs577072105 Chr.17: 3471743-3471743 on Build GRCh38
rs941774255 Chr.17: 3471487-3471487 on Build GRCh38
rs963276726 Chr.17: 3471772-3471772 on Build GRCh38
rs989250341 Chr.17: 3471414-3471414 on Build GRCh38
rs992802452 Chr.17: 3471655-3471655 on Build GRCh38
rs545386504 Chr.17: 3471551-3471551 on Build GRCh38
rs549046664 Chr.17: 3471669-3471669 on Build GRCh38
rs116582520 Chr.17: 3471437-3471437 on Build GRCh38
rs149789275 Chr.17: 3471519-3471520 on Build GRCh38
rs892345123 Chr.17: 3471598-3471598 on Build GRCh38
rs910047633 Chr.17: 3471739-3471739 on Build GRCh38
rs146848166 Chr.17: 3471434-3471434 on Build GRCh38
rs767057008 Chr.17: 3471749-3471749 on Build GRCh38
rs921656082 Chr.17: 3471643-3471643 on Build GRCh38
rs973189671 Chr.17: 3471773-3471773 on Build GRCh38
rs145316936 Chr.17: 3471401-3471401 on Build GRCh38
rs386794623 Chr.17: 3471677-3471680 on Build GRCh38
rs770293221 Chr.17: 3471597-3471597 on Build GRCh38
rs1014705104 Chr.17: 3471612-3471612 on Build GRCh38
rs148969784 Chr.17: 3471507-3471507 on Build GRCh38
rs1006968763 Chr.17: 3471341-3471341 on Build GRCh38
rs141856026 Chr.17: 3471622-3471622 on Build GRCh38
rs62071294 Chr.17: 3471801-3471801 on Build GRCh38
rs887144114 Chr.17: 3471744-3471744 on Build GRCh38
rs1055987546 Chr.17: 3471728-3471728 on Build GRCh38
rs980193912 Chr.17: 3471621-3471621 on Build GRCh38
rs1010377281 Chr.17: 3471373-3471373 on Build GRCh38
rs142784503 Chr.17: 3471524-3471525 on Build GRCh38
rs984320784 Chr.17: 3471729-3471729 on Build GRCh38
rs148166787 Chr.17: 3471550-3471550 on Build GRCh38
rs1041897694 Chr.17: 3471522-3471522 on Build GRCh38
rs535603400 Chr.17: 3471653-3471653 on Build GRCh38
rs563678248 Chr.17: 3471562-3471562 on Build GRCh38
rs911559893 Chr.17: 3471800-3471800 on Build GRCh38
rs572926140 Chr.17: 3471729-3471729 on Build GRCh38
rs553170539 Chr.17: 3471469-3471469 on Build GRCh38

View Full Product Details