Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3471718-3472224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA22
Entrez Gene ID
84690
Gene Name
spermatogenesis associated 22
Gene Aliases
NYD-SP20, NYDSP20
UniGene
Hs.351068
Gene Chromosome Location
Chr.17: 3440011-3513852 on Build GRCh38
Transcripts
NM_001170695.1 NM_001170696.1 NM_001170697.1 NM_001170698.1 NM_001170699.1 NM_001321336.1 NM_001321337.1 NM_032598.4

SNP Detail

SNP ID Chromosome Location
rs1002420119 Chr.17: 3471821-3471821 on Build GRCh38
rs964611347 Chr.17: 3472094-3472094 on Build GRCh38
rs73305600 Chr.17: 3471872-3471872 on Build GRCh38
rs539697214 Chr.17: 3471765-3471765 on Build GRCh38
rs1052017343 Chr.17: 3471768-3471768 on Build GRCh38
rs749911341 Chr.17: 3471905-3471905 on Build GRCh38
rs1004139923 Chr.17: 3471844-3471844 on Build GRCh38
rs900889728 Chr.17: 3471759-3471759 on Build GRCh38
rs35217820 Chr.17: 3472054-3472055 on Build GRCh38
rs1017204636 Chr.17: 3472070-3472070 on Build GRCh38
rs1043664510 Chr.17: 3471911-3471911 on Build GRCh38
rs143134811 Chr.17: 3471852-3471852 on Build GRCh38
rs866366119 Chr.17: 3471757-3471757 on Build GRCh38
rs1005527290 Chr.17: 3472053-3472053 on Build GRCh38
rs977093831 Chr.17: 3471813-3471813 on Build GRCh38
rs113047175 Chr.17: 3472163-3472163 on Build GRCh38
rs386794624 Chr.17: 3471837-3471838 on Build GRCh38
rs926165717 Chr.17: 3471825-3471825 on Build GRCh38
rs745848398 Chr.17: 3471952-3471952 on Build GRCh38
rs539784553 Chr.17: 3472080-3472080 on Build GRCh38
rs960358512 Chr.17: 3471835-3471835 on Build GRCh38
rs908608548 Chr.17: 3472101-3472101 on Build GRCh38
rs752159395 Chr.17: 3471774-3471774 on Build GRCh38
rs953146591 Chr.17: 3471782-3471782 on Build GRCh38
rs1039442546 Chr.17: 3471750-3471750 on Build GRCh38
rs1008734493 Chr.17: 3471876-3471876 on Build GRCh38
rs1052089988 Chr.17: 3471898-3471898 on Build GRCh38
rs933661814 Chr.17: 3471760-3471760 on Build GRCh38
rs905087548 Chr.17: 3471918-3471918 on Build GRCh38
rs1010777343 Chr.17: 3471783-3471783 on Build GRCh38
rs905014575 Chr.17: 3471802-3471802 on Build GRCh38
rs539842584 Chr.17: 3471829-3471829 on Build GRCh38
rs1028821036 Chr.17: 3471855-3471855 on Build GRCh38
rs895935969 Chr.17: 3472015-3472015 on Build GRCh38
rs138039883 Chr.17: 3472013-3472013 on Build GRCh38
rs962357285 Chr.17: 3471837-3471838 on Build GRCh38
rs139000849 Chr.17: 3471845-3471845 on Build GRCh38
rs577072105 Chr.17: 3471743-3471743 on Build GRCh38
rs62071295 Chr.17: 3471932-3471932 on Build GRCh38
rs149507854 Chr.17: 3472052-3472052 on Build GRCh38
rs1025674107 Chr.17: 3472031-3472031 on Build GRCh38
rs963276726 Chr.17: 3471772-3471772 on Build GRCh38
rs556481617 Chr.17: 3472116-3472116 on Build GRCh38
rs529225782 Chr.17: 3471902-3471902 on Build GRCh38
rs780831265 Chr.17: 3472004-3472004 on Build GRCh38
rs1025641398 Chr.17: 3471839-3471839 on Build GRCh38
rs115184036 Chr.17: 3471838-3471838 on Build GRCh38
rs767057008 Chr.17: 3471749-3471749 on Build GRCh38
rs973189671 Chr.17: 3471773-3471773 on Build GRCh38
rs975948407 Chr.17: 3471860-3471860 on Build GRCh38
rs73305597 Chr.17: 3471827-3471827 on Build GRCh38
rs62071294 Chr.17: 3471801-3471801 on Build GRCh38
rs887144114 Chr.17: 3471744-3471744 on Build GRCh38
rs147471461 Chr.17: 3471912-3471912 on Build GRCh38
rs549329398 Chr.17: 3471924-3471924 on Build GRCh38
rs957600632 Chr.17: 3471985-3471985 on Build GRCh38
rs781446100 Chr.17: 3472198-3472198 on Build GRCh38
rs940134664 Chr.17: 3472140-3472140 on Build GRCh38
rs543706564 Chr.17: 3471887-3471887 on Build GRCh38
rs916166730 Chr.17: 3472023-3472023 on Build GRCh38
rs910069637 Chr.17: 3471846-3471846 on Build GRCh38
rs182723905 Chr.17: 3471901-3471901 on Build GRCh38
rs533451353 Chr.17: 3471886-3471886 on Build GRCh38
rs76698640 Chr.17: 3471837-3471837 on Build GRCh38
rs1014344109 Chr.17: 3472028-3472028 on Build GRCh38
rs386794625 Chr.17: 3471837-3471840 on Build GRCh38
rs911559893 Chr.17: 3471800-3471800 on Build GRCh38
rs951647371 Chr.17: 3471840-3471840 on Build GRCh38
rs73305602 Chr.17: 3472004-3472004 on Build GRCh38
rs975641786 Chr.17: 3472186-3472186 on Build GRCh38

View Full Product Details