Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5210526-5211028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100130950
Entrez Gene ID
100130950
Gene Name
uncharacterized LOC100130950
Gene Aliases
-
UniGene
Hs.668646
Gene Chromosome Location
Chr.17: 5192084-5235636 on Build GRCh38
Transcripts
NR_034082.1 NM_001271842.1 NM_207103.3

Gene Symbol
SCIMP
Entrez Gene ID
388325
Gene Name
SLP adaptor and CSK interacting membrane protein
Gene Aliases
C17orf87, UNQ5783
UniGene
Hs.462080
Gene Chromosome Location
Chr.17: 5208920-5234860 on Build GRCh38
Transcripts
NR_034082.1 NM_001271842.1 NM_207103.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
79562 c.366A>C p.P122P Chr.17: 5210873-5210873 on Build GRCh38
5538565 c.403G>A p.E135K Chr.17: 5210836-5210836 on Build GRCh38
5733705 c.321C>A p.Y107* Chr.17: 5210918-5210918 on Build GRCh38
7388796 c.342A>T p.K114N Chr.17: 5210897-5210897 on Build GRCh38
3519653 c.377A>C p.E126A Chr.17: 5210862-5210862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368421148 Chr.17: 5210963-5210963 on Build GRCh38
rs372375934 Chr.17: 5210931-5210931 on Build GRCh38
rs913435955 Chr.17: 5210920-5210920 on Build GRCh38
rs761274771 Chr.17: 5210956-5210956 on Build GRCh38
rs746896578 Chr.17: 5210755-5210755 on Build GRCh38
rs780120054 Chr.17: 5211004-5211004 on Build GRCh38
rs367619956 Chr.17: 5210900-5210900 on Build GRCh38
rs759825932 Chr.17: 5210927-5210928 on Build GRCh38
rs911815885 Chr.17: 5210608-5210608 on Build GRCh38
rs750084422 Chr.17: 5210887-5210889 on Build GRCh38
rs201474608 Chr.17: 5210982-5210982 on Build GRCh38
rs754913591 Chr.17: 5210896-5210896 on Build GRCh38
rs1049020767 Chr.17: 5210984-5210984 on Build GRCh38
rs1040653784 Chr.17: 5210950-5210950 on Build GRCh38
rs1055018488 Chr.17: 5210667-5210667 on Build GRCh38
rs920471182 Chr.17: 5210575-5210575 on Build GRCh38
rs747978306 Chr.17: 5210796-5210796 on Build GRCh38
rs770409940 Chr.17: 5210831-5210831 on Build GRCh38
rs149560316 Chr.17: 5210590-5210590 on Build GRCh38
rs887867043 Chr.17: 5210998-5210998 on Build GRCh38
rs569423985 Chr.17: 5210972-5210972 on Build GRCh38
rs781174325 Chr.17: 5210897-5210897 on Build GRCh38
rs748934299 Chr.17: 5210927-5210927 on Build GRCh38
rs769853168 Chr.17: 5210999-5210999 on Build GRCh38
rs779009502 Chr.17: 5210891-5210891 on Build GRCh38
rs531695180 Chr.17: 5210629-5210629 on Build GRCh38
rs986085926 Chr.17: 5210591-5210591 on Build GRCh38
rs769523111 Chr.17: 5210902-5210902 on Build GRCh38
rs374828071 Chr.17: 5210843-5210843 on Build GRCh38
rs932000017 Chr.17: 5210954-5210954 on Build GRCh38
rs527963667 Chr.17: 5210696-5210696 on Build GRCh38
rs374444572 Chr.17: 5210926-5210926 on Build GRCh38
rs961840593 Chr.17: 5210572-5210572 on Build GRCh38
rs375621740 Chr.17: 5210777-5210777 on Build GRCh38
rs201961696 Chr.17: 5210842-5210842 on Build GRCh38
rs201163648 Chr.17: 5210824-5210824 on Build GRCh38
rs962089243 Chr.17: 5210766-5210766 on Build GRCh38
rs748157119 Chr.17: 5210864-5210864 on Build GRCh38
rs996115567 Chr.17: 5210673-5210673 on Build GRCh38
rs761184817 Chr.17: 5210869-5210869 on Build GRCh38
rs762609412 Chr.17: 5210827-5210827 on Build GRCh38
rs531628627 Chr.17: 5210736-5210736 on Build GRCh38
rs1054584590 Chr.17: 5210973-5210975 on Build GRCh38
rs781018296 Chr.17: 5210795-5210795 on Build GRCh38
rs750491742 Chr.17: 5210894-5210894 on Build GRCh38
rs772624931 Chr.17: 5210940-5210940 on Build GRCh38
rs754054475 Chr.17: 5210882-5210882 on Build GRCh38
rs979735410 Chr.17: 5210877-5210877 on Build GRCh38
rs764395405 Chr.17: 5210879-5210879 on Build GRCh38
rs973628539 Chr.17: 5210575-5210576 on Build GRCh38
rs183322183 Chr.17: 5210713-5210713 on Build GRCh38
rs1009192718 Chr.17: 5210626-5210626 on Build GRCh38
rs892044308 Chr.17: 5210594-5210594 on Build GRCh38
rs758464510 Chr.17: 5211001-5211001 on Build GRCh38
rs772908985 Chr.17: 5210820-5210820 on Build GRCh38
rs76257348 Chr.17: 5210719-5210719 on Build GRCh38
rs767833712 Chr.17: 5210967-5210967 on Build GRCh38
rs188679694 Chr.17: 5210952-5210952 on Build GRCh38
rs988963346 Chr.17: 5210914-5210914 on Build GRCh38
rs370565939 Chr.17: 5210799-5210799 on Build GRCh38
rs577947011 Chr.17: 5210552-5210552 on Build GRCh38
rs961493446 Chr.17: 5210764-5210764 on Build GRCh38
rs757423379 Chr.17: 5210883-5210883 on Build GRCh38
rs563019820 Chr.17: 5210625-5210625 on Build GRCh38
rs1053256438 Chr.17: 5210568-5210568 on Build GRCh38
rs767954295 Chr.17: 5210865-5210865 on Build GRCh38
rs117955736 Chr.17: 5210630-5210630 on Build GRCh38
rs200826404 Chr.17: 5210928-5210928 on Build GRCh38
rs533393325 Chr.17: 5210750-5210750 on Build GRCh38
rs1003132887 Chr.17: 5210742-5210742 on Build GRCh38
rs1057235 Chr.17: 5210646-5210646 on Build GRCh38
rs764820048 Chr.17: 5210764-5210764 on Build GRCh38

View Full Product Details