Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7173881-7174378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASGR1
Entrez Gene ID
432
Gene Name
asialoglycoprotein receptor 1
Gene Aliases
ASGPR, ASGPR1, CLEC4H1, HL-1
UniGene
Hs.12056
Gene Chromosome Location
Chr.17: 7173431-7179564 on Build GRCh38
Transcripts
NM_001197216.2 NM_001671.4 XM_011523861.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3820424 c.554A>G p.E185G Chr.17: 7174178-7174178 on Build GRCh38
5037203 c.599T>G p.F200C Chr.17: 7174063-7174063 on Build GRCh38
4849151 c.695G>T p.G232V Chr.17: 7173967-7173967 on Build GRCh38
4069162 c.680C>T p.T227M Chr.17: 7173982-7173982 on Build GRCh38
5422665 c.622G>A p.V208M Chr.17: 7174040-7174040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770128000 Chr.17: 7173915-7173915 on Build GRCh38
rs778085799 Chr.17: 7173917-7173917 on Build GRCh38
rs765939894 Chr.17: 7174077-7174077 on Build GRCh38
rs776116098 Chr.17: 7174153-7174153 on Build GRCh38
rs754264930 Chr.17: 7174167-7174167 on Build GRCh38
rs570514081 Chr.17: 7174352-7174352 on Build GRCh38
rs1006002949 Chr.17: 7174113-7174113 on Build GRCh38
rs771437084 Chr.17: 7174152-7174152 on Build GRCh38
rs759635816 Chr.17: 7174322-7174322 on Build GRCh38
rs765494787 Chr.17: 7174232-7174232 on Build GRCh38
rs144251057 Chr.17: 7174273-7174273 on Build GRCh38
rs554737782 Chr.17: 7174022-7174022 on Build GRCh38
rs772195636 Chr.17: 7174072-7174072 on Build GRCh38
rs1020153338 Chr.17: 7174244-7174244 on Build GRCh38
rs531767592 Chr.17: 7173999-7173999 on Build GRCh38
rs1030088013 Chr.17: 7174149-7174149 on Build GRCh38
rs1006033937 Chr.17: 7174318-7174318 on Build GRCh38
rs763364916 Chr.17: 7174256-7174257 on Build GRCh38
rs370567183 Chr.17: 7174341-7174341 on Build GRCh38
rs201068122 Chr.17: 7174121-7174121 on Build GRCh38
rs761325900 Chr.17: 7174225-7174225 on Build GRCh38
rs369316332 Chr.17: 7174266-7174266 on Build GRCh38
rs762258005 Chr.17: 7174229-7174229 on Build GRCh38
rs748385803 Chr.17: 7173912-7173912 on Build GRCh38
rs769527434 Chr.17: 7174055-7174055 on Build GRCh38
rs757977596 Chr.17: 7174275-7174275 on Build GRCh38
rs768195516 Chr.17: 7174073-7174074 on Build GRCh38
rs763318667 Chr.17: 7174313-7174313 on Build GRCh38
rs139511614 Chr.17: 7174268-7174268 on Build GRCh38
rs774881423 Chr.17: 7173973-7173974 on Build GRCh38
rs1046245164 Chr.17: 7174181-7174181 on Build GRCh38
rs773435237 Chr.17: 7174310-7174310 on Build GRCh38
rs760058896 Chr.17: 7174335-7174336 on Build GRCh38
rs745604612 Chr.17: 7174274-7174274 on Build GRCh38
rs764624231 Chr.17: 7174156-7174156 on Build GRCh38
rs748053905 Chr.17: 7174122-7174122 on Build GRCh38
rs778941160 Chr.17: 7174104-7174104 on Build GRCh38
rs957273892 Chr.17: 7173924-7173924 on Build GRCh38
rs373411111 Chr.17: 7174267-7174267 on Build GRCh38
rs200527750 Chr.17: 7174305-7174305 on Build GRCh38
rs769285748 Chr.17: 7174228-7174228 on Build GRCh38
rs771656399 Chr.17: 7174213-7174213 on Build GRCh38
rs764872797 Chr.17: 7173935-7173935 on Build GRCh38
rs768074691 Chr.17: 7174088-7174088 on Build GRCh38
rs150525835 Chr.17: 7174212-7174212 on Build GRCh38
rs768648594 Chr.17: 7174020-7174020 on Build GRCh38
rs984897770 Chr.17: 7174304-7174304 on Build GRCh38
rs777324517 Chr.17: 7174298-7174298 on Build GRCh38
rs766866981 Chr.17: 7174260-7174261 on Build GRCh38
rs747965845 Chr.17: 7174048-7174048 on Build GRCh38
rs141693553 Chr.17: 7174015-7174015 on Build GRCh38
rs751651865 Chr.17: 7174264-7174264 on Build GRCh38
rs776470584 Chr.17: 7173931-7173931 on Build GRCh38
rs767830073 Chr.17: 7173970-7173970 on Build GRCh38
rs765525009 Chr.17: 7174174-7174174 on Build GRCh38
rs749476449 Chr.17: 7173986-7173986 on Build GRCh38
rs975812290 Chr.17: 7173968-7173968 on Build GRCh38
rs778173821 Chr.17: 7173984-7173984 on Build GRCh38
rs368109417 Chr.17: 7173939-7173939 on Build GRCh38
rs752100016 Chr.17: 7174305-7174306 on Build GRCh38
rs776363236 Chr.17: 7174046-7174046 on Build GRCh38
rs777629829 Chr.17: 7174196-7174196 on Build GRCh38
rs749087233 Chr.17: 7174198-7174198 on Build GRCh38
rs757552158 Chr.17: 7174099-7174099 on Build GRCh38
rs761453831 Chr.17: 7173934-7173934 on Build GRCh38
rs773566687 Chr.17: 7174103-7174104 on Build GRCh38
rs755346951 Chr.17: 7173909-7173909 on Build GRCh38
rs757827804 Chr.17: 7173961-7173961 on Build GRCh38
rs766537010 Chr.17: 7174257-7174257 on Build GRCh38
rs545839544 Chr.17: 7174141-7174141 on Build GRCh38
rs140200078 Chr.17: 7173926-7173926 on Build GRCh38
rs753106586 Chr.17: 7174089-7174089 on Build GRCh38
rs766592988 Chr.17: 7174315-7174315 on Build GRCh38
rs914151429 Chr.17: 7174184-7174184 on Build GRCh38
rs763079626 Chr.17: 7174239-7174239 on Build GRCh38
rs757140165 Chr.17: 7174333-7174333 on Build GRCh38
rs540643770 Chr.17: 7174332-7174332 on Build GRCh38
rs756483196 Chr.17: 7173976-7173976 on Build GRCh38
rs201237165 Chr.17: 7174178-7174178 on Build GRCh38
rs748746332 Chr.17: 7174346-7174346 on Build GRCh38
rs753119514 Chr.17: 7173974-7173974 on Build GRCh38
rs984433142 Chr.17: 7174079-7174079 on Build GRCh38
rs372044113 Chr.17: 7174210-7174210 on Build GRCh38
rs759118980 Chr.17: 7174146-7174146 on Build GRCh38
rs760333217 Chr.17: 7173991-7173991 on Build GRCh38
rs746973272 Chr.17: 7174007-7174007 on Build GRCh38
rs779782015 Chr.17: 7174295-7174295 on Build GRCh38
rs755504341 Chr.17: 7173969-7173969 on Build GRCh38
rs752046089 Chr.17: 7173959-7173959 on Build GRCh38
rs745961817 Chr.17: 7174105-7174105 on Build GRCh38
rs757465551 Chr.17: 7174002-7174002 on Build GRCh38
rs750559574 Chr.17: 7174175-7174175 on Build GRCh38
rs979580087 Chr.17: 7174235-7174235 on Build GRCh38
rs202036859 Chr.17: 7174091-7174091 on Build GRCh38
rs1037188062 Chr.17: 7174117-7174117 on Build GRCh38
rs754156915 Chr.17: 7174095-7174095 on Build GRCh38
rs1010115481 Chr.17: 7173925-7173925 on Build GRCh38
rs762198444 Chr.17: 7174172-7174172 on Build GRCh38
rs750202356 Chr.17: 7174337-7174337 on Build GRCh38
rs34632742 Chr.17: 7174336-7174336 on Build GRCh38
rs774516771 Chr.17: 7174319-7174319 on Build GRCh38
rs369527058 Chr.17: 7174185-7174185 on Build GRCh38
rs748998144 Chr.17: 7174128-7174128 on Build GRCh38
rs779023527 Chr.17: 7174005-7174005 on Build GRCh38
rs746546279 Chr.17: 7174296-7174296 on Build GRCh38
rs762471369 Chr.17: 7173951-7173951 on Build GRCh38
rs117542665 Chr.17: 7174323-7174323 on Build GRCh38
rs761235750 Chr.17: 7174154-7174154 on Build GRCh38
rs368755766 Chr.17: 7174353-7174353 on Build GRCh38
rs950228901 Chr.17: 7174340-7174340 on Build GRCh38
rs771238304 Chr.17: 7174357-7174357 on Build GRCh38
rs560189186 Chr.17: 7174123-7174123 on Build GRCh38
rs762561375 Chr.17: 7174061-7174061 on Build GRCh38
rs777539552 Chr.17: 7174126-7174126 on Build GRCh38
rs372327550 Chr.17: 7174086-7174086 on Build GRCh38
rs376738856 Chr.17: 7174120-7174120 on Build GRCh38
rs1054460959 Chr.17: 7174209-7174209 on Build GRCh38
rs367753365 Chr.17: 7174085-7174085 on Build GRCh38
rs775139544 Chr.17: 7174219-7174219 on Build GRCh38
rs867393231 Chr.17: 7173981-7173981 on Build GRCh38
rs113006607 Chr.17: 7174261-7174261 on Build GRCh38
rs768379651 Chr.17: 7173930-7173930 on Build GRCh38
rs779871802 Chr.17: 7174338-7174338 on Build GRCh38
rs372176894 Chr.17: 7174090-7174090 on Build GRCh38
rs771753310 Chr.17: 7173928-7173928 on Build GRCh38
rs544829539 Chr.17: 7173952-7173952 on Build GRCh38
rs532376531 Chr.17: 7174133-7174133 on Build GRCh38
rs780843328 Chr.17: 7174344-7174344 on Build GRCh38
rs775593723 Chr.17: 7173929-7173929 on Build GRCh38
rs753856371 Chr.17: 7174326-7174326 on Build GRCh38
rs770593227 Chr.17: 7174205-7174205 on Build GRCh38

View Full Product Details