Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7219453-7219990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADVL
Entrez Gene ID
37
Gene Name
acyl-CoA dehydrogenase, very long chain
Gene Aliases
ACAD6, LCACD, VLCAD
UniGene
Hs.437178
Gene Chromosome Location
Chr.17: 7217125-7225267 on Build GRCh38
Transcripts
NM_000018.3 NM_001033859.2 NM_001270447.1 NM_001270448.1 XM_006721516.2 XM_011523829.1 XM_011523830.1 NM_001321074.1 NM_001365.4 NR_135527.1 XM_011523698.1 XM_011523699.2

Gene Symbol
DLG4
Entrez Gene ID
1742
Gene Name
discs large MAGUK scaffold protein 4
Gene Aliases
PSD95, SAP-90, SAP90
UniGene
Hs.463928
Gene Chromosome Location
Chr.17: 7189890-7220050 on Build GRCh38
Transcripts
NM_000018.3 NM_001033859.2 NM_001270447.1 NM_001270448.1 XM_006721516.2 XM_011523829.1 XM_011523830.1 NM_001321074.1 NM_001365.4 NR_135527.1 XM_011523698.1 XM_011523699.2

SNP Detail

SNP ID Chromosome Location
rs370487268 Chr.17: 7219830-7219830 on Build GRCh38
rs988173181 Chr.17: 7219825-7219825 on Build GRCh38
rs773972425 Chr.17: 7219680-7219680 on Build GRCh38
rs745801522 Chr.17: 7219965-7219965 on Build GRCh38
rs3835013 Chr.17: 7219925-7219926 on Build GRCh38
rs1032863954 Chr.17: 7219806-7219806 on Build GRCh38
rs531336193 Chr.17: 7219549-7219549 on Build GRCh38
rs1042431830 Chr.17: 7219964-7219964 on Build GRCh38
rs559165545 Chr.17: 7219780-7219780 on Build GRCh38
rs886053371 Chr.17: 7219853-7219853 on Build GRCh38
rs771984326 Chr.17: 7219879-7219879 on Build GRCh38
rs933168254 Chr.17: 7219930-7219930 on Build GRCh38
rs1021216337 Chr.17: 7219586-7219586 on Build GRCh38
rs775169805 Chr.17: 7219548-7219548 on Build GRCh38
rs1042379199 Chr.17: 7219785-7219785 on Build GRCh38
rs796293831 Chr.17: 7219937-7219938 on Build GRCh38
rs956949999 Chr.17: 7219605-7219605 on Build GRCh38
rs77051465 Chr.17: 7219921-7219921 on Build GRCh38
rs921097477 Chr.17: 7219682-7219682 on Build GRCh38
rs79281055 Chr.17: 7219691-7219691 on Build GRCh38
rs761465026 Chr.17: 7219742-7219742 on Build GRCh38
rs767746963 Chr.17: 7219943-7219943 on Build GRCh38
rs756203774 Chr.17: 7219956-7219956 on Build GRCh38
rs773696021 Chr.17: 7219937-7219937 on Build GRCh38
rs950368507 Chr.17: 7219667-7219667 on Build GRCh38
rs752243826 Chr.17: 7219556-7219557 on Build GRCh38
rs762906819 Chr.17: 7219602-7219602 on Build GRCh38
rs969979537 Chr.17: 7219792-7219792 on Build GRCh38
rs945354338 Chr.17: 7219782-7219782 on Build GRCh38
rs773362488 Chr.17: 7219875-7219875 on Build GRCh38
rs975752159 Chr.17: 7219853-7219853 on Build GRCh38
rs965332279 Chr.17: 7219840-7219840 on Build GRCh38
rs1015161160 Chr.17: 7219503-7219503 on Build GRCh38
rs895489463 Chr.17: 7219912-7219912 on Build GRCh38
rs1051165757 Chr.17: 7219767-7219767 on Build GRCh38
rs1013800278 Chr.17: 7219778-7219778 on Build GRCh38
rs550196368 Chr.17: 7219922-7219922 on Build GRCh38
rs756798696 Chr.17: 7219926-7219927 on Build GRCh38
rs6145976 Chr.17: 7219936-7219937 on Build GRCh38
rs956772134 Chr.17: 7219817-7219817 on Build GRCh38
rs564821395 Chr.17: 7219883-7219883 on Build GRCh38
rs941922535 Chr.17: 7219733-7219733 on Build GRCh38
rs1032502314 Chr.17: 7219968-7219968 on Build GRCh38
rs372592554 Chr.17: 7219949-7219949 on Build GRCh38
rs377448912 Chr.17: 7219715-7219715 on Build GRCh38
rs1024625989 Chr.17: 7219642-7219642 on Build GRCh38
rs796568889 Chr.17: 7219858-7219858 on Build GRCh38
rs557698043 Chr.17: 7219585-7219585 on Build GRCh38
rs763375468 Chr.17: 7219938-7219938 on Build GRCh38
rs9915319 Chr.17: 7219739-7219739 on Build GRCh38
rs773007123 Chr.17: 7219812-7219812 on Build GRCh38
rs771198524 Chr.17: 7219913-7219914 on Build GRCh38
rs755170147 Chr.17: 7219815-7219815 on Build GRCh38
rs1033814639 Chr.17: 7219620-7219620 on Build GRCh38
rs748217172 Chr.17: 7219860-7219860 on Build GRCh38
rs775362595 Chr.17: 7219891-7219891 on Build GRCh38
rs1013747831 Chr.17: 7219519-7219519 on Build GRCh38
rs879906860 Chr.17: 7219902-7219902 on Build GRCh38
rs1056775771 Chr.17: 7219890-7219890 on Build GRCh38
rs577538299 Chr.17: 7219630-7219630 on Build GRCh38
rs369086959 Chr.17: 7219912-7219913 on Build GRCh38
rs78514016 Chr.17: 7219875-7219875 on Build GRCh38
rs867758455 Chr.17: 7219937-7219938 on Build GRCh38
rs12951682 Chr.17: 7219641-7219641 on Build GRCh38
rs542756786 Chr.17: 7219748-7219748 on Build GRCh38
rs548553878 Chr.17: 7219966-7219966 on Build GRCh38
rs779056818 Chr.17: 7219963-7219963 on Build GRCh38
rs117559220 Chr.17: 7219913-7219913 on Build GRCh38
rs753389263 Chr.17: 7219922-7219923 on Build GRCh38
rs79809891 Chr.17: 7219637-7219637 on Build GRCh38
rs1001479462 Chr.17: 7219959-7219959 on Build GRCh38
rs921164942 Chr.17: 7219854-7219854 on Build GRCh38
rs766742361 Chr.17: 7219940-7219940 on Build GRCh38
rs750033388 Chr.17: 7219878-7219878 on Build GRCh38
rs866382039 Chr.17: 7219557-7219557 on Build GRCh38
rs751863859 Chr.17: 7219937-7219938 on Build GRCh38
rs1024676782 Chr.17: 7219834-7219834 on Build GRCh38
rs983370772 Chr.17: 7219683-7219684 on Build GRCh38
rs1001027299 Chr.17: 7219801-7219801 on Build GRCh38
rs778969037 Chr.17: 7219871-7219871 on Build GRCh38
rs899922240 Chr.17: 7219534-7219534 on Build GRCh38
rs1003858790 Chr.17: 7219734-7219734 on Build GRCh38
rs886851847 Chr.17: 7219725-7219725 on Build GRCh38
rs755185338 Chr.17: 7219941-7219941 on Build GRCh38
rs895303813 Chr.17: 7219803-7219803 on Build GRCh38
rs746658509 Chr.17: 7219898-7219898 on Build GRCh38
rs1001450141 Chr.17: 7219617-7219617 on Build GRCh38
rs1051549817 Chr.17: 7219559-7219559 on Build GRCh38
rs1024216537 Chr.17: 7219523-7219523 on Build GRCh38
rs986919510 Chr.17: 7219948-7219948 on Build GRCh38
rs1048633979 Chr.17: 7219717-7219717 on Build GRCh38
rs41283403 Chr.17: 7219509-7219509 on Build GRCh38
rs12951648 Chr.17: 7219604-7219604 on Build GRCh38
rs75850352 Chr.17: 7219490-7219490 on Build GRCh38
rs943926768 Chr.17: 7219654-7219654 on Build GRCh38
rs757178968 Chr.17: 7219868-7219868 on Build GRCh38
rs758357222 Chr.17: 7219967-7219967 on Build GRCh38
rs1110994 Chr.17: 7219537-7219537 on Build GRCh38
rs139223575 Chr.17: 7219921-7219922 on Build GRCh38
rs765784191 Chr.17: 7219929-7219929 on Build GRCh38
rs891100872 Chr.17: 7219561-7219561 on Build GRCh38
rs1001101895 Chr.17: 7219797-7219797 on Build GRCh38

View Full Product Details