Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28560633-28561137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRNAU1AP
Entrez Gene ID
54952
Gene Name
tRNA selenocysteine 1 associated protein 1
Gene Aliases
PRO1902, SECP43, TRSPAP1
UniGene
Hs.533626
Gene Chromosome Location
Chr.1: 28553017-28578545 on Build GRCh38
Transcripts
NM_017846.4 NR_003109.1 XM_017001542.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
907988 c.167C>T p.A56V Chr.1: 28560674-28560674 on Build GRCh38
288359 c.221C>A p.T74K Chr.1: 28560728-28560728 on Build GRCh38
1687293 c.208C>T p.L70F Chr.1: 28560715-28560715 on Build GRCh38
6239355 c.225+6G>T p.? Chr.1: 28560738-28560738 on Build GRCh38
7166753 c.143G>A p.C48Y Chr.1: 28560650-28560650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779053306 Chr.1: 28560657-28560657 on Build GRCh38
rs374374301 Chr.1: 28560745-28560745 on Build GRCh38
rs780022102 Chr.1: 28560770-28560770 on Build GRCh38
rs758543320 Chr.1: 28560677-28560677 on Build GRCh38
rs996102421 Chr.1: 28560902-28560902 on Build GRCh38
rs985348505 Chr.1: 28560941-28560941 on Build GRCh38
rs905262301 Chr.1: 28561086-28561086 on Build GRCh38
rs370847791 Chr.1: 28560802-28560802 on Build GRCh38
rs780554877 Chr.1: 28560919-28560919 on Build GRCh38
rs768930687 Chr.1: 28561082-28561082 on Build GRCh38
rs967506855 Chr.1: 28561072-28561072 on Build GRCh38
rs144594170 Chr.1: 28560749-28560749 on Build GRCh38
rs1020570703 Chr.1: 28561037-28561037 on Build GRCh38
rs957267120 Chr.1: 28560807-28560807 on Build GRCh38
rs1001769156 Chr.1: 28560759-28560759 on Build GRCh38
rs775040485 Chr.1: 28560736-28560736 on Build GRCh38
rs751619477 Chr.1: 28560771-28560771 on Build GRCh38
rs570824411 Chr.1: 28560806-28560806 on Build GRCh38
rs368286631 Chr.1: 28560752-28560752 on Build GRCh38
rs921674486 Chr.1: 28560903-28560903 on Build GRCh38
rs909405456 Chr.1: 28561011-28561011 on Build GRCh38
rs750687980 Chr.1: 28560756-28560756 on Build GRCh38
rs751347662 Chr.1: 28560914-28560914 on Build GRCh38
rs771521043 Chr.1: 28560732-28560732 on Build GRCh38
rs757787668 Chr.1: 28560674-28560674 on Build GRCh38
rs769724753 Chr.1: 28560706-28560706 on Build GRCh38
rs749592857 Chr.1: 28561025-28561025 on Build GRCh38
rs1032739136 Chr.1: 28561085-28561085 on Build GRCh38
rs367597232 Chr.1: 28560746-28560746 on Build GRCh38
rs1032876150 Chr.1: 28560788-28560788 on Build GRCh38
rs777460712 Chr.1: 28560713-28560713 on Build GRCh38
rs370368589 Chr.1: 28560728-28560728 on Build GRCh38
rs375366255 Chr.1: 28560763-28560763 on Build GRCh38
rs975764077 Chr.1: 28560895-28560895 on Build GRCh38
rs759256223 Chr.1: 28560730-28560730 on Build GRCh38
rs941204815 Chr.1: 28560852-28560852 on Build GRCh38
rs917885991 Chr.1: 28561027-28561027 on Build GRCh38
rs200127909 Chr.1: 28560773-28560773 on Build GRCh38
rs756011550 Chr.1: 28560772-28560772 on Build GRCh38
rs940899086 Chr.1: 28561014-28561014 on Build GRCh38
rs762226214 Chr.1: 28560750-28560750 on Build GRCh38
rs539800778 Chr.1: 28560862-28560862 on Build GRCh38
rs886932630 Chr.1: 28560846-28560846 on Build GRCh38
rs548347794 Chr.1: 28560688-28560688 on Build GRCh38
rs890487839 Chr.1: 28560959-28560959 on Build GRCh38
rs896504796 Chr.1: 28560702-28560702 on Build GRCh38
rs746346334 Chr.1: 28560855-28560855 on Build GRCh38
rs796310546 Chr.1: 28561015-28561015 on Build GRCh38
rs1029393639 Chr.1: 28560912-28560912 on Build GRCh38
rs899272200 Chr.1: 28560871-28560871 on Build GRCh38
rs568489685 Chr.1: 28560714-28560714 on Build GRCh38
rs750633725 Chr.1: 28560662-28560662 on Build GRCh38
rs201097715 Chr.1: 28560687-28560687 on Build GRCh38
rs1010128233 Chr.1: 28560848-28560848 on Build GRCh38
rs1009564061 Chr.1: 28560964-28560964 on Build GRCh38

View Full Product Details