Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17212345-17212894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLCN
Entrez Gene ID
201163
Gene Name
folliculin
Gene Aliases
BHD, FLCL
UniGene
Hs.31652
Gene Chromosome Location
Chr.17: 17206945-17237188 on Build GRCh38
Transcripts
NM_144997.5 XM_011523714.2 XM_011523716.1 XM_011523718.2 XM_011523719.2 XM_011523721.2 XM_017024304.1 XM_017024305.1 XM_017024306.1 XM_017024307.1 XM_017024309.1

SNP Detail

SNP ID Chromosome Location
rs937173603 Chr.17: 17212704-17212704 on Build GRCh38
rs886052657 Chr.17: 17212768-17212768 on Build GRCh38
rs960040392 Chr.17: 17212768-17212768 on Build GRCh38
rs960030868 Chr.17: 17212523-17212523 on Build GRCh38
rs972285089 Chr.17: 17212619-17212619 on Build GRCh38
rs747663295 Chr.17: 17212421-17212421 on Build GRCh38
rs991538455 Chr.17: 17212537-17212537 on Build GRCh38
rs1042019775 Chr.17: 17212767-17212767 on Build GRCh38
rs929595318 Chr.17: 17212666-17212666 on Build GRCh38
rs886052655 Chr.17: 17212475-17212476 on Build GRCh38
rs895708601 Chr.17: 17212398-17212398 on Build GRCh38
rs750900491 Chr.17: 17212827-17212827 on Build GRCh38
rs1011380772 Chr.17: 17212501-17212501 on Build GRCh38
rs7223831 Chr.17: 17212741-17212741 on Build GRCh38
rs1035720677 Chr.17: 17212522-17212522 on Build GRCh38
rs1021972178 Chr.17: 17212515-17212515 on Build GRCh38
rs1033116770 Chr.17: 17212837-17212837 on Build GRCh38
rs770643935 Chr.17: 17212834-17212834 on Build GRCh38
rs1021480303 Chr.17: 17212740-17212740 on Build GRCh38
rs909597510 Chr.17: 17212701-17212701 on Build GRCh38
rs563644388 Chr.17: 17212610-17212610 on Build GRCh38
rs973574118 Chr.17: 17212568-17212568 on Build GRCh38
rs943645264 Chr.17: 17212655-17212655 on Build GRCh38
rs1039479974 Chr.17: 17212679-17212679 on Build GRCh38
rs963455195 Chr.17: 17212853-17212853 on Build GRCh38
rs748168795 Chr.17: 17212804-17212804 on Build GRCh38
rs541543491 Chr.17: 17212409-17212409 on Build GRCh38
rs139529824 Chr.17: 17212861-17212861 on Build GRCh38
rs919468352 Chr.17: 17212862-17212862 on Build GRCh38
rs991422285 Chr.17: 17212795-17212795 on Build GRCh38
rs1011841230 Chr.17: 17212709-17212709 on Build GRCh38
rs62064407 Chr.17: 17212504-17212504 on Build GRCh38
rs532627636 Chr.17: 17212654-17212654 on Build GRCh38
rs1046751595 Chr.17: 17212415-17212415 on Build GRCh38
rs953574331 Chr.17: 17212539-17212539 on Build GRCh38
rs1038369584 Chr.17: 17212386-17212386 on Build GRCh38
rs967295683 Chr.17: 17212517-17212517 on Build GRCh38
rs771952210 Chr.17: 17212593-17212593 on Build GRCh38
rs866377403 Chr.17: 17212603-17212603 on Build GRCh38
rs886052656 Chr.17: 17212688-17212688 on Build GRCh38
rs566019526 Chr.17: 17212620-17212620 on Build GRCh38
rs879377193 Chr.17: 17212628-17212628 on Build GRCh38
rs199535675 Chr.17: 17212472-17212473 on Build GRCh38
rs977721165 Chr.17: 17212518-17212518 on Build GRCh38
rs903180933 Chr.17: 17212477-17212477 on Build GRCh38
rs898049182 Chr.17: 17212389-17212389 on Build GRCh38
rs141650706 Chr.17: 17212699-17212699 on Build GRCh38
rs867735833 Chr.17: 17212594-17212594 on Build GRCh38
rs397932764 Chr.17: 17212501-17212502 on Build GRCh38
rs1052438501 Chr.17: 17212617-17212617 on Build GRCh38
rs185759963 Chr.17: 17212805-17212805 on Build GRCh38
rs543608114 Chr.17: 17212554-17212554 on Build GRCh38
rs886052654 Chr.17: 17212382-17212382 on Build GRCh38
rs1035535196 Chr.17: 17212750-17212750 on Build GRCh38
rs574733214 Chr.17: 17212540-17212540 on Build GRCh38
rs1008014224 Chr.17: 17212430-17212430 on Build GRCh38
rs1020309073 Chr.17: 17212451-17212451 on Build GRCh38
rs554751440 Chr.17: 17212411-17212411 on Build GRCh38
rs540609895 Chr.17: 17212476-17212476 on Build GRCh38
rs143768653 Chr.17: 17212869-17212869 on Build GRCh38
rs571893996 Chr.17: 17212744-17212744 on Build GRCh38
rs1018033454 Chr.17: 17212452-17212452 on Build GRCh38

View Full Product Details