Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17843831-17844336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOM1L2
Entrez Gene ID
146691
Gene Name
target of myb1 like 2 membrane trafficking protein
Gene Aliases
-
UniGene
Hs.462379
Gene Chromosome Location
Chr.17: 17843508-17972470 on Build GRCh38
Transcripts
NM_001033551.2 NM_001082968.1 NM_001288786.1 NM_001288787.1 NM_001288788.1 NM_001288789.1 XM_005256461.2 XM_005256462.1 XM_005256463.2 XM_005256466.1 XM_011523662.1 XM_017024201.1

SNP Detail

SNP ID Chromosome Location
rs1049803232 Chr.17: 17843957-17843957 on Build GRCh38
rs555754425 Chr.17: 17844279-17844279 on Build GRCh38
rs901223418 Chr.17: 17843938-17843938 on Build GRCh38
rs1031057821 Chr.17: 17843992-17843992 on Build GRCh38
rs192764265 Chr.17: 17843954-17843954 on Build GRCh38
rs1009511143 Chr.17: 17843979-17843979 on Build GRCh38
rs767068906 Chr.17: 17843988-17843988 on Build GRCh38
rs186015839 Chr.17: 17844128-17844128 on Build GRCh38
rs988658614 Chr.17: 17843894-17843894 on Build GRCh38
rs908015628 Chr.17: 17843953-17843953 on Build GRCh38
rs980555541 Chr.17: 17843868-17843868 on Build GRCh38
rs1021986209 Chr.17: 17843903-17843903 on Build GRCh38
rs751043539 Chr.17: 17843891-17843891 on Build GRCh38
rs1012683746 Chr.17: 17843854-17843854 on Build GRCh38
rs892342888 Chr.17: 17844231-17844231 on Build GRCh38
rs114926222 Chr.17: 17844233-17844233 on Build GRCh38
rs559417740 Chr.17: 17844125-17844125 on Build GRCh38
rs936112752 Chr.17: 17844198-17844198 on Build GRCh38
rs1000682342 Chr.17: 17843967-17843967 on Build GRCh38
rs1005734497 Chr.17: 17844299-17844299 on Build GRCh38
rs996469697 Chr.17: 17844265-17844265 on Build GRCh38
rs376201287 Chr.17: 17844259-17844259 on Build GRCh38
rs1050208757 Chr.17: 17844161-17844161 on Build GRCh38
rs1033774235 Chr.17: 17844278-17844278 on Build GRCh38
rs545650271 Chr.17: 17843982-17843982 on Build GRCh38
rs908768960 Chr.17: 17843910-17843910 on Build GRCh38
rs544790896 Chr.17: 17843961-17843961 on Build GRCh38
rs1033395651 Chr.17: 17844012-17844012 on Build GRCh38
rs536279654 Chr.17: 17843878-17843878 on Build GRCh38
rs1045337499 Chr.17: 17844221-17844221 on Build GRCh38
rs115901542 Chr.17: 17843890-17843890 on Build GRCh38
rs920779203 Chr.17: 17844196-17844196 on Build GRCh38
rs926774608 Chr.17: 17843876-17843876 on Build GRCh38
rs892370221 Chr.17: 17843976-17843976 on Build GRCh38
rs565177115 Chr.17: 17844244-17844244 on Build GRCh38
rs747046878 Chr.17: 17844254-17844254 on Build GRCh38
rs532057096 Chr.17: 17844056-17844056 on Build GRCh38
rs116262834 Chr.17: 17843937-17843937 on Build GRCh38
rs970446641 Chr.17: 17843862-17843862 on Build GRCh38
rs111653510 Chr.17: 17844234-17844234 on Build GRCh38
rs190779850 Chr.17: 17844050-17844050 on Build GRCh38
rs1023715319 Chr.17: 17843856-17843856 on Build GRCh38
rs2236513 Chr.17: 17844052-17844052 on Build GRCh38
rs891617002 Chr.17: 17844035-17844035 on Build GRCh38
rs527926503 Chr.17: 17844171-17844171 on Build GRCh38
rs150960424 Chr.17: 17844044-17844044 on Build GRCh38
rs961754329 Chr.17: 17844295-17844295 on Build GRCh38
rs181471332 Chr.17: 17844106-17844106 on Build GRCh38
rs974690689 Chr.17: 17844055-17844055 on Build GRCh38
rs756571338 Chr.17: 17844312-17844312 on Build GRCh38
rs1015757212 Chr.17: 17844300-17844300 on Build GRCh38
rs987208128 Chr.17: 17844114-17844117 on Build GRCh38
rs573646540 Chr.17: 17843857-17843857 on Build GRCh38
rs779274554 Chr.17: 17844085-17844085 on Build GRCh38
rs1037937872 Chr.17: 17843956-17843956 on Build GRCh38
rs940288194 Chr.17: 17843921-17843921 on Build GRCh38
rs892585437 Chr.17: 17844243-17844243 on Build GRCh38
rs79705505 Chr.17: 17844236-17844236 on Build GRCh38
rs933020284 Chr.17: 17843923-17843923 on Build GRCh38
rs35052993 Chr.17: 17843904-17843905 on Build GRCh38
rs968411397 Chr.17: 17843907-17843907 on Build GRCh38
rs185431675 Chr.17: 17843987-17843987 on Build GRCh38
rs1028940653 Chr.17: 17844271-17844271 on Build GRCh38
rs3183702 Chr.17: 17843975-17843975 on Build GRCh38
rs977008602 Chr.17: 17844057-17844057 on Build GRCh38
rs572156157 Chr.17: 17843981-17843981 on Build GRCh38
rs907647510 Chr.17: 17844118-17844118 on Build GRCh38
rs9915248 Chr.17: 17844200-17844200 on Build GRCh38
rs974849093 Chr.17: 17843909-17843909 on Build GRCh38
rs973177748 Chr.17: 17844298-17844298 on Build GRCh38
rs932617616 Chr.17: 17844091-17844091 on Build GRCh38
rs1014672536 Chr.17: 17844291-17844291 on Build GRCh38

View Full Product Details