Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18858030-18858567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPSAP2
Entrez Gene ID
5636
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 2
Gene Aliases
PAP41
UniGene
Hs.632236
Gene Chromosome Location
Chr.17: 18856299-18931287 on Build GRCh38
Transcripts
NM_001243936.1 NM_001243941.1 NM_001243942.1 NM_002767.3 XM_005256725.2 XM_005256726.1 XM_005256729.4 XM_017024864.1 XM_017024865.1 XM_017024866.1 XM_017024867.1 XM_017024869.1 XM_017024870.1 XM_017024871.1 XM_017024872.1 XM_017024873.1 XM_017024874.1 XM_017024876.1

SNP Detail

SNP ID Chromosome Location
rs999262648 Chr.17: 18858181-18858181 on Build GRCh38
rs946139097 Chr.17: 18858153-18858153 on Build GRCh38
rs1027093192 Chr.17: 18858267-18858267 on Build GRCh38
rs1040693604 Chr.17: 18858138-18858138 on Build GRCh38
rs1012606162 Chr.17: 18858371-18858371 on Build GRCh38
rs1013840037 Chr.17: 18858197-18858197 on Build GRCh38
rs969922152 Chr.17: 18858067-18858067 on Build GRCh38
rs9905483 Chr.17: 18858084-18858084 on Build GRCh38
rs535077337 Chr.17: 18858113-18858113 on Build GRCh38
rs545962548 Chr.17: 18858393-18858393 on Build GRCh38
rs11548434 Chr.17: 18858195-18858195 on Build GRCh38
rs59633160 Chr.17: 18858083-18858083 on Build GRCh38
rs1027145075 Chr.17: 18858519-18858519 on Build GRCh38
rs978842816 Chr.17: 18858384-18858384 on Build GRCh38
rs890092882 Chr.17: 18858073-18858073 on Build GRCh38
rs1056937292 Chr.17: 18858271-18858271 on Build GRCh38
rs982824192 Chr.17: 18858304-18858304 on Build GRCh38
rs897591269 Chr.17: 18858250-18858250 on Build GRCh38
rs916794270 Chr.17: 18858064-18858064 on Build GRCh38
rs903680179 Chr.17: 18858400-18858400 on Build GRCh38
rs1052144762 Chr.17: 18858429-18858429 on Build GRCh38
rs951495789 Chr.17: 18858284-18858284 on Build GRCh38
rs951549015 Chr.17: 18858525-18858525 on Build GRCh38
rs900339405 Chr.17: 18858141-18858141 on Build GRCh38
rs1013641921 Chr.17: 18858449-18858449 on Build GRCh38
rs896427865 Chr.17: 18858517-18858517 on Build GRCh38
rs1023908390 Chr.17: 18858483-18858483 on Build GRCh38
rs1043499965 Chr.17: 18858167-18858167 on Build GRCh38
rs28374894 Chr.17: 18858346-18858346 on Build GRCh38
rs8068683 Chr.17: 18858514-18858514 on Build GRCh38
rs28540773 Chr.17: 18858441-18858441 on Build GRCh38
rs867746333 Chr.17: 18858089-18858089 on Build GRCh38
rs1050072694 Chr.17: 18858070-18858070 on Build GRCh38
rs913342450 Chr.17: 18858145-18858145 on Build GRCh38
rs944005831 Chr.17: 18858342-18858342 on Build GRCh38
rs1024273856 Chr.17: 18858213-18858213 on Build GRCh38
rs930396604 Chr.17: 18858481-18858481 on Build GRCh38
rs763101691 Chr.17: 18858471-18858472 on Build GRCh38
rs867473315 Chr.17: 18858406-18858406 on Build GRCh38
rs994570983 Chr.17: 18858254-18858254 on Build GRCh38
rs1030705962 Chr.17: 18858189-18858189 on Build GRCh38
rs912605246 Chr.17: 18858321-18858321 on Build GRCh38
rs1043980764 Chr.17: 18858395-18858395 on Build GRCh38
rs969597315 Chr.17: 18858253-18858253 on Build GRCh38
rs1014664021 Chr.17: 18858082-18858082 on Build GRCh38
rs931777199 Chr.17: 18858160-18858160 on Build GRCh38
rs1036002546 Chr.17: 18858466-18858466 on Build GRCh38
rs934823398 Chr.17: 18858252-18858252 on Build GRCh38
rs1035972725 Chr.17: 18858220-18858220 on Build GRCh38
rs867480456 Chr.17: 18858127-18858127 on Build GRCh38
rs879719536 Chr.17: 18858142-18858142 on Build GRCh38
rs1020719373 Chr.17: 18858546-18858546 on Build GRCh38
rs982834164 Chr.17: 18858535-18858535 on Build GRCh38
rs59889351 Chr.17: 18858112-18858113 on Build GRCh38
rs905466883 Chr.17: 18858499-18858499 on Build GRCh38
rs1015922023 Chr.17: 18858486-18858486 on Build GRCh38
rs921819228 Chr.17: 18858358-18858358 on Build GRCh38
rs888368881 Chr.17: 18858388-18858388 on Build GRCh38
rs1019625909 Chr.17: 18858537-18858537 on Build GRCh38
rs541612534 Chr.17: 18858526-18858526 on Build GRCh38
rs980655199 Chr.17: 18858136-18858136 on Build GRCh38
rs565572248 Chr.17: 18858076-18858076 on Build GRCh38
rs758419160 Chr.17: 18858098-18858099 on Build GRCh38
rs974624675 Chr.17: 18858258-18858258 on Build GRCh38
rs1002122928 Chr.17: 18858268-18858268 on Build GRCh38
rs903627743 Chr.17: 18858169-18858169 on Build GRCh38
rs9905495 Chr.17: 18858106-18858106 on Build GRCh38
rs1044055935 Chr.17: 18858377-18858377 on Build GRCh38
rs979363388 Chr.17: 18858159-18858159 on Build GRCh38

View Full Product Details