Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18928672-18929073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPSAP2
Entrez Gene ID
5636
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 2
Gene Aliases
PAP41
UniGene
Hs.632236
Gene Chromosome Location
Chr.17: 18856299-18931287 on Build GRCh38
Transcripts
NM_001243936.1 NM_001243940.1 NM_001243941.1 NM_001243942.1 NM_002767.3 XM_005256725.2 XM_005256726.1 XM_005256729.4 XM_017024864.1 XM_017024865.1 XM_017024866.1 XM_017024867.1 XM_017024868.1 XM_017024869.1 XM_017024870.1 XM_017024871.1 XM_017024872.1 XM_017024873.1 XM_017024874.1 XM_017024875.1 XM_017024876.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3819002 c.931G>C p.E311Q Chr.17: 18928937-18928937 on Build GRCh38
6756750 c.916_917insC p.R308fs*4 Chr.17: 18928922-18928923 on Build GRCh38
6756751 c.908C>A p.S303Y Chr.17: 18928914-18928914 on Build GRCh38
5644112 c.944T>C p.I315T Chr.17: 18928950-18928950 on Build GRCh38
4966524 c.898G>T p.G300C Chr.17: 18928904-18928904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192961172 Chr.17: 18928732-18928732 on Build GRCh38
rs866854095 Chr.17: 18928713-18928713 on Build GRCh38
rs373764009 Chr.17: 18928826-18928826 on Build GRCh38
rs780158912 Chr.17: 18928761-18928761 on Build GRCh38
rs766956925 Chr.17: 18928736-18928736 on Build GRCh38
rs550128304 Chr.17: 18929046-18929046 on Build GRCh38
rs753529551 Chr.17: 18928692-18928692 on Build GRCh38
rs772145230 Chr.17: 18928750-18928750 on Build GRCh38
rs765955896 Chr.17: 18928887-18928887 on Build GRCh38
rs763926308 Chr.17: 18928785-18928785 on Build GRCh38
rs148635892 Chr.17: 18928909-18928909 on Build GRCh38
rs775441214 Chr.17: 18928752-18928752 on Build GRCh38
rs188247143 Chr.17: 18928728-18928728 on Build GRCh38
rs528134793 Chr.17: 18928735-18928736 on Build GRCh38
rs757865754 Chr.17: 18928717-18928717 on Build GRCh38
rs760575856 Chr.17: 18928784-18928784 on Build GRCh38
rs777264608 Chr.17: 18928984-18928985 on Build GRCh38
rs764862299 Chr.17: 18928715-18928715 on Build GRCh38
rs115108920 Chr.17: 18928716-18928716 on Build GRCh38
rs547809682 Chr.17: 18928691-18928691 on Build GRCh38
rs561030100 Chr.17: 18928972-18928972 on Build GRCh38
rs569964249 Chr.17: 18929049-18929049 on Build GRCh38
rs916526376 Chr.17: 18928994-18928994 on Build GRCh38
rs772179015 Chr.17: 18928949-18928949 on Build GRCh38
rs768578695 Chr.17: 18928967-18928967 on Build GRCh38
rs779029267 Chr.17: 18928931-18928931 on Build GRCh38
rs748625282 Chr.17: 18928725-18928725 on Build GRCh38
rs745951274 Chr.17: 18928744-18928744 on Build GRCh38
rs533582887 Chr.17: 18929040-18929040 on Build GRCh38
rs916890298 Chr.17: 18929043-18929044 on Build GRCh38
rs774568371 Chr.17: 18928738-18928738 on Build GRCh38
rs529900273 Chr.17: 18928983-18928983 on Build GRCh38
rs367595675 Chr.17: 18928828-18928828 on Build GRCh38
rs752190173 Chr.17: 18928763-18928764 on Build GRCh38
rs947949686 Chr.17: 18928995-18928995 on Build GRCh38
rs973936358 Chr.17: 18929009-18929009 on Build GRCh38
rs910109862 Chr.17: 18928820-18928820 on Build GRCh38
rs756839493 Chr.17: 18928709-18928709 on Build GRCh38
rs750042279 Chr.17: 18928873-18928873 on Build GRCh38
rs371840452 Chr.17: 18928932-18928932 on Build GRCh38
rs778345713 Chr.17: 18928924-18928924 on Build GRCh38
rs781770054 Chr.17: 18928724-18928724 on Build GRCh38
rs747203450 Chr.17: 18929042-18929042 on Build GRCh38
rs776399488 Chr.17: 18928808-18928808 on Build GRCh38
rs760490026 Chr.17: 18928690-18928690 on Build GRCh38
rs755836328 Chr.17: 18928780-18928781 on Build GRCh38
rs370317871 Chr.17: 18928775-18928775 on Build GRCh38
rs769590983 Chr.17: 18928985-18928985 on Build GRCh38
rs771051238 Chr.17: 18928736-18928736 on Build GRCh38
rs747008799 Chr.17: 18928965-18928965 on Build GRCh38
rs149229926 Chr.17: 18928837-18928837 on Build GRCh38
rs200655672 Chr.17: 18928964-18928964 on Build GRCh38
rs977910346 Chr.17: 18928899-18928899 on Build GRCh38
rs772779345 Chr.17: 18928990-18928990 on Build GRCh38
rs984349088 Chr.17: 18928789-18928789 on Build GRCh38
rs757877327 Chr.17: 18928878-18928878 on Build GRCh38
rs749576809 Chr.17: 18928735-18928735 on Build GRCh38
rs749621665 Chr.17: 18928929-18928929 on Build GRCh38

View Full Product Details