Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28341119-28341624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFAIP1
Entrez Gene ID
7126
Gene Name
TNF alpha induced protein 1
Gene Aliases
B12, B61, BTBD34, EDP1, hBACURD2
UniGene
Hs.76090
Gene Chromosome Location
Chr.17: 28335522-28347009 on Build GRCh38
Transcripts
NM_021137.4 XM_017024993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6230419 c.518+10G>A p.? Chr.17: 28341466-28341466 on Build GRCh38
6858860 c.465+10delC p.? Chr.17: 28341336-28341336 on Build GRCh38
6055749 c.381G>A p.K127K Chr.17: 28341242-28341242 on Build GRCh38
1381632 c.469G>A p.V157M Chr.17: 28341407-28341407 on Build GRCh38
1288530 c.489C>A p.N163K Chr.17: 28341427-28341427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548727631 Chr.17: 28341532-28341532 on Build GRCh38
rs782401079 Chr.17: 28341297-28341297 on Build GRCh38
rs782321000 Chr.17: 28341439-28341439 on Build GRCh38
rs150081648 Chr.17: 28341406-28341406 on Build GRCh38
rs530156551 Chr.17: 28341513-28341513 on Build GRCh38
rs781920351 Chr.17: 28341428-28341428 on Build GRCh38
rs781914118 Chr.17: 28341283-28341283 on Build GRCh38
rs782324647 Chr.17: 28341282-28341282 on Build GRCh38
rs377497252 Chr.17: 28341213-28341213 on Build GRCh38
rs974731957 Chr.17: 28341159-28341159 on Build GRCh38
rs782007362 Chr.17: 28341461-28341461 on Build GRCh38
rs905515250 Chr.17: 28341591-28341591 on Build GRCh38
rs371221886 Chr.17: 28341424-28341424 on Build GRCh38
rs376818659 Chr.17: 28341503-28341503 on Build GRCh38
rs782669222 Chr.17: 28341394-28341394 on Build GRCh38
rs782594918 Chr.17: 28341498-28341498 on Build GRCh38
rs782046108 Chr.17: 28341475-28341475 on Build GRCh38
rs781879429 Chr.17: 28341481-28341481 on Build GRCh38
rs932537359 Chr.17: 28341203-28341203 on Build GRCh38
rs895665318 Chr.17: 28341565-28341565 on Build GRCh38
rs368681613 Chr.17: 28341466-28341466 on Build GRCh38
rs781805107 Chr.17: 28341243-28341243 on Build GRCh38
rs527546038 Chr.17: 28341182-28341182 on Build GRCh38
rs781835131 Chr.17: 28341354-28341354 on Build GRCh38
rs770222458 Chr.17: 28341328-28341328 on Build GRCh38
rs375542704 Chr.17: 28341527-28341527 on Build GRCh38
rs974848180 Chr.17: 28341491-28341491 on Build GRCh38
rs1044140033 Chr.17: 28341588-28341588 on Build GRCh38
rs949794012 Chr.17: 28341573-28341573 on Build GRCh38
rs782486646 Chr.17: 28341550-28341550 on Build GRCh38
rs2234117 Chr.17: 28341534-28341534 on Build GRCh38
rs905567589 Chr.17: 28341379-28341379 on Build GRCh38
rs782288971 Chr.17: 28341291-28341291 on Build GRCh38
rs782162965 Chr.17: 28341432-28341432 on Build GRCh38
rs1052955818 Chr.17: 28341535-28341535 on Build GRCh38
rs782574676 Chr.17: 28341370-28341370 on Build GRCh38
rs766727864 Chr.17: 28341576-28341576 on Build GRCh38
rs782207773 Chr.17: 28341264-28341264 on Build GRCh38
rs782814763 Chr.17: 28341479-28341479 on Build GRCh38
rs781907314 Chr.17: 28341333-28341333 on Build GRCh38
rs781859032 Chr.17: 28341214-28341214 on Build GRCh38
rs112851427 Chr.17: 28341144-28341144 on Build GRCh38
rs372239760 Chr.17: 28341487-28341487 on Build GRCh38
rs781807638 Chr.17: 28341467-28341467 on Build GRCh38
rs782285126 Chr.17: 28341255-28341255 on Build GRCh38
rs369434450 Chr.17: 28341465-28341465 on Build GRCh38
rs1043022513 Chr.17: 28341205-28341205 on Build GRCh38
rs374538209 Chr.17: 28341528-28341528 on Build GRCh38
rs782122584 Chr.17: 28341311-28341311 on Build GRCh38
rs782729293 Chr.17: 28341231-28341231 on Build GRCh38
rs782169440 Chr.17: 28341380-28341380 on Build GRCh38
rs782404890 Chr.17: 28341454-28341454 on Build GRCh38
rs782745620 Chr.17: 28341316-28341316 on Build GRCh38
rs773583442 Chr.17: 28341451-28341451 on Build GRCh38
rs782502673 Chr.17: 28341258-28341258 on Build GRCh38
rs782706865 Chr.17: 28341330-28341330 on Build GRCh38
rs782188116 Chr.17: 28341180-28341180 on Build GRCh38
rs782007608 Chr.17: 28341304-28341304 on Build GRCh38
rs782758721 Chr.17: 28341350-28341350 on Build GRCh38
rs782105862 Chr.17: 28341444-28341444 on Build GRCh38
rs782476643 Chr.17: 28341495-28341495 on Build GRCh38
rs569039127 Chr.17: 28341502-28341502 on Build GRCh38
rs782539470 Chr.17: 28341349-28341349 on Build GRCh38
rs34565131 Chr.17: 28341587-28341588 on Build GRCh38
rs529904785 Chr.17: 28341230-28341230 on Build GRCh38
rs782258471 Chr.17: 28341401-28341401 on Build GRCh38
rs782145811 Chr.17: 28341186-28341186 on Build GRCh38
rs782623465 Chr.17: 28341263-28341263 on Build GRCh38
rs560108839 Chr.17: 28341151-28341151 on Build GRCh38
rs782441029 Chr.17: 28341245-28341245 on Build GRCh38
rs782272204 Chr.17: 28341205-28341206 on Build GRCh38
rs782686534 Chr.17: 28341249-28341249 on Build GRCh38
rs782537796 Chr.17: 28341389-28341389 on Build GRCh38
rs782641034 Chr.17: 28341488-28341488 on Build GRCh38
rs534456199 Chr.17: 28341597-28341597 on Build GRCh38
rs371779108 Chr.17: 28341355-28341355 on Build GRCh38
rs781848113 Chr.17: 28341492-28341492 on Build GRCh38

View Full Product Details