Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44011870-44012375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PYY
Entrez Gene ID
5697
Gene Name
peptide YY
Gene Aliases
PYY-I, PYY1
UniGene
Hs.169249
Gene Chromosome Location
Chr.17: 43952733-44024260 on Build GRCh38
Transcripts
XM_011525035.1 NM_001304813.1 NM_001304814.1 NM_032376.3 XM_005257737.2 XM_011525346.1 XM_011525347.1 XM_011525349.1 XM_011525351.1 XM_011525353.1

Gene Symbol
TMEM101
Entrez Gene ID
84336
Gene Name
transmembrane protein 101
Gene Aliases
-
UniGene
Hs.514211
Gene Chromosome Location
Chr.17: 44011188-44024260 on Build GRCh38
Transcripts
XM_011525035.1 NM_001304813.1 NM_001304814.1 NM_032376.3 XM_005257737.2 XM_011525346.1 XM_011525347.1 XM_011525349.1 XM_011525351.1 XM_011525353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5364432 c.498G>A p.R166R Chr.17: 44012204-44012204 on Build GRCh38
1267986 c.698T>C p.F233S Chr.17: 44012004-44012004 on Build GRCh38
3717400 c.530A>T p.E177V Chr.17: 44012172-44012172 on Build GRCh38
3701224 c.582T>C p.F194F Chr.17: 44012120-44012120 on Build GRCh38
3517979 c.613C>T p.Q205* Chr.17: 44012089-44012089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761402909 Chr.17: 44012095-44012095 on Build GRCh38
rs559613170 Chr.17: 44012332-44012332 on Build GRCh38
rs945851725 Chr.17: 44012213-44012213 on Build GRCh38
rs147985387 Chr.17: 44012019-44012019 on Build GRCh38
rs775083910 Chr.17: 44012281-44012295 on Build GRCh38
rs761128095 Chr.17: 44011983-44011983 on Build GRCh38
rs773756005 Chr.17: 44012202-44012202 on Build GRCh38
rs749586725 Chr.17: 44012063-44012064 on Build GRCh38
rs757474050 Chr.17: 44012171-44012171 on Build GRCh38
rs770151253 Chr.17: 44012235-44012235 on Build GRCh38
rs535084079 Chr.17: 44012014-44012014 on Build GRCh38
rs775190389 Chr.17: 44011977-44011977 on Build GRCh38
rs556952458 Chr.17: 44012018-44012018 on Build GRCh38
rs751760680 Chr.17: 44012150-44012150 on Build GRCh38
rs373505021 Chr.17: 44012017-44012017 on Build GRCh38
rs781010038 Chr.17: 44012173-44012173 on Build GRCh38
rs777510963 Chr.17: 44011939-44011939 on Build GRCh38
rs775137559 Chr.17: 44012072-44012072 on Build GRCh38
rs764215750 Chr.17: 44012248-44012248 on Build GRCh38
rs201602637 Chr.17: 44011953-44011953 on Build GRCh38
rs764113941 Chr.17: 44012149-44012149 on Build GRCh38
rs141644804 Chr.17: 44012106-44012106 on Build GRCh38
rs746886148 Chr.17: 44011946-44011946 on Build GRCh38
rs138911524 Chr.17: 44012351-44012351 on Build GRCh38
rs772081824 Chr.17: 44012176-44012199 on Build GRCh38
rs1031710009 Chr.17: 44011980-44011980 on Build GRCh38
rs775937879 Chr.17: 44012239-44012239 on Build GRCh38
rs769383566 Chr.17: 44011975-44011975 on Build GRCh38
rs3744422 Chr.17: 44012329-44012329 on Build GRCh38
rs770648264 Chr.17: 44011951-44011951 on Build GRCh38
rs766657064 Chr.17: 44012016-44012016 on Build GRCh38
rs979737149 Chr.17: 44012048-44012048 on Build GRCh38
rs765798208 Chr.17: 44012119-44012119 on Build GRCh38
rs766891277 Chr.17: 44012051-44012051 on Build GRCh38
rs757041057 Chr.17: 44012061-44012061 on Build GRCh38
rs577035736 Chr.17: 44012302-44012302 on Build GRCh38
rs756109414 Chr.17: 44012197-44012197 on Build GRCh38
rs768241435 Chr.17: 44012199-44012199 on Build GRCh38
rs745796955 Chr.17: 44012066-44012066 on Build GRCh38
rs766716635 Chr.17: 44012096-44012096 on Build GRCh38
rs747203975 Chr.17: 44012045-44012045 on Build GRCh38
rs778126583 Chr.17: 44011996-44011996 on Build GRCh38
rs575103631 Chr.17: 44012343-44012343 on Build GRCh38
rs768292806 Chr.17: 44012083-44012083 on Build GRCh38
rs868195475 Chr.17: 44012070-44012070 on Build GRCh38
rs778994182 Chr.17: 44011911-44011911 on Build GRCh38
rs779179435 Chr.17: 44012077-44012078 on Build GRCh38
rs746740266 Chr.17: 44011913-44011913 on Build GRCh38
rs751632248 Chr.17: 44012271-44012271 on Build GRCh38
rs776657338 Chr.17: 44012032-44012032 on Build GRCh38
rs757377702 Chr.17: 44012064-44012064 on Build GRCh38
rs776905114 Chr.17: 44012105-44012105 on Build GRCh38
rs748317469 Chr.17: 44011916-44011916 on Build GRCh38
rs758611877 Chr.17: 44012024-44012024 on Build GRCh38
rs760001507 Chr.17: 44012113-44012113 on Build GRCh38
rs376280871 Chr.17: 44012198-44012198 on Build GRCh38
rs201655817 Chr.17: 44012063-44012063 on Build GRCh38
rs773435165 Chr.17: 44012013-44012013 on Build GRCh38
rs866096663 Chr.17: 44011938-44011938 on Build GRCh38
rs77130005 Chr.17: 44012289-44012289 on Build GRCh38
rs990471163 Chr.17: 44012114-44012114 on Build GRCh38
rs777916551 Chr.17: 44012038-44012038 on Build GRCh38
rs150743650 Chr.17: 44012141-44012141 on Build GRCh38
rs369422095 Chr.17: 44012185-44012185 on Build GRCh38
rs755018786 Chr.17: 44011899-44011899 on Build GRCh38
rs769892318 Chr.17: 44012071-44012071 on Build GRCh38
rs1012568067 Chr.17: 44011948-44011948 on Build GRCh38
rs773985527 Chr.17: 44012091-44012091 on Build GRCh38
rs763477159 Chr.17: 44012242-44012242 on Build GRCh38
rs369622814 Chr.17: 44011997-44011997 on Build GRCh38
rs748866523 Chr.17: 44012079-44012079 on Build GRCh38
rs944716279 Chr.17: 44012335-44012335 on Build GRCh38
rs762663874 Chr.17: 44011989-44011989 on Build GRCh38
rs116587312 Chr.17: 44011961-44011961 on Build GRCh38
rs200746192 Chr.17: 44012144-44012144 on Build GRCh38
rs747809911 Chr.17: 44012205-44012205 on Build GRCh38
rs780668431 Chr.17: 44011896-44011896 on Build GRCh38
rs772325811 Chr.17: 44011921-44011921 on Build GRCh38
rs771658120 Chr.17: 44012206-44012206 on Build GRCh38
rs745746454 Chr.17: 44012118-44012121 on Build GRCh38
rs759948544 Chr.17: 44012225-44012225 on Build GRCh38
rs1012680619 Chr.17: 44012261-44012261 on Build GRCh38
rs777047079 Chr.17: 44012211-44012211 on Build GRCh38
rs780876294 Chr.17: 44012064-44012064 on Build GRCh38
rs938515156 Chr.17: 44012183-44012183 on Build GRCh38
rs754313465 Chr.17: 44011898-44011898 on Build GRCh38
rs752811584 Chr.17: 44012022-44012022 on Build GRCh38

View Full Product Details