Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44316277-44316813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RUNDC3A
Entrez Gene ID
10900
Gene Name
RUN domain containing 3A
Gene Aliases
RAP2IP, RPIP-8, RPIP8
UniGene
Hs.500197
Gene Chromosome Location
Chr.17: 44308559-44318670 on Build GRCh38
Transcripts
NM_001144825.1 NM_001144826.1 NM_006695.4 XM_005256964.3 XM_017024035.1 XM_017024036.1 XM_017024037.1 XM_017024038.1 XM_017024039.1 XM_017024040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3712334 c.1081C>A p.L361I Chr.17: 44316512-44316512 on Build GRCh38
5722628 c.1143G>A p.S381S Chr.17: 44316670-44316670 on Build GRCh38
3518045 c.1003G>A p.E335K Chr.17: 44316434-44316434 on Build GRCh38
4623573 c.1167C>A p.G389G Chr.17: 44316694-44316694 on Build GRCh38
5701434 c.980_981insC p.L329fs*23 Chr.17: 44316411-44316412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749133117 Chr.17: 44316555-44316555 on Build GRCh38
rs765600100 Chr.17: 44316500-44316500 on Build GRCh38
rs990694117 Chr.17: 44316470-44316470 on Build GRCh38
rs939200381 Chr.17: 44316339-44316339 on Build GRCh38
rs776739126 Chr.17: 44316464-44316464 on Build GRCh38
rs187888156 Chr.17: 44316736-44316736 on Build GRCh38
rs979503843 Chr.17: 44316720-44316720 on Build GRCh38
rs199988752 Chr.17: 44316497-44316497 on Build GRCh38
rs377485717 Chr.17: 44316467-44316467 on Build GRCh38
rs34004571 Chr.17: 44316782-44316782 on Build GRCh38
rs767007093 Chr.17: 44316493-44316493 on Build GRCh38
rs1017317302 Chr.17: 44316750-44316750 on Build GRCh38
rs753459866 Chr.17: 44316600-44316600 on Build GRCh38
rs934852731 Chr.17: 44316420-44316420 on Build GRCh38
rs748663000 Chr.17: 44316425-44316425 on Build GRCh38
rs775366864 Chr.17: 44316485-44316485 on Build GRCh38
rs757311105 Chr.17: 44316526-44316526 on Build GRCh38
rs747499593 Chr.17: 44316525-44316525 on Build GRCh38
rs775599925 Chr.17: 44316336-44316336 on Build GRCh38
rs768545722 Chr.17: 44316561-44316561 on Build GRCh38
rs766087214 Chr.17: 44316587-44316587 on Build GRCh38
rs771802762 Chr.17: 44316446-44316446 on Build GRCh38
rs746060248 Chr.17: 44316466-44316466 on Build GRCh38
rs758759015 Chr.17: 44316511-44316511 on Build GRCh38
rs202167609 Chr.17: 44316564-44316564 on Build GRCh38
rs770195573 Chr.17: 44316538-44316538 on Build GRCh38
rs746350288 Chr.17: 44316343-44316345 on Build GRCh38
rs1031667354 Chr.17: 44316445-44316445 on Build GRCh38
rs775847125 Chr.17: 44316543-44316543 on Build GRCh38
rs946368740 Chr.17: 44316507-44316507 on Build GRCh38
rs993992145 Chr.17: 44316569-44316569 on Build GRCh38
rs751084181 Chr.17: 44316371-44316371 on Build GRCh38
rs1011953281 Chr.17: 44316626-44316626 on Build GRCh38
rs922738707 Chr.17: 44316452-44316452 on Build GRCh38
rs763358865 Chr.17: 44316601-44316601 on Build GRCh38
rs772724876 Chr.17: 44316574-44316574 on Build GRCh38
rs774181293 Chr.17: 44316563-44316563 on Build GRCh38
rs1038329894 Chr.17: 44316557-44316557 on Build GRCh38
rs967772007 Chr.17: 44316681-44316681 on Build GRCh38
rs753118081 Chr.17: 44316510-44316510 on Build GRCh38
rs62640053 Chr.17: 44316631-44316631 on Build GRCh38
rs746090324 Chr.17: 44316529-44316529 on Build GRCh38
rs960504863 Chr.17: 44316615-44316615 on Build GRCh38
rs767111929 Chr.17: 44316377-44316377 on Build GRCh38
rs890063863 Chr.17: 44316731-44316731 on Build GRCh38
rs761275987 Chr.17: 44316491-44316491 on Build GRCh38
rs370610838 Chr.17: 44316337-44316337 on Build GRCh38
rs902190906 Chr.17: 44316532-44316532 on Build GRCh38
rs747126693 Chr.17: 44316448-44316448 on Build GRCh38
rs762915955 Chr.17: 44316338-44316338 on Build GRCh38
rs781537555 Chr.17: 44316751-44316751 on Build GRCh38
rs760165342 Chr.17: 44316575-44316575 on Build GRCh38
rs529238243 Chr.17: 44316672-44316672 on Build GRCh38
rs758846620 Chr.17: 44316444-44316444 on Build GRCh38
rs779213235 Chr.17: 44316416-44316416 on Build GRCh38
rs866369860 Chr.17: 44316381-44316381 on Build GRCh38
rs748844830 Chr.17: 44316559-44316559 on Build GRCh38
rs866941152 Chr.17: 44316446-44316447 on Build GRCh38
rs756785212 Chr.17: 44316375-44316375 on Build GRCh38
rs367613256 Chr.17: 44316572-44316572 on Build GRCh38
rs771014523 Chr.17: 44316459-44316459 on Build GRCh38
rs1043351009 Chr.17: 44316508-44316508 on Build GRCh38
rs961407124 Chr.17: 44316776-44316776 on Build GRCh38
rs750077745 Chr.17: 44316495-44316495 on Build GRCh38
rs763707716 Chr.17: 44316355-44316355 on Build GRCh38
rs774311284 Chr.17: 44316489-44316489 on Build GRCh38
rs1028568271 Chr.17: 44316685-44316685 on Build GRCh38
rs374238223 Chr.17: 44316488-44316488 on Build GRCh38
rs374637818 Chr.17: 44316402-44316402 on Build GRCh38
rs779506801 Chr.17: 44316376-44316376 on Build GRCh38
rs762824530 Chr.17: 44316487-44316487 on Build GRCh38
rs778185034 Chr.17: 44316518-44316518 on Build GRCh38
rs926187574 Chr.17: 44316409-44316409 on Build GRCh38
rs781265956 Chr.17: 44316528-44316528 on Build GRCh38
rs916530588 Chr.17: 44316474-44316474 on Build GRCh38
rs755388948 Chr.17: 44316568-44316568 on Build GRCh38

View Full Product Details