Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63314296-63314803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TANC2
Entrez Gene ID
26115
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2
Gene Aliases
ROLSA, rols
UniGene
Hs.410889
Gene Chromosome Location
Chr.17: 62966523-63427706 on Build GRCh38
Transcripts
NM_025185.3 XM_005257203.4 XM_006721806.3 XM_006721807.3 XM_006721810.3 XM_006721811.3 XM_011524597.2 XM_011524598.2 XM_011524599.1 XM_011524600.2 XM_011524601.2 XM_011524602.2 XM_017024429.1 XM_017024430.1 XM_017024431.1 XM_017024432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6323362 c.1016C>A p.S339* Chr.17: 63314466-63314466 on Build GRCh38
6147669 c.967A>G p.I323V Chr.17: 63314417-63314417 on Build GRCh38
2794578 c.1079C>G p.A360G Chr.17: 63314529-63314529 on Build GRCh38
3520596 c.1202C>T p.P401L Chr.17: 63314652-63314652 on Build GRCh38
6195417 c.1193C>G p.S398* Chr.17: 63314643-63314643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75148027 Chr.17: 63314343-63314343 on Build GRCh38
rs200973824 Chr.17: 63314608-63314608 on Build GRCh38
rs765441426 Chr.17: 63314664-63314664 on Build GRCh38
rs777212672 Chr.17: 63314638-63314638 on Build GRCh38
rs367733202 Chr.17: 63314493-63314493 on Build GRCh38
rs896000036 Chr.17: 63314413-63314413 on Build GRCh38
rs769207252 Chr.17: 63314344-63314344 on Build GRCh38
rs749094947 Chr.17: 63314481-63314481 on Build GRCh38
rs372158189 Chr.17: 63314533-63314533 on Build GRCh38
rs772644087 Chr.17: 63314486-63314486 on Build GRCh38
rs773055904 Chr.17: 63314446-63314446 on Build GRCh38
rs753214101 Chr.17: 63314407-63314407 on Build GRCh38
rs755941584 Chr.17: 63314689-63314689 on Build GRCh38
rs773996052 Chr.17: 63314498-63314498 on Build GRCh38
rs777331405 Chr.17: 63314560-63314560 on Build GRCh38
rs766703437 Chr.17: 63314390-63314390 on Build GRCh38
rs150364213 Chr.17: 63314585-63314585 on Build GRCh38
rs756060797 Chr.17: 63314555-63314555 on Build GRCh38
rs778444346 Chr.17: 63314714-63314714 on Build GRCh38
rs373599249 Chr.17: 63314399-63314399 on Build GRCh38
rs756714102 Chr.17: 63314709-63314709 on Build GRCh38
rs769313187 Chr.17: 63314482-63314482 on Build GRCh38
rs752864811 Chr.17: 63314696-63314697 on Build GRCh38
rs767889438 Chr.17: 63314506-63314506 on Build GRCh38
rs774755576 Chr.17: 63314391-63314391 on Build GRCh38
rs895963137 Chr.17: 63314762-63314762 on Build GRCh38
rs757422430 Chr.17: 63314534-63314534 on Build GRCh38
rs759835968 Chr.17: 63314505-63314505 on Build GRCh38
rs748092332 Chr.17: 63314554-63314554 on Build GRCh38
rs867829706 Chr.17: 63314488-63314489 on Build GRCh38
rs191595364 Chr.17: 63314569-63314569 on Build GRCh38
rs764221772 Chr.17: 63314526-63314526 on Build GRCh38
rs756181350 Chr.17: 63314415-63314415 on Build GRCh38
rs781229986 Chr.17: 63314455-63314455 on Build GRCh38
rs764122149 Chr.17: 63314418-63314418 on Build GRCh38
rs776036126 Chr.17: 63314512-63314512 on Build GRCh38
rs757478572 Chr.17: 63314440-63314440 on Build GRCh38
rs775775457 Chr.17: 63314610-63314610 on Build GRCh38
rs754837305 Chr.17: 63314453-63314453 on Build GRCh38
rs904942471 Chr.17: 63314327-63314327 on Build GRCh38
rs777513617 Chr.17: 63314695-63314695 on Build GRCh38
rs36082892 Chr.17: 63314440-63314441 on Build GRCh38
rs565290187 Chr.17: 63314429-63314429 on Build GRCh38
rs145116450 Chr.17: 63314668-63314668 on Build GRCh38
rs761240810 Chr.17: 63314622-63314622 on Build GRCh38
rs937759894 Chr.17: 63314728-63314728 on Build GRCh38
rs750699899 Chr.17: 63314546-63314546 on Build GRCh38
rs765501350 Chr.17: 63314542-63314542 on Build GRCh38
rs760032068 Chr.17: 63314397-63314397 on Build GRCh38
rs770822150 Chr.17: 63314500-63314500 on Build GRCh38
rs762549502 Chr.17: 63314382-63314382 on Build GRCh38
rs746738452 Chr.17: 63314452-63314452 on Build GRCh38
rs184939935 Chr.17: 63314342-63314342 on Build GRCh38
rs770439367 Chr.17: 63314572-63314572 on Build GRCh38
rs754777978 Chr.17: 63314548-63314548 on Build GRCh38
rs774309244 Chr.17: 63314549-63314549 on Build GRCh38
rs575715567 Chr.17: 63314444-63314444 on Build GRCh38
rs964160016 Chr.17: 63314588-63314588 on Build GRCh38
rs745560986 Chr.17: 63314606-63314606 on Build GRCh38
rs369559231 Chr.17: 63314646-63314646 on Build GRCh38
rs779994723 Chr.17: 63314448-63314448 on Build GRCh38
rs979117644 Chr.17: 63314673-63314673 on Build GRCh38
rs772878165 Chr.17: 63314357-63314357 on Build GRCh38
rs372517614 Chr.17: 63314669-63314669 on Build GRCh38
rs188282938 Chr.17: 63314411-63314411 on Build GRCh38
rs770637490 Chr.17: 63314497-63314497 on Build GRCh38
rs367962603 Chr.17: 63314748-63314748 on Build GRCh38
rs753834001 Chr.17: 63314700-63314700 on Build GRCh38
rs750638841 Chr.17: 63314665-63314665 on Build GRCh38
rs768980674 Chr.17: 63314635-63314635 on Build GRCh38
rs887084046 Chr.17: 63314502-63314502 on Build GRCh38
rs761135261 Chr.17: 63314520-63314520 on Build GRCh38

View Full Product Details