Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45081552-45082048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMT1
Entrez Gene ID
4836
Gene Name
N-myristoyltransferase 1
Gene Aliases
NMT
UniGene
Hs.532790
Gene Chromosome Location
Chr.17: 45061291-45109016 on Build GRCh38
Transcripts
NM_021079.4 XM_011524858.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146927 c.138G>C p.L46F Chr.17: 45081650-45081650 on Build GRCh38
5644122 c.186A>G p.K62K Chr.17: 45081698-45081698 on Build GRCh38
3819764 c.177G>T p.K59N Chr.17: 45081689-45081689 on Build GRCh38
5364362 c.207T>C p.S69S Chr.17: 45081719-45081719 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs562367944 Chr.17: 45081951-45081951 on Build GRCh38
rs761306458 Chr.17: 45081747-45081747 on Build GRCh38
rs762000577 Chr.17: 45081974-45081974 on Build GRCh38
rs995007933 Chr.17: 45081973-45081973 on Build GRCh38
rs536053629 Chr.17: 45081610-45081610 on Build GRCh38
rs963579027 Chr.17: 45081972-45081972 on Build GRCh38
rs777218079 Chr.17: 45081771-45081771 on Build GRCh38
rs557630208 Chr.17: 45081641-45081641 on Build GRCh38
rs766623394 Chr.17: 45081613-45081613 on Build GRCh38
rs201559014 Chr.17: 45081703-45081703 on Build GRCh38
rs759619751 Chr.17: 45081626-45081626 on Build GRCh38
rs959132728 Chr.17: 45081992-45081992 on Build GRCh38
rs780748535 Chr.17: 45081673-45081673 on Build GRCh38
rs529187710 Chr.17: 45081815-45081815 on Build GRCh38
rs890433812 Chr.17: 45081885-45081885 on Build GRCh38
rs938548698 Chr.17: 45082009-45082009 on Build GRCh38
rs777402320 Chr.17: 45081653-45081653 on Build GRCh38
rs141656657 Chr.17: 45081713-45081713 on Build GRCh38
rs202032923 Chr.17: 45081589-45081589 on Build GRCh38
rs996603447 Chr.17: 45081830-45081830 on Build GRCh38
rs755738171 Chr.17: 45081768-45081768 on Build GRCh38
rs758247752 Chr.17: 45081648-45081648 on Build GRCh38
rs984466916 Chr.17: 45081913-45081913 on Build GRCh38
rs573160396 Chr.17: 45081776-45081776 on Build GRCh38
rs908964919 Chr.17: 45081944-45081944 on Build GRCh38
rs923780752 Chr.17: 45081764-45081764 on Build GRCh38
rs752296085 Chr.17: 45081758-45081758 on Build GRCh38
rs771565194 Chr.17: 45081757-45081757 on Build GRCh38
rs781069116 Chr.17: 45081602-45081603 on Build GRCh38
rs3087878 Chr.17: 45081693-45081693 on Build GRCh38
rs140455291 Chr.17: 45081660-45081660 on Build GRCh38
rs570730383 Chr.17: 45081855-45081855 on Build GRCh38
rs111289001 Chr.17: 45082004-45082004 on Build GRCh38
rs948533653 Chr.17: 45081959-45081959 on Build GRCh38
rs753958254 Chr.17: 45081615-45081615 on Build GRCh38
rs528723469 Chr.17: 45081927-45081927 on Build GRCh38
rs774451911 Chr.17: 45081676-45081676 on Build GRCh38
rs533000655 Chr.17: 45082011-45082011 on Build GRCh38
rs184508172 Chr.17: 45081734-45081734 on Build GRCh38
rs543985247 Chr.17: 45081870-45081870 on Build GRCh38
rs757177482 Chr.17: 45081661-45081661 on Build GRCh38
rs779784731 Chr.17: 45081705-45081705 on Build GRCh38
rs369434915 Chr.17: 45081722-45081722 on Build GRCh38
rs1005144939 Chr.17: 45081916-45081916 on Build GRCh38
rs757745898 Chr.17: 45081800-45081800 on Build GRCh38
rs752810524 Chr.17: 45081646-45081646 on Build GRCh38
rs375158125 Chr.17: 45081698-45081698 on Build GRCh38
rs754661249 Chr.17: 45081603-45081603 on Build GRCh38
rs189266479 Chr.17: 45081782-45081782 on Build GRCh38
rs928513268 Chr.17: 45082000-45082000 on Build GRCh38
rs561642277 Chr.17: 45081805-45081805 on Build GRCh38
rs753007039 Chr.17: 45081791-45081791 on Build GRCh38
rs745547877 Chr.17: 45081797-45081797 on Build GRCh38

View Full Product Details