Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49160467-49160972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT2
Entrez Gene ID
124872
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 2
Gene Aliases
B4GALT, GALGT2
UniGene
Hs.374679
Gene Chromosome Location
Chr.17: 49120347-49170722 on Build GRCh38
Transcripts
NM_001159387.1 NM_001159388.1 NM_153446.2 XM_011524314.2 XM_017024173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4608673 c.895C>A p.P299T Chr.17: 49160590-49160590 on Build GRCh38
1184331 c.907C>T p.R303C Chr.17: 49160602-49160602 on Build GRCh38
6609388 c.908G>A p.R303H Chr.17: 49160603-49160603 on Build GRCh38
4950294 c.931T>C p.Y311H Chr.17: 49160626-49160626 on Build GRCh38
4067462 c.887C>T p.A296V Chr.17: 49160582-49160582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914792586 Chr.17: 49160746-49160746 on Build GRCh38
rs145173178 Chr.17: 49160498-49160498 on Build GRCh38
rs1054127814 Chr.17: 49160779-49160779 on Build GRCh38
rs900395295 Chr.17: 49160788-49160788 on Build GRCh38
rs756097824 Chr.17: 49160531-49160531 on Build GRCh38
rs796194080 Chr.17: 49160904-49160905 on Build GRCh38
rs560483031 Chr.17: 49160723-49160723 on Build GRCh38
rs1005918872 Chr.17: 49160875-49160875 on Build GRCh38
rs185410355 Chr.17: 49160861-49160861 on Build GRCh38
rs1014610274 Chr.17: 49160841-49160841 on Build GRCh38
rs768970250 Chr.17: 49160647-49160647 on Build GRCh38
rs995971011 Chr.17: 49160800-49160800 on Build GRCh38
rs1008224705 Chr.17: 49160794-49160794 on Build GRCh38
rs8071836 Chr.17: 49160666-49160666 on Build GRCh38
rs555823384 Chr.17: 49160577-49160577 on Build GRCh38
rs373854762 Chr.17: 49160611-49160611 on Build GRCh38
rs775069696 Chr.17: 49160690-49160690 on Build GRCh38
rs200640469 Chr.17: 49160510-49160510 on Build GRCh38
rs993277301 Chr.17: 49160842-49160842 on Build GRCh38
rs759206868 Chr.17: 49160674-49160674 on Build GRCh38
rs770534725 Chr.17: 49160670-49160670 on Build GRCh38
rs781716479 Chr.17: 49160657-49160657 on Build GRCh38
rs764085984 Chr.17: 49160618-49160618 on Build GRCh38
rs540732540 Chr.17: 49160722-49160722 on Build GRCh38
rs542090787 Chr.17: 49160747-49160747 on Build GRCh38
rs889883065 Chr.17: 49160783-49160783 on Build GRCh38
rs189934978 Chr.17: 49160884-49160884 on Build GRCh38
rs771853495 Chr.17: 49160684-49160684 on Build GRCh38
rs915701955 Chr.17: 49160699-49160699 on Build GRCh38
rs527564859 Chr.17: 49160903-49160903 on Build GRCh38
rs368477173 Chr.17: 49160640-49160640 on Build GRCh38
rs200167882 Chr.17: 49160540-49160540 on Build GRCh38
rs754808700 Chr.17: 49160516-49160516 on Build GRCh38
rs141565774 Chr.17: 49160739-49160739 on Build GRCh38
rs762521960 Chr.17: 49160583-49160583 on Build GRCh38
rs1007424768 Chr.17: 49160613-49160613 on Build GRCh38
rs544523119 Chr.17: 49160644-49160644 on Build GRCh38
rs1051513726 Chr.17: 49160806-49160806 on Build GRCh38
rs760502999 Chr.17: 49160692-49160692 on Build GRCh38
rs758092732 Chr.17: 49160790-49160790 on Build GRCh38
rs752642086 Chr.17: 49160530-49160530 on Build GRCh38
rs746054405 Chr.17: 49160632-49160632 on Build GRCh38
rs758644590 Chr.17: 49160639-49160639 on Build GRCh38
rs981610238 Chr.17: 49160714-49160714 on Build GRCh38
rs777654820 Chr.17: 49160533-49160533 on Build GRCh38
rs777393599 Chr.17: 49160813-49160813 on Build GRCh38
rs774125323 Chr.17: 49160590-49160590 on Build GRCh38
rs767470275 Chr.17: 49160527-49160527 on Build GRCh38
rs990963131 Chr.17: 49160707-49160707 on Build GRCh38
rs1004629948 Chr.17: 49160819-49160819 on Build GRCh38
rs148383545 Chr.17: 49160568-49160568 on Build GRCh38
rs984845411 Chr.17: 49160678-49160678 on Build GRCh38
rs752552356 Chr.17: 49160603-49160603 on Build GRCh38
rs550321488 Chr.17: 49160934-49160934 on Build GRCh38
rs757345539 Chr.17: 49160538-49160538 on Build GRCh38
rs62079775 Chr.17: 49160708-49160708 on Build GRCh38
rs949411932 Chr.17: 49160710-49160710 on Build GRCh38
rs767370487 Chr.17: 49160602-49160602 on Build GRCh38
rs753774586 Chr.17: 49160625-49160625 on Build GRCh38
rs368777036 Chr.17: 49160653-49160653 on Build GRCh38
rs772390801 Chr.17: 49160552-49160552 on Build GRCh38
rs747428968 Chr.17: 49160570-49160570 on Build GRCh38
rs867793742 Chr.17: 49160879-49160879 on Build GRCh38
rs1027667722 Chr.17: 49160942-49160942 on Build GRCh38
rs138230946 Chr.17: 49160628-49160628 on Build GRCh38
rs866052976 Chr.17: 49160856-49160856 on Build GRCh38
rs772532114 Chr.17: 49160581-49160581 on Build GRCh38
rs376144943 Chr.17: 49160913-49160913 on Build GRCh38
rs35625808 Chr.17: 49160629-49160629 on Build GRCh38
rs907462471 Chr.17: 49160500-49160500 on Build GRCh38
rs951773005 Chr.17: 49160676-49160676 on Build GRCh38
rs371277157 Chr.17: 49160585-49160585 on Build GRCh38
rs567504906 Chr.17: 49160489-49160489 on Build GRCh38
rs759303018 Chr.17: 49160599-49160599 on Build GRCh38
rs368123425 Chr.17: 49160547-49160547 on Build GRCh38

View Full Product Details