Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49989927-49990421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLX3
Entrez Gene ID
1747
Gene Name
distal-less homeobox 3
Gene Aliases
AI4, TDO
UniGene
Hs.134194
Gene Chromosome Location
Chr.17: 49990005-49995224 on Build GRCh38
Transcripts
NM_005220.2

SNP Detail

SNP ID Chromosome Location
rs979955241 Chr.17: 49990099-49990099 on Build GRCh38
rs898320565 Chr.17: 49990255-49990255 on Build GRCh38
rs990354073 Chr.17: 49990214-49990214 on Build GRCh38
rs752377359 Chr.17: 49990268-49990268 on Build GRCh38
rs924831068 Chr.17: 49990056-49990056 on Build GRCh38
rs544277873 Chr.17: 49990104-49990104 on Build GRCh38
rs894486710 Chr.17: 49990301-49990301 on Build GRCh38
rs546770430 Chr.17: 49990158-49990158 on Build GRCh38
rs1001851721 Chr.17: 49990030-49990030 on Build GRCh38
rs957888800 Chr.17: 49990198-49990198 on Build GRCh38
rs904899838 Chr.17: 49990164-49990164 on Build GRCh38
rs117813438 Chr.17: 49990221-49990221 on Build GRCh38
rs913778726 Chr.17: 49990088-49990088 on Build GRCh38
rs1018108381 Chr.17: 49990235-49990235 on Build GRCh38
rs1034900148 Chr.17: 49990059-49990059 on Build GRCh38
rs1027691001 Chr.17: 49990112-49990112 on Build GRCh38
rs549037087 Chr.17: 49990185-49990185 on Build GRCh38
rs749305695 Chr.17: 49989997-49989997 on Build GRCh38
rs1002282858 Chr.17: 49990168-49990168 on Build GRCh38
rs3803876 Chr.17: 49990075-49990075 on Build GRCh38
rs61087392 Chr.17: 49990348-49990349 on Build GRCh38
rs960576274 Chr.17: 49990068-49990068 on Build GRCh38
rs192378506 Chr.17: 49990245-49990245 on Build GRCh38
rs1013005698 Chr.17: 49990127-49990127 on Build GRCh38
rs1024414003 Chr.17: 49990353-49990353 on Build GRCh38
rs3803879 Chr.17: 49990265-49990265 on Build GRCh38
rs397952563 Chr.17: 49990362-49990363 on Build GRCh38
rs35795438 Chr.17: 49990111-49990112 on Build GRCh38
rs768920190 Chr.17: 49990012-49990012 on Build GRCh38
rs912274099 Chr.17: 49990223-49990223 on Build GRCh38
rs568427405 Chr.17: 49990189-49990189 on Build GRCh38
rs886053128 Chr.17: 49990292-49990293 on Build GRCh38
rs542196522 Chr.17: 49990031-49990031 on Build GRCh38
rs564135525 Chr.17: 49990107-49990107 on Build GRCh38
rs533081185 Chr.17: 49990111-49990111 on Build GRCh38
rs977708696 Chr.17: 49990050-49990050 on Build GRCh38
rs1012891164 Chr.17: 49990337-49990337 on Build GRCh38
rs774623575 Chr.17: 49990013-49990013 on Build GRCh38
rs983716938 Chr.17: 49990288-49990288 on Build GRCh38
rs886053129 Chr.17: 49990312-49990312 on Build GRCh38
rs3803878 Chr.17: 49990217-49990217 on Build GRCh38
rs932281608 Chr.17: 49990220-49990220 on Build GRCh38
rs958805769 Chr.17: 49989968-49989968 on Build GRCh38
rs186772609 Chr.17: 49990170-49990170 on Build GRCh38
rs529185809 Chr.17: 49990182-49990182 on Build GRCh38
rs1027638711 Chr.17: 49989948-49989948 on Build GRCh38
rs530450412 Chr.17: 49990094-49990094 on Build GRCh38
rs3803877 Chr.17: 49990191-49990191 on Build GRCh38
rs538662689 Chr.17: 49990349-49990349 on Build GRCh38
rs868762263 Chr.17: 49990179-49990179 on Build GRCh38
rs1038546193 Chr.17: 49990160-49990160 on Build GRCh38
rs764837417 Chr.17: 49990256-49990256 on Build GRCh38
rs950222807 Chr.17: 49990015-49990016 on Build GRCh38
rs879159872 Chr.17: 49990054-49990054 on Build GRCh38
rs3840066 Chr.17: 49990293-49990293 on Build GRCh38
rs1006360522 Chr.17: 49990144-49990144 on Build GRCh38
rs1047989983 Chr.17: 49990123-49990123 on Build GRCh38
rs886053130 Chr.17: 49990349-49990350 on Build GRCh38
rs887973049 Chr.17: 49990133-49990133 on Build GRCh38
rs879090995 Chr.17: 49990134-49990134 on Build GRCh38
rs1020816555 Chr.17: 49990197-49990197 on Build GRCh38
rs772459781 Chr.17: 49990109-49990109 on Build GRCh38
rs910610518 Chr.17: 49990103-49990103 on Build GRCh38
rs879005386 Chr.17: 49990385-49990385 on Build GRCh38
rs971600195 Chr.17: 49990181-49990181 on Build GRCh38
rs1024359947 Chr.17: 49990163-49990163 on Build GRCh38

View Full Product Details