Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36450140-36450643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSCP1
Entrez Gene ID
127700
Gene Name
organic solute carrier partner 1
Gene Aliases
C1orf102, NOR1
UniGene
Hs.202207
Gene Chromosome Location
Chr.1: 36417906-36450485 on Build GRCh38
Transcripts
NM_145047.4 NM_206837.2 XM_005270461.1 XM_005270462.1 XM_005270463.3 XM_011540680.2 XM_011540681.2 XM_017000303.1 XM_017000304.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
426193 c.97G>C p.D33H Chr.1: 36450273-36450273 on Build GRCh38
4007493 c.101A>C p.K34T Chr.1: 36450269-36450269 on Build GRCh38
6126402 c.40G>A p.G14S Chr.1: 36450330-36450330 on Build GRCh38
3985018 c.48G>A p.E16E Chr.1: 36450322-36450322 on Build GRCh38
3985019 c.48G>A p.E16E Chr.1: 36450322-36450322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11547025 Chr.1: 36450278-36450278 on Build GRCh38
rs770702649 Chr.1: 36450394-36450394 on Build GRCh38
rs1025899827 Chr.1: 36450561-36450561 on Build GRCh38
rs1056050777 Chr.1: 36450170-36450170 on Build GRCh38
rs201037496 Chr.1: 36450398-36450398 on Build GRCh38
rs545503622 Chr.1: 36450491-36450491 on Build GRCh38
rs906258517 Chr.1: 36450535-36450535 on Build GRCh38
rs1031353425 Chr.1: 36450343-36450343 on Build GRCh38
rs543300807 Chr.1: 36450450-36450450 on Build GRCh38
rs894900389 Chr.1: 36450249-36450249 on Build GRCh38
rs767592647 Chr.1: 36450465-36450465 on Build GRCh38
rs748897051 Chr.1: 36450270-36450270 on Build GRCh38
rs755473902 Chr.1: 36450248-36450248 on Build GRCh38
rs750619496 Chr.1: 36450227-36450227 on Build GRCh38
rs959409585 Chr.1: 36450577-36450577 on Build GRCh38
rs34360868 Chr.1: 36450563-36450564 on Build GRCh38
rs922821453 Chr.1: 36450401-36450401 on Build GRCh38
rs561613890 Chr.1: 36450611-36450611 on Build GRCh38
rs200304061 Chr.1: 36450257-36450257 on Build GRCh38
rs997853238 Chr.1: 36450553-36450553 on Build GRCh38
rs761657140 Chr.1: 36450411-36450411 on Build GRCh38
rs760563226 Chr.1: 36450292-36450292 on Build GRCh38
rs986409800 Chr.1: 36450219-36450219 on Build GRCh38
rs957950189 Chr.1: 36450550-36450550 on Build GRCh38
rs200812460 Chr.1: 36450413-36450413 on Build GRCh38
rs755882757 Chr.1: 36450215-36450215 on Build GRCh38
rs942825277 Chr.1: 36450383-36450383 on Build GRCh38
rs1045704297 Chr.1: 36450437-36450437 on Build GRCh38
rs527694642 Chr.1: 36450520-36450520 on Build GRCh38
rs565358283 Chr.1: 36450408-36450408 on Build GRCh38
rs751803676 Chr.1: 36450452-36450452 on Build GRCh38
rs1012027312 Chr.1: 36450489-36450489 on Build GRCh38
rs777436455 Chr.1: 36450222-36450222 on Build GRCh38
rs370263831 Chr.1: 36450251-36450251 on Build GRCh38
rs760639855 Chr.1: 36450406-36450406 on Build GRCh38
rs991650130 Chr.1: 36450589-36450589 on Build GRCh38
rs888679420 Chr.1: 36450197-36450197 on Build GRCh38
rs554453784 Chr.1: 36450443-36450443 on Build GRCh38
rs957195482 Chr.1: 36450173-36450173 on Build GRCh38
rs1030757689 Chr.1: 36450547-36450547 on Build GRCh38
rs747135773 Chr.1: 36450244-36450244 on Build GRCh38
rs778173516 Chr.1: 36450387-36450387 on Build GRCh38
rs950296802 Chr.1: 36450605-36450605 on Build GRCh38
rs560478357 Chr.1: 36450507-36450507 on Build GRCh38
rs773071945 Chr.1: 36450480-36450480 on Build GRCh38
rs76250426 Chr.1: 36450612-36450612 on Build GRCh38
rs536095898 Chr.1: 36450430-36450430 on Build GRCh38
rs758591420 Chr.1: 36450318-36450318 on Build GRCh38
rs781753637 Chr.1: 36450250-36450250 on Build GRCh38
rs201190612 Chr.1: 36450306-36450306 on Build GRCh38
rs781133965 Chr.1: 36450367-36450367 on Build GRCh38
rs150491917 Chr.1: 36450277-36450277 on Build GRCh38
rs976758966 Chr.1: 36450595-36450595 on Build GRCh38
rs578063883 Chr.1: 36450478-36450478 on Build GRCh38
rs1005905013 Chr.1: 36450211-36450211 on Build GRCh38
rs111667086 Chr.1: 36450607-36450607 on Build GRCh38
rs546026706 Chr.1: 36450583-36450583 on Build GRCh38
rs773476229 Chr.1: 36450262-36450262 on Build GRCh38
rs377445734 Chr.1: 36450405-36450405 on Build GRCh38
rs756597040 Chr.1: 36450381-36450381 on Build GRCh38
rs957919095 Chr.1: 36450371-36450371 on Build GRCh38
rs761483264 Chr.1: 36450404-36450405 on Build GRCh38
rs910012720 Chr.1: 36450388-36450388 on Build GRCh38
rs1047745767 Chr.1: 36450164-36450164 on Build GRCh38
rs151311655 Chr.1: 36450271-36450271 on Build GRCh38
rs763901874 Chr.1: 36450302-36450302 on Build GRCh38
rs76502678 Chr.1: 36450564-36450564 on Build GRCh38
rs556601250 Chr.1: 36450471-36450471 on Build GRCh38
rs752428300 Chr.1: 36450207-36450207 on Build GRCh38
rs758642416 Chr.1: 36450240-36450240 on Build GRCh38
rs903193404 Chr.1: 36450462-36450462 on Build GRCh38
rs751627554 Chr.1: 36450334-36450334 on Build GRCh38
rs748646276 Chr.1: 36450375-36450375 on Build GRCh38
rs893258456 Chr.1: 36450499-36450499 on Build GRCh38
rs916080217 Chr.1: 36450592-36450592 on Build GRCh38
rs989446501 Chr.1: 36450373-36450373 on Build GRCh38
rs549842821 Chr.1: 36450206-36450206 on Build GRCh38
rs978058314 Chr.1: 36450326-36450326 on Build GRCh38
rs576178166 Chr.1: 36450447-36450447 on Build GRCh38
rs745521447 Chr.1: 36450404-36450404 on Build GRCh38
rs950388103 Chr.1: 36450563-36450563 on Build GRCh38
rs766658594 Chr.1: 36450324-36450324 on Build GRCh38
rs1018262464 Chr.1: 36450567-36450567 on Build GRCh38
rs765033327 Chr.1: 36450412-36450412 on Build GRCh38
rs999777823 Chr.1: 36450545-36450545 on Build GRCh38
rs202098892 Chr.1: 36450328-36450328 on Build GRCh38
rs911181512 Chr.1: 36450382-36450382 on Build GRCh38
rs922692743 Chr.1: 36450604-36450604 on Build GRCh38
rs371410461 Chr.1: 36450393-36450393 on Build GRCh38
rs754996882 Chr.1: 36450346-36450346 on Build GRCh38
rs908954298 Chr.1: 36450568-36450568 on Build GRCh38
rs950370725 Chr.1: 36450434-36450434 on Build GRCh38

View Full Product Details